Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 75016056-75016561 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CARNMT1
Entrez Gene ID
138199
Gene Name
carnosine N-methyltransferase 1
Gene Aliases
C9orf41, UPF0586
UniGene
Hs.567688
Gene Chromosome Location
Chr.9: 74980794-75028456 on Build GRCh38
Transcripts
NM_001320497.1 NM_152420.2 NR_135282.1 XM_005251701.3 XM_011518223.2 XM_017014269.1 XM_017014270.1 XM_017014271.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
69970 c.479C>T p.S160F Chr.9: 75016379-75016379 on Build GRCh38
1569923 c.572_573insA p.N191fs*11 Chr.9: 75016285-75016286 on Build GRCh38
3413730 c.513A>C p.E171D Chr.9: 75016345-75016345 on Build GRCh38
6350853 c.483G>A p.T161T Chr.9: 75016375-75016375 on Build GRCh38
332521 c.520A>T p.K174* Chr.9: 75016338-75016338 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs746241409 Chr.9: 75016273-75016273 on Build GRCh38
rs755829890 Chr.9: 75016427-75016427 on Build GRCh38
rs753950646 Chr.9: 75016360-75016360 on Build GRCh38
rs767651375 Chr.9: 75016217-75016217 on Build GRCh38
rs941015794 Chr.9: 75016308-75016308 on Build GRCh38
rs375404122 Chr.9: 75016092-75016092 on Build GRCh38
rs922777899 Chr.9: 75016201-75016201 on Build GRCh38
rs745813333 Chr.9: 75016448-75016448 on Build GRCh38
rs758329610 Chr.9: 75016156-75016156 on Build GRCh38
rs760935443 Chr.9: 75016333-75016333 on Build GRCh38
rs1013978154 Chr.9: 75016453-75016453 on Build GRCh38
rs760424605 Chr.9: 75016462-75016462 on Build GRCh38
rs902076285 Chr.9: 75016538-75016548 on Build GRCh38
rs746808873 Chr.9: 75016280-75016280 on Build GRCh38
rs951420820 Chr.9: 75016177-75016177 on Build GRCh38
rs764153468 Chr.9: 75016243-75016243 on Build GRCh38
rs756370570 Chr.9: 75016432-75016433 on Build GRCh38
rs759295740 Chr.9: 75016313-75016313 on Build GRCh38
rs771021329 Chr.9: 75016301-75016301 on Build GRCh38
rs1043682312 Chr.9: 75016474-75016474 on Build GRCh38
rs866741210 Chr.9: 75016189-75016189 on Build GRCh38
rs779354198 Chr.9: 75016270-75016270 on Build GRCh38
rs751816424 Chr.9: 75016408-75016408 on Build GRCh38
rs778755398 Chr.9: 75016434-75016434 on Build GRCh38
rs372633105 Chr.9: 75016376-75016376 on Build GRCh38
rs576698033 Chr.9: 75016160-75016160 on Build GRCh38
rs747836388 Chr.9: 75016286-75016286 on Build GRCh38
rs368573143 Chr.9: 75016523-75016523 on Build GRCh38
rs367763445 Chr.9: 75016361-75016361 on Build GRCh38
rs992884647 Chr.9: 75016162-75016162 on Build GRCh38
rs1002438283 Chr.9: 75016143-75016143 on Build GRCh38
rs757768005 Chr.9: 75016262-75016262 on Build GRCh38
rs758768242 Chr.9: 75016279-75016279 on Build GRCh38
rs998722922 Chr.9: 75016521-75016521 on Build GRCh38
rs777524962 Chr.9: 75016428-75016428 on Build GRCh38
rs34980997 Chr.9: 75016188-75016188 on Build GRCh38
rs372689805 Chr.9: 75016483-75016483 on Build GRCh38
rs377279854 Chr.9: 75016402-75016402 on Build GRCh38
rs945035710 Chr.9: 75016437-75016438 on Build GRCh38
rs186297202 Chr.9: 75016153-75016153 on Build GRCh38
rs775640456 Chr.9: 75016328-75016328 on Build GRCh38
rs910118837 Chr.9: 75016206-75016206 on Build GRCh38
rs748737629 Chr.9: 75016299-75016299 on Build GRCh38
rs1024320545 Chr.9: 75016167-75016167 on Build GRCh38
rs1047742167 Chr.9: 75016242-75016242 on Build GRCh38
rs1033342229 Chr.9: 75016530-75016530 on Build GRCh38
rs371188840 Chr.9: 75016232-75016232 on Build GRCh38
rs748825598 Chr.9: 75016429-75016429 on Build GRCh38
rs573849024 Chr.9: 75016140-75016140 on Build GRCh38
rs752941642 Chr.9: 75016220-75016220 on Build GRCh38
rs536493008 Chr.9: 75016148-75016148 on Build GRCh38
rs903756332 Chr.9: 75016482-75016482 on Build GRCh38
rs529898007 Chr.9: 75016494-75016494 on Build GRCh38
rs776812081 Chr.9: 75016481-75016481 on Build GRCh38
rs894189232 Chr.9: 75016419-75016419 on Build GRCh38
rs541087984 Chr.9: 75016334-75016334 on Build GRCh38
rs572086678 Chr.9: 75016291-75016291 on Build GRCh38
rs911303817 Chr.9: 75016133-75016133 on Build GRCh38
rs771819384 Chr.9: 75016450-75016450 on Build GRCh38
rs369519571 Chr.9: 75016466-75016466 on Build GRCh38
rs1034457531 Chr.9: 75016149-75016149 on Build GRCh38
rs750376185 Chr.9: 75016080-75016080 on Build GRCh38
rs375058342 Chr.9: 75016234-75016234 on Build GRCh38
rs774291008 Chr.9: 75016309-75016309 on Build GRCh38
rs780455946 Chr.9: 75016405-75016405 on Build GRCh38
rs767894750 Chr.9: 75016349-75016349 on Build GRCh38
rs767311544 Chr.9: 75016316-75016316 on Build GRCh38
rs145889052 Chr.9: 75016375-75016375 on Build GRCh38
rs962941520 Chr.9: 75016208-75016208 on Build GRCh38
rs544969137 Chr.9: 75016176-75016176 on Build GRCh38
rs770539113 Chr.9: 75016433-75016433 on Build GRCh38
rs903002589 Chr.9: 75016502-75016502 on Build GRCh38
rs754398588 Chr.9: 75016248-75016248 on Build GRCh38
rs1022591760 Chr.9: 75016125-75016125 on Build GRCh38
rs930186016 Chr.9: 75016228-75016228 on Build GRCh38
rs750625178 Chr.9: 75016242-75016243 on Build GRCh38
rs780751743 Chr.9: 75016281-75016281 on Build GRCh38

View Full Product Details