Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 137050816-137051318 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ENTPD2
Entrez Gene ID
954
Gene Name
ectonucleoside triphosphate diphosphohydrolase 2
Gene Aliases
CD39L1, NTPDase-2
UniGene
Hs.123036
Gene Chromosome Location
Chr.9: 137048098-137054051 on Build GRCh38
Transcripts
NM_001246.3 NM_203468.2 XM_011519212.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
239709 c.716_724delGCTTCCTCT p.F240_C242delFLC Chr.9: 137050952-137050960 on Build GRCh38
1739559 c.733C>T p.R245C Chr.9: 137050943-137050943 on Build GRCh38
422431 c.747C>G p.L249L Chr.9: 137050929-137050929 on Build GRCh38
5962500 c.730G>C p.G244R Chr.9: 137050946-137050946 on Build GRCh38
422430 c.707A>G p.Y236C Chr.9: 137050969-137050969 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs772794241 Chr.9: 137051264-137051264 on Build GRCh38
rs139027787 Chr.9: 137051284-137051284 on Build GRCh38
rs116635795 Chr.9: 137050962-137050962 on Build GRCh38
rs774636661 Chr.9: 137051024-137051024 on Build GRCh38
rs760409236 Chr.9: 137051270-137051270 on Build GRCh38
rs879376155 Chr.9: 137050888-137050888 on Build GRCh38
rs768876583 Chr.9: 137050954-137050954 on Build GRCh38
rs368559678 Chr.9: 137050863-137050863 on Build GRCh38
rs756649518 Chr.9: 137050902-137050902 on Build GRCh38
rs750747096 Chr.9: 137050901-137050901 on Build GRCh38
rs757106831 Chr.9: 137051010-137051010 on Build GRCh38
rs369887698 Chr.9: 137051139-137051139 on Build GRCh38
rs975901403 Chr.9: 137051156-137051156 on Build GRCh38
rs753620776 Chr.9: 137051208-137051208 on Build GRCh38
rs780800770 Chr.9: 137051011-137051011 on Build GRCh38
rs776007084 Chr.9: 137051117-137051117 on Build GRCh38
rs563908073 Chr.9: 137051099-137051099 on Build GRCh38
rs770836776 Chr.9: 137050861-137050861 on Build GRCh38
rs777913963 Chr.9: 137051155-137051155 on Build GRCh38
rs145171719 Chr.9: 137051127-137051127 on Build GRCh38
rs763472190 Chr.9: 137051078-137051078 on Build GRCh38
rs753341928 Chr.9: 137050908-137050908 on Build GRCh38
rs938101304 Chr.9: 137051026-137051026 on Build GRCh38
rs377578336 Chr.9: 137051182-137051182 on Build GRCh38
rs965890788 Chr.9: 137050960-137050960 on Build GRCh38
rs571941272 Chr.9: 137051067-137051067 on Build GRCh38
rs201122803 Chr.9: 137050989-137050989 on Build GRCh38
rs555069562 Chr.9: 137051259-137051259 on Build GRCh38
rs767060838 Chr.9: 137051132-137051132 on Build GRCh38
rs201629581 Chr.9: 137051119-137051119 on Build GRCh38
rs749636417 Chr.9: 137051242-137051242 on Build GRCh38
rs765730832 Chr.9: 137051073-137051073 on Build GRCh38
rs748652176 Chr.9: 137051258-137051258 on Build GRCh38
rs76730730 Chr.9: 137051225-137051225 on Build GRCh38
rs756798666 Chr.9: 137051081-137051081 on Build GRCh38
rs763674017 Chr.9: 137051124-137051124 on Build GRCh38
rs199705392 Chr.9: 137050968-137050968 on Build GRCh38
rs778704168 Chr.9: 137051220-137051220 on Build GRCh38
rs770820178 Chr.9: 137050938-137050938 on Build GRCh38
rs757870499 Chr.9: 137051075-137051075 on Build GRCh38
rs751245390 Chr.9: 137051080-137051080 on Build GRCh38
rs528726375 Chr.9: 137051167-137051167 on Build GRCh38
rs889427359 Chr.9: 137051077-137051077 on Build GRCh38
rs780458762 Chr.9: 137051238-137051238 on Build GRCh38
rs761409849 Chr.9: 137050915-137050915 on Build GRCh38
rs776802560 Chr.9: 137051068-137051068 on Build GRCh38
rs768004591 Chr.9: 137050898-137050898 on Build GRCh38
rs35983961 Chr.9: 137051098-137051098 on Build GRCh38
rs756175007 Chr.9: 137051088-137051088 on Build GRCh38
rs763064182 Chr.9: 137050877-137050877 on Build GRCh38
rs768216778 Chr.9: 137051275-137051275 on Build GRCh38
rs141638137 Chr.9: 137051103-137051103 on Build GRCh38
rs766126391 Chr.9: 137051272-137051272 on Build GRCh38
rs200918154 Chr.9: 137051293-137051293 on Build GRCh38
rs201716888 Chr.9: 137051164-137051164 on Build GRCh38
rs780600887 Chr.9: 137050905-137050905 on Build GRCh38
rs145240352 Chr.9: 137050998-137050998 on Build GRCh38
rs535221430 Chr.9: 137050890-137050890 on Build GRCh38
rs777380815 Chr.9: 137051005-137051005 on Build GRCh38
rs771657320 Chr.9: 137050922-137050922 on Build GRCh38
rs1055203543 Chr.9: 137051044-137051044 on Build GRCh38
rs752251479 Chr.9: 137051074-137051074 on Build GRCh38
rs141307913 Chr.9: 137050988-137050988 on Build GRCh38
rs371276900 Chr.9: 137051105-137051105 on Build GRCh38
rs761081418 Chr.9: 137050977-137050977 on Build GRCh38
rs781198847 Chr.9: 137051229-137051229 on Build GRCh38
rs772473354 Chr.9: 137051034-137051034 on Build GRCh38
rs773510818 Chr.9: 137050976-137050976 on Build GRCh38
rs755010690 Chr.9: 137051217-137051217 on Build GRCh38
rs745643137 Chr.9: 137051086-137051086 on Build GRCh38
rs369416410 Chr.9: 137051115-137051116 on Build GRCh38
rs759351054 Chr.9: 137050943-137050943 on Build GRCh38
rs767845036 Chr.9: 137050975-137050975 on Build GRCh38
rs200191389 Chr.9: 137051032-137051032 on Build GRCh38
rs760772555 Chr.9: 137051052-137051052 on Build GRCh38
rs375426284 Chr.9: 137051178-137051178 on Build GRCh38
rs1037175258 Chr.9: 137051186-137051186 on Build GRCh38
rs993138967 Chr.9: 137051106-137051106 on Build GRCh38
rs78069653 Chr.9: 137051179-137051179 on Build GRCh38
rs760544909 Chr.9: 137050855-137050855 on Build GRCh38
rs777549961 Chr.9: 137050924-137050924 on Build GRCh38
rs747269047 Chr.9: 137051109-137051109 on Build GRCh38
rs760042607 Chr.9: 137051069-137051069 on Build GRCh38
rs993142652 Chr.9: 137050847-137050847 on Build GRCh38
rs769190505 Chr.9: 137050982-137050982 on Build GRCh38
rs767588454 Chr.9: 137051290-137051290 on Build GRCh38
rs367870548 Chr.9: 137051009-137051009 on Build GRCh38
rs762486822 Chr.9: 137051233-137051234 on Build GRCh38
rs143374800 Chr.9: 137051277-137051277 on Build GRCh38
rs750133912 Chr.9: 137051185-137051185 on Build GRCh38
rs868337912 Chr.9: 137051065-137051065 on Build GRCh38
rs372911345 Chr.9: 137051170-137051170 on Build GRCh38
rs770323046 Chr.9: 137051114-137051114 on Build GRCh38
rs199604605 Chr.9: 137051177-137051177 on Build GRCh38
rs760358331 Chr.9: 137051201-137051201 on Build GRCh38
rs937986692 Chr.9: 137051171-137051171 on Build GRCh38
rs773456383 Chr.9: 137051049-137051049 on Build GRCh38
rs779959577 Chr.9: 137051145-137051145 on Build GRCh38
rs769524705 Chr.9: 137051203-137051203 on Build GRCh38
rs754814569 Chr.9: 137051150-137051150 on Build GRCh38
rs769936130 Chr.9: 137051234-137051234 on Build GRCh38
rs775570161 Chr.9: 137050910-137050910 on Build GRCh38
rs774142681 Chr.9: 137051246-137051247 on Build GRCh38
rs557629801 Chr.9: 137051223-137051223 on Build GRCh38
rs896680985 Chr.9: 137051070-137051070 on Build GRCh38
rs2292925 Chr.9: 137050911-137050911 on Build GRCh38
rs753853340 Chr.9: 137051148-137051148 on Build GRCh38
rs780942303 Chr.9: 137051064-137051065 on Build GRCh38
rs559166791 Chr.9: 137051165-137051165 on Build GRCh38
rs201198065 Chr.9: 137051244-137051244 on Build GRCh38
rs781245560 Chr.9: 137050975-137050976 on Build GRCh38
rs756382655 Chr.9: 137051294-137051294 on Build GRCh38
rs781040746 Chr.9: 137051083-137051083 on Build GRCh38
rs766294964 Chr.9: 137051205-137051205 on Build GRCh38
rs752597686 Chr.9: 137051285-137051285 on Build GRCh38
rs541605947 Chr.9: 137051000-137051000 on Build GRCh38
rs373580613 Chr.9: 137051181-137051181 on Build GRCh38
rs374065956 Chr.9: 137050942-137050942 on Build GRCh38
rs774691572 Chr.9: 137051252-137051252 on Build GRCh38
rs768782675 Chr.9: 137051238-137051245 on Build GRCh38
rs747820624 Chr.9: 137050914-137050914 on Build GRCh38
rs746804655 Chr.9: 137050932-137050932 on Build GRCh38
rs764450531 Chr.9: 137050884-137050884 on Build GRCh38
rs138022111 Chr.9: 137051113-137051113 on Build GRCh38
rs746103337 Chr.9: 137051233-137051233 on Build GRCh38
rs770993547 Chr.9: 137051111-137051111 on Build GRCh38
rs769785647 Chr.9: 137051021-137051021 on Build GRCh38
rs182051175 Chr.9: 137051137-137051137 on Build GRCh38
rs566162999 Chr.9: 137051166-137051166 on Build GRCh38
rs552032181 Chr.9: 137051217-137051219 on Build GRCh38

View Full Product Details