Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 137244555-137245048 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM166A
Entrez Gene ID
401565
Gene Name
family with sequence similarity 166 member A
Gene Aliases
HSD46
UniGene
Hs.522530
Gene Chromosome Location
Chr.9: 137243583-137251193 on Build GRCh38
Transcripts
NM_001001710.2 XM_006717108.2 XM_011518694.1 XM_011518695.1 XM_011518696.2 XM_011518697.1 XM_017014718.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7291898 c.747+10C>T p.? Chr.9: 137244632-137244632 on Build GRCh38
6217175 c.666C>A p.F222L Chr.9: 137244723-137244723 on Build GRCh38
6007569 c.691G>T p.G231W Chr.9: 137244698-137244698 on Build GRCh38
5475269 c.724G>A p.E242K Chr.9: 137244907-137244907 on Build GRCh38
3848114 c.772G>A p.A258T Chr.9: 137244859-137244859 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs749911273 Chr.9: 137244674-137244674 on Build GRCh38
rs774799739 Chr.9: 137244898-137244898 on Build GRCh38
rs780288828 Chr.9: 137244804-137244804 on Build GRCh38
rs746501786 Chr.9: 137244649-137244670 on Build GRCh38
rs762029125 Chr.9: 137244989-137244989 on Build GRCh38
rs769293357 Chr.9: 137244948-137244948 on Build GRCh38
rs370530610 Chr.9: 137244649-137244649 on Build GRCh38
rs371393584 Chr.9: 137244730-137244730 on Build GRCh38
rs746077864 Chr.9: 137244755-137244755 on Build GRCh38
rs764226015 Chr.9: 137244978-137244978 on Build GRCh38
rs925212529 Chr.9: 137244903-137244903 on Build GRCh38
rs762563818 Chr.9: 137244952-137244952 on Build GRCh38
rs759608224 Chr.9: 137244700-137244700 on Build GRCh38
rs569822523 Chr.9: 137244842-137244842 on Build GRCh38
rs769505038 Chr.9: 137244896-137244896 on Build GRCh38
rs778836216 Chr.9: 137244683-137244683 on Build GRCh38
rs762118119 Chr.9: 137244727-137244727 on Build GRCh38
rs755260501 Chr.9: 137244873-137244873 on Build GRCh38
rs771397399 Chr.9: 137244892-137244892 on Build GRCh38
rs115662231 Chr.9: 137244672-137244672 on Build GRCh38
rs138556037 Chr.9: 137244859-137244859 on Build GRCh38
rs758586501 Chr.9: 137244972-137244972 on Build GRCh38
rs771956234 Chr.9: 137245020-137245020 on Build GRCh38
rs766142646 Chr.9: 137244773-137244773 on Build GRCh38
rs777352483 Chr.9: 137244609-137244609 on Build GRCh38
rs777762789 Chr.9: 137244932-137244932 on Build GRCh38
rs779200322 Chr.9: 137244925-137244925 on Build GRCh38
rs781014744 Chr.9: 137244866-137244866 on Build GRCh38
rs765326421 Chr.9: 137244967-137244967 on Build GRCh38
rs760577423 Chr.9: 137245023-137245023 on Build GRCh38
rs757068865 Chr.9: 137244860-137244860 on Build GRCh38
rs746837435 Chr.9: 137244792-137244792 on Build GRCh38
rs746392437 Chr.9: 137244745-137244745 on Build GRCh38
rs533681403 Chr.9: 137244680-137244680 on Build GRCh38
rs773693344 Chr.9: 137244768-137244768 on Build GRCh38
rs576410794 Chr.9: 137244578-137244578 on Build GRCh38
rs143899198 Chr.9: 137244726-137244726 on Build GRCh38
rs749032276 Chr.9: 137244640-137244640 on Build GRCh38
rs544084778 Chr.9: 137244938-137244938 on Build GRCh38
rs142358325 Chr.9: 137244686-137244686 on Build GRCh38
rs768183011 Chr.9: 137244955-137244955 on Build GRCh38
rs771039724 Chr.9: 137244799-137244809 on Build GRCh38
rs760547937 Chr.9: 137244965-137244965 on Build GRCh38
rs769654267 Chr.9: 137244888-137244914 on Build GRCh38
rs953990320 Chr.9: 137244999-137244999 on Build GRCh38
rs764601226 Chr.9: 137244724-137244724 on Build GRCh38
rs778373649 Chr.9: 137244880-137244880 on Build GRCh38
rs111918484 Chr.9: 137244812-137244812 on Build GRCh38
rs1016253793 Chr.9: 137244714-137244714 on Build GRCh38
rs745753262 Chr.9: 137244862-137244865 on Build GRCh38
rs150138458 Chr.9: 137244740-137244740 on Build GRCh38
rs773794006 Chr.9: 137245012-137245012 on Build GRCh38
rs141610152 Chr.9: 137244951-137244951 on Build GRCh38
rs1037261178 Chr.9: 137244772-137244772 on Build GRCh38
rs776441745 Chr.9: 137244843-137244843 on Build GRCh38
rs181951804 Chr.9: 137244907-137244907 on Build GRCh38
rs1001945062 Chr.9: 137244930-137244930 on Build GRCh38
rs758524481 Chr.9: 137244931-137244931 on Build GRCh38
rs769735726 Chr.9: 137244708-137244708 on Build GRCh38
rs561414478 Chr.9: 137245014-137245014 on Build GRCh38
rs779706317 Chr.9: 137244735-137244735 on Build GRCh38
rs896604914 Chr.9: 137244816-137244816 on Build GRCh38
rs1015852019 Chr.9: 137244632-137244632 on Build GRCh38
rs898676185 Chr.9: 137244795-137244795 on Build GRCh38
rs781152822 Chr.9: 137244840-137244841 on Build GRCh38
rs141597818 Chr.9: 137244665-137244665 on Build GRCh38
rs920339044 Chr.9: 137244648-137244648 on Build GRCh38
rs777422431 Chr.9: 137244791-137244791 on Build GRCh38
rs968295040 Chr.9: 137244854-137244854 on Build GRCh38
rs949413166 Chr.9: 137244827-137244827 on Build GRCh38
rs770241333 Chr.9: 137244811-137244811 on Build GRCh38
rs748967239 Chr.9: 137245011-137245011 on Build GRCh38
rs781773389 Chr.9: 137244984-137244984 on Build GRCh38
rs746188806 Chr.9: 137244989-137244989 on Build GRCh38
rs150244600 Chr.9: 137244653-137244653 on Build GRCh38
rs949523161 Chr.9: 137244936-137244936 on Build GRCh38
rs946529257 Chr.9: 137244725-137244725 on Build GRCh38
rs374721889 Chr.9: 137244589-137244589 on Build GRCh38
rs773153741 Chr.9: 137245021-137245021 on Build GRCh38
rs201286537 Chr.9: 137244715-137244715 on Build GRCh38
rs62584882 Chr.9: 137244654-137244654 on Build GRCh38
rs768557893 Chr.9: 137244643-137244643 on Build GRCh38
rs752677469 Chr.9: 137244929-137244929 on Build GRCh38
rs765687644 Chr.9: 137244599-137244599 on Build GRCh38
rs757774935 Chr.9: 137244982-137244982 on Build GRCh38
rs986260555 Chr.9: 137244738-137244738 on Build GRCh38
rs567844089 Chr.9: 137244733-137244733 on Build GRCh38
rs760230102 Chr.9: 137244769-137244769 on Build GRCh38
rs772692588 Chr.9: 137244817-137244817 on Build GRCh38
rs767150154 Chr.9: 137244728-137244728 on Build GRCh38
rs370092815 Chr.9: 137244716-137244716 on Build GRCh38
rs912615818 Chr.9: 137245017-137245017 on Build GRCh38
rs115339115 Chr.9: 137244681-137244681 on Build GRCh38
rs529469092 Chr.9: 137244957-137244957 on Build GRCh38
rs750276561 Chr.9: 137244910-137244910 on Build GRCh38
rs1027821158 Chr.9: 137244798-137244798 on Build GRCh38
rs765214832 Chr.9: 137244785-137244785 on Build GRCh38
rs764804250 Chr.9: 137244848-137244848 on Build GRCh38
rs758397494 Chr.9: 137244789-137244789 on Build GRCh38
rs1051784913 Chr.9: 137244610-137244610 on Build GRCh38
rs764441736 Chr.9: 137244664-137244664 on Build GRCh38
rs368100373 Chr.9: 137244677-137244677 on Build GRCh38
rs995653054 Chr.9: 137244795-137244795 on Build GRCh38
rs555243390 Chr.9: 137244659-137244659 on Build GRCh38
rs777490014 Chr.9: 137244597-137244598 on Build GRCh38
rs772279098 Chr.9: 137245010-137245012 on Build GRCh38
rs775090209 Chr.9: 137244956-137244957 on Build GRCh38
rs913719934 Chr.9: 137244716-137244716 on Build GRCh38
rs746243971 Chr.9: 137244940-137244940 on Build GRCh38
rs751743409 Chr.9: 137244979-137244979 on Build GRCh38
rs772391357 Chr.9: 137244879-137244879 on Build GRCh38
rs758147160 Chr.9: 137244750-137244750 on Build GRCh38
rs372288312 Chr.9: 137244753-137244753 on Build GRCh38
rs753023802 Chr.9: 137244600-137244600 on Build GRCh38
rs368482857 Chr.9: 137244742-137244742 on Build GRCh38
rs781645362 Chr.9: 137244939-137244939 on Build GRCh38
rs371296055 Chr.9: 137244591-137244591 on Build GRCh38
rs768817013 Chr.9: 137244908-137244908 on Build GRCh38
rs748272201 Chr.9: 137244878-137244878 on Build GRCh38
rs1019809981 Chr.9: 137244968-137244968 on Build GRCh38
rs750151670 Chr.9: 137244959-137244959 on Build GRCh38
rs747325528 Chr.9: 137244890-137244890 on Build GRCh38
rs754601959 Chr.9: 137244660-137244660 on Build GRCh38
rs542889685 Chr.9: 137245007-137245007 on Build GRCh38
rs375089903 Chr.9: 137244756-137244756 on Build GRCh38
rs890075144 Chr.9: 137244962-137244962 on Build GRCh38
rs776747004 Chr.9: 137244698-137244698 on Build GRCh38
rs774862230 Chr.9: 137244762-137244762 on Build GRCh38
rs1002143217 Chr.9: 137244594-137244594 on Build GRCh38
rs117505852 Chr.9: 137244913-137244913 on Build GRCh38
rs141120386 Chr.9: 137244646-137244646 on Build GRCh38
rs542316575 Chr.9: 137244612-137244612 on Build GRCh38
rs754028808 Chr.9: 137244868-137244868 on Build GRCh38
rs766018341 Chr.9: 137244651-137244651 on Build GRCh38
rs752550919 Chr.9: 137244786-137244786 on Build GRCh38
rs897133646 Chr.9: 137244950-137244950 on Build GRCh38
rs4073074 Chr.9: 137244732-137244732 on Build GRCh38
rs769729242 Chr.9: 137244623-137244623 on Build GRCh38
rs541401756 Chr.9: 137244902-137244902 on Build GRCh38
rs749377056 Chr.9: 137244757-137244757 on Build GRCh38
rs1017762277 Chr.9: 137244864-137244864 on Build GRCh38
rs547007247 Chr.9: 137244780-137244780 on Build GRCh38
rs572606971 Chr.9: 137244895-137244895 on Build GRCh38
rs774354877 Chr.9: 137244644-137244644 on Build GRCh38
rs759307299 Chr.9: 137244846-137244846 on Build GRCh38
rs769114207 Chr.9: 137244759-137244759 on Build GRCh38
rs375483164 Chr.9: 137244607-137244607 on Build GRCh38
rs780715028 Chr.9: 137244673-137244673 on Build GRCh38
rs747906919 Chr.9: 137244685-137244685 on Build GRCh38
rs767743227 Chr.9: 137244764-137244764 on Build GRCh38
rs200646970 Chr.9: 137244621-137244621 on Build GRCh38
rs777173061 Chr.9: 137244894-137244894 on Build GRCh38
rs762432218 Chr.9: 137244763-137244763 on Build GRCh38
rs373939553 Chr.9: 137244661-137244661 on Build GRCh38
rs752939435 Chr.9: 137244970-137244970 on Build GRCh38
rs749305754 Chr.9: 137244832-137244832 on Build GRCh38
rs778845227 Chr.9: 137244877-137244877 on Build GRCh38
rs780324533 Chr.9: 137245008-137245008 on Build GRCh38
rs570387652 Chr.9: 137244809-137244809 on Build GRCh38
rs374564504 Chr.9: 137244837-137244837 on Build GRCh38
rs535334326 Chr.9: 137244853-137244853 on Build GRCh38
rs760309367 Chr.9: 137244631-137244631 on Build GRCh38
rs753330552 Chr.9: 137244774-137244774 on Build GRCh38
rs754903051 Chr.9: 137244741-137244741 on Build GRCh38
rs777669248 Chr.9: 137244754-137244754 on Build GRCh38
rs35907539 Chr.9: 137244736-137244736 on Build GRCh38
rs756266027 Chr.9: 137244800-137244800 on Build GRCh38

View Full Product Details