Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 70199732-70200237 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DGAT2L6
Entrez Gene ID
347516
Gene Name
diacylglycerol O-acyltransferase 2 like 6
Gene Aliases
DC3
UniGene
Hs.454355
Gene Chromosome Location
Chr.X: 70177483-70205703 on Build GRCh38
Transcripts
NM_198512.2

SNP Detail

SNP ID Chromosome Location
rs376345103 Chr.X: 70199843-70199843 on Build GRCh38
rs369983719 Chr.X: 70199899-70199899 on Build GRCh38
rs199964484 Chr.X: 70199820-70199820 on Build GRCh38
rs182094563 Chr.X: 70199978-70199978 on Build GRCh38
rs1040364172 Chr.X: 70199937-70199937 on Build GRCh38
rs891570849 Chr.X: 70200162-70200162 on Build GRCh38
rs199868415 Chr.X: 70199858-70199858 on Build GRCh38
rs1013899633 Chr.X: 70200163-70200163 on Build GRCh38
rs905565868 Chr.X: 70200195-70200195 on Build GRCh38
rs757866664 Chr.X: 70200091-70200091 on Build GRCh38
rs774585003 Chr.X: 70199835-70199835 on Build GRCh38
rs775563554 Chr.X: 70199897-70199897 on Build GRCh38
rs745993159 Chr.X: 70199777-70199777 on Build GRCh38
rs750491748 Chr.X: 70199890-70199891 on Build GRCh38
rs999702928 Chr.X: 70200099-70200099 on Build GRCh38
rs767903713 Chr.X: 70200212-70200212 on Build GRCh38
rs756504906 Chr.X: 70200015-70200015 on Build GRCh38
rs756263037 Chr.X: 70199925-70199925 on Build GRCh38
rs1052815940 Chr.X: 70200100-70200100 on Build GRCh38
rs779675665 Chr.X: 70200098-70200098 on Build GRCh38
rs776212908 Chr.X: 70200174-70200174 on Build GRCh38
rs368777263 Chr.X: 70199992-70199992 on Build GRCh38
rs746564718 Chr.X: 70200128-70200128 on Build GRCh38
rs749703157 Chr.X: 70200068-70200068 on Build GRCh38
rs763293794 Chr.X: 70199821-70199821 on Build GRCh38
rs757989917 Chr.X: 70199867-70199867 on Build GRCh38
rs777581571 Chr.X: 70199883-70199883 on Build GRCh38
rs902439669 Chr.X: 70200056-70200056 on Build GRCh38
rs780215628 Chr.X: 70200212-70200212 on Build GRCh38
rs932075593 Chr.X: 70199975-70199975 on Build GRCh38
rs373796174 Chr.X: 70199887-70199887 on Build GRCh38
rs750739912 Chr.X: 70200149-70200149 on Build GRCh38
rs200516088 Chr.X: 70199898-70199898 on Build GRCh38
rs962580410 Chr.X: 70199935-70199935 on Build GRCh38
rs1001849196 Chr.X: 70200202-70200202 on Build GRCh38
rs745947183 Chr.X: 70199888-70199888 on Build GRCh38
rs752077822 Chr.X: 70199993-70199993 on Build GRCh38
rs753377531 Chr.X: 70199849-70199849 on Build GRCh38
rs868565368 Chr.X: 70199877-70199877 on Build GRCh38
rs1044023375 Chr.X: 70199979-70199979 on Build GRCh38
rs140805163 Chr.X: 70199863-70199863 on Build GRCh38
rs112865288 Chr.X: 70199890-70199890 on Build GRCh38
rs764465072 Chr.X: 70199824-70199824 on Build GRCh38
rs776719642 Chr.X: 70199762-70199762 on Build GRCh38
rs767778314 Chr.X: 70199962-70199962 on Build GRCh38
rs769976796 Chr.X: 70199797-70199797 on Build GRCh38
rs762343876 Chr.X: 70199836-70199836 on Build GRCh38
rs1024205820 Chr.X: 70199756-70199756 on Build GRCh38
rs186969997 Chr.X: 70200058-70200058 on Build GRCh38
rs1040892623 Chr.X: 70199861-70199861 on Build GRCh38
rs935219193 Chr.X: 70200087-70200087 on Build GRCh38
rs944683788 Chr.X: 70199934-70199934 on Build GRCh38
rs906722604 Chr.X: 70200201-70200201 on Build GRCh38
rs774731510 Chr.X: 70199902-70199902 on Build GRCh38
rs768407034 Chr.X: 70200171-70200172 on Build GRCh38
rs369660367 Chr.X: 70199884-70199884 on Build GRCh38
rs1041390191 Chr.X: 70200053-70200053 on Build GRCh38

View Full Product Details