Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 11326971-11327471 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GALNT18
Entrez Gene ID
374378
Gene Name
polypeptide N-acetylgalactosaminyltransferase 18
Gene Aliases
GALNACT18, GALNT15, GALNTL4, GalNAc-T15, GalNAc-T18
UniGene
Hs.655152
Gene Chromosome Location
Chr.11: 11270874-11622045 on Build GRCh38
Transcripts
NM_198516.2 XM_006718224.2 XM_011520068.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3443561 c.1482C>T p.I494I Chr.11: 11327116-11327116 on Build GRCh38
686331 c.1489A>G p.I497V Chr.11: 11327109-11327109 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs901524785 Chr.11: 11327160-11327160 on Build GRCh38
rs1044319340 Chr.11: 11327115-11327115 on Build GRCh38
rs747107371 Chr.11: 11327052-11327052 on Build GRCh38
rs745624376 Chr.11: 11327178-11327178 on Build GRCh38
rs866769216 Chr.11: 11327324-11327324 on Build GRCh38
rs369750746 Chr.11: 11327068-11327068 on Build GRCh38
rs180864514 Chr.11: 11327204-11327204 on Build GRCh38
rs755562293 Chr.11: 11327158-11327158 on Build GRCh38
rs946987932 Chr.11: 11327114-11327114 on Build GRCh38
rs754708808 Chr.11: 11327139-11327139 on Build GRCh38
rs1023714254 Chr.11: 11327436-11327436 on Build GRCh38
rs200806645 Chr.11: 11327056-11327056 on Build GRCh38
rs929853537 Chr.11: 11327315-11327315 on Build GRCh38
rs184968469 Chr.11: 11327259-11327259 on Build GRCh38
rs1025975657 Chr.11: 11327193-11327193 on Build GRCh38
rs974041696 Chr.11: 11327285-11327285 on Build GRCh38
rs892890512 Chr.11: 11327346-11327346 on Build GRCh38
rs929039131 Chr.11: 11327432-11327432 on Build GRCh38
rs757231037 Chr.11: 11327093-11327093 on Build GRCh38
rs1035029166 Chr.11: 11327314-11327314 on Build GRCh38
rs545918654 Chr.11: 11327429-11327429 on Build GRCh38
rs866944679 Chr.11: 11327169-11327169 on Build GRCh38
rs754618893 Chr.11: 11327054-11327054 on Build GRCh38
rs35895990 Chr.11: 11327264-11327265 on Build GRCh38
rs756008280 Chr.11: 11327210-11327210 on Build GRCh38
rs745181 Chr.11: 11327281-11327281 on Build GRCh38
rs574438446 Chr.11: 11327351-11327351 on Build GRCh38
rs770704827 Chr.11: 11327092-11327092 on Build GRCh38
rs772140469 Chr.11: 11327105-11327105 on Build GRCh38
rs752299713 Chr.11: 11327157-11327157 on Build GRCh38
rs879513700 Chr.11: 11327235-11327235 on Build GRCh38
rs901629567 Chr.11: 11327431-11327431 on Build GRCh38
rs576992821 Chr.11: 11327426-11327426 on Build GRCh38
rs942404317 Chr.11: 11327101-11327101 on Build GRCh38
rs896674394 Chr.11: 11327412-11327412 on Build GRCh38
rs762431580 Chr.11: 11327228-11327228 on Build GRCh38
rs777830967 Chr.11: 11327164-11327164 on Build GRCh38
rs115791155 Chr.11: 11327132-11327132 on Build GRCh38
rs75793419 Chr.11: 11327247-11327247 on Build GRCh38
rs80045991 Chr.11: 11327311-11327311 on Build GRCh38
rs1051339106 Chr.11: 11327095-11327095 on Build GRCh38
rs188596818 Chr.11: 11327278-11327278 on Build GRCh38
rs531898533 Chr.11: 11327110-11327110 on Build GRCh38
rs116436785 Chr.11: 11327174-11327174 on Build GRCh38
rs760643052 Chr.11: 11327109-11327109 on Build GRCh38
rs962729177 Chr.11: 11327437-11327437 on Build GRCh38
rs866373578 Chr.11: 11327064-11327064 on Build GRCh38
rs776786478 Chr.11: 11327215-11327215 on Build GRCh38
rs973989324 Chr.11: 11327024-11327024 on Build GRCh38
rs749136483 Chr.11: 11327078-11327078 on Build GRCh38
rs750767311 Chr.11: 11327130-11327130 on Build GRCh38
rs7110342 Chr.11: 11327390-11327390 on Build GRCh38
rs17435705 Chr.11: 11327122-11327122 on Build GRCh38
rs558443515 Chr.11: 11327280-11327280 on Build GRCh38
rs1056699249 Chr.11: 11327402-11327402 on Build GRCh38
rs943491506 Chr.11: 11327403-11327403 on Build GRCh38
rs767044253 Chr.11: 11327146-11327146 on Build GRCh38
rs373282244 Chr.11: 11327076-11327076 on Build GRCh38
rs1026613977 Chr.11: 11327019-11327019 on Build GRCh38
rs778706000 Chr.11: 11327176-11327176 on Build GRCh38
rs35924905 Chr.11: 11327377-11327378 on Build GRCh38
rs986670643 Chr.11: 11327320-11327320 on Build GRCh38
rs775554401 Chr.11: 11327201-11327201 on Build GRCh38
rs942499125 Chr.11: 11327330-11327330 on Build GRCh38
rs1039508755 Chr.11: 11327345-11327345 on Build GRCh38
rs527964041 Chr.11: 11327128-11327128 on Build GRCh38
rs770385467 Chr.11: 11327232-11327232 on Build GRCh38
rs116157475 Chr.11: 11327003-11327003 on Build GRCh38
rs985067375 Chr.11: 11327049-11327049 on Build GRCh38
rs1041744798 Chr.11: 11327357-11327357 on Build GRCh38
rs747569937 Chr.11: 11327061-11327061 on Build GRCh38
rs775672652 Chr.11: 11327107-11327107 on Build GRCh38
rs779503493 Chr.11: 11327213-11327214 on Build GRCh38
rs887723889 Chr.11: 11327233-11327233 on Build GRCh38
rs144034068 Chr.11: 11327113-11327113 on Build GRCh38
rs768614555 Chr.11: 11327214-11327214 on Build GRCh38
rs756115965 Chr.11: 11327073-11327073 on Build GRCh38
rs941345488 Chr.11: 11327023-11327023 on Build GRCh38
rs554894051 Chr.11: 11327326-11327326 on Build GRCh38
rs537570994 Chr.11: 11327099-11327099 on Build GRCh38
rs372280659 Chr.11: 11327179-11327179 on Build GRCh38
rs146669960 Chr.11: 11327414-11327414 on Build GRCh38
rs1000160194 Chr.11: 11327392-11327392 on Build GRCh38

View Full Product Details