Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 27654805-27655320 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BDNF
Entrez Gene ID
627
Gene Name
brain derived neurotrophic factor
Gene Aliases
ANON2, BULN2
UniGene
Hs.502182
Gene Chromosome Location
Chr.11: 27654893-27722058 on Build GRCh38
Transcripts
NM_001143805.1 NM_001143806.1 NM_001143807.1 NM_001143808.1 NM_001143809.1 NM_001143810.1 NM_001143811.1 NM_001143812.1 NM_001143813.1 NM_001143814.1 NM_001143816.1 NM_001709.4 NM_170731.4 NM_170732.4 NM_170733.3 NM_170734.3 NM_170735.5 XM_011520280.2 NR_002832.2 NR_033312.1 NR_033313.1 NR_033314.1 NR_033315.1

Gene Symbol
BDNF-AS
Entrez Gene ID
497258
Gene Name
BDNF antisense RNA
Gene Aliases
ANTI-BDNF, BDNF, BDNF-AS1, BDNFOS, NCRNA00049
UniGene
Hs.675323
Gene Chromosome Location
Chr.11: 27506852-27698171 on Build GRCh38
Transcripts
NM_001143805.1 NM_001143806.1 NM_001143807.1 NM_001143808.1 NM_001143809.1 NM_001143810.1 NM_001143811.1 NM_001143812.1 NM_001143813.1 NM_001143814.1 NM_001143816.1 NM_001709.4 NM_170731.4 NM_170732.4 NM_170733.3 NM_170734.3 NM_170735.5 XM_011520280.2 NR_002832.2 NR_033312.1 NR_033313.1 NR_033314.1 NR_033315.1

SNP Detail

SNP ID Chromosome Location
rs959833998 Chr.11: 27654874-27654874 on Build GRCh38
rs1015831484 Chr.11: 27654867-27654867 on Build GRCh38
rs571512915 Chr.11: 27655009-27655009 on Build GRCh38
rs368036351 Chr.11: 27655262-27655262 on Build GRCh38
rs930942895 Chr.11: 27655168-27655168 on Build GRCh38
rs979164730 Chr.11: 27654953-27654953 on Build GRCh38
rs760691328 Chr.11: 27655109-27655109 on Build GRCh38
rs543296443 Chr.11: 27655008-27655008 on Build GRCh38
rs1017152417 Chr.11: 27655077-27655077 on Build GRCh38
rs572229151 Chr.11: 27654948-27654952 on Build GRCh38
rs563108932 Chr.11: 27655027-27655028 on Build GRCh38
rs1022849062 Chr.11: 27654926-27654926 on Build GRCh38
rs547324985 Chr.11: 27655111-27655111 on Build GRCh38
rs1048518594 Chr.11: 27655178-27655178 on Build GRCh38
rs939697536 Chr.11: 27655199-27655199 on Build GRCh38
rs895571052 Chr.11: 27655252-27655252 on Build GRCh38
rs969603027 Chr.11: 27655114-27655114 on Build GRCh38
rs942574013 Chr.11: 27655265-27655265 on Build GRCh38
rs901279557 Chr.11: 27654912-27654912 on Build GRCh38
rs959795947 Chr.11: 27655162-27655162 on Build GRCh38
rs980944675 Chr.11: 27655153-27655153 on Build GRCh38
rs1036762140 Chr.11: 27655229-27655229 on Build GRCh38
rs576161770 Chr.11: 27654994-27654994 on Build GRCh38
rs371776547 Chr.11: 27654982-27654982 on Build GRCh38
rs1039585931 Chr.11: 27655291-27655291 on Build GRCh38
rs538304747 Chr.11: 27655010-27655010 on Build GRCh38
rs905777857 Chr.11: 27655269-27655269 on Build GRCh38
rs992566737 Chr.11: 27655213-27655213 on Build GRCh38
rs1035138256 Chr.11: 27655161-27655161 on Build GRCh38
rs557600370 Chr.11: 27654986-27654986 on Build GRCh38
rs3178739 Chr.11: 27655096-27655096 on Build GRCh38
rs1033053340 Chr.11: 27654978-27654978 on Build GRCh38
rs912892886 Chr.11: 27655222-27655222 on Build GRCh38
rs754338213 Chr.11: 27655261-27655261 on Build GRCh38
rs1006218681 Chr.11: 27655050-27655050 on Build GRCh38
rs191137757 Chr.11: 27655126-27655126 on Build GRCh38
rs888150804 Chr.11: 27655192-27655192 on Build GRCh38
rs80244532 Chr.11: 27655263-27655263 on Build GRCh38
rs780584976 Chr.11: 27654864-27654864 on Build GRCh38

View Full Product Details