Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 39758010-39758509 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IKZF3
Entrez Gene ID
22806
Gene Name
IKAROS family zinc finger 3
Gene Aliases
AIO, AIOLOS, ZNFN1A3
UniGene
Hs.371680
Gene Chromosome Location
Chr.17: 39757715-39864188 on Build GRCh38
Transcripts
NM_001257408.1 NM_001257409.1 NM_001257410.1 NM_001257411.1 NM_001257412.1 NM_001257413.1 NM_001257414.1 NM_001284514.1 NM_001284515.1 NM_001284516.1 NM_012481.4 NM_183228.2 NM_183229.2 NM_183230.2 NM_183231.2 NM_183232.2

SNP Detail

SNP ID Chromosome Location
rs148998770 Chr.17: 39758038-39758038 on Build GRCh38
rs751860124 Chr.17: 39758200-39758200 on Build GRCh38
rs913410812 Chr.17: 39758366-39758366 on Build GRCh38
rs190888830 Chr.17: 39758049-39758049 on Build GRCh38
rs181552870 Chr.17: 39758392-39758392 on Build GRCh38
rs934257671 Chr.17: 39758051-39758051 on Build GRCh38
rs980543540 Chr.17: 39758326-39758326 on Build GRCh38
rs34873352 Chr.17: 39758254-39758255 on Build GRCh38
rs911499160 Chr.17: 39758413-39758413 on Build GRCh38
rs990197214 Chr.17: 39758110-39758110 on Build GRCh38
rs887440943 Chr.17: 39758414-39758414 on Build GRCh38
rs767773895 Chr.17: 39758341-39758341 on Build GRCh38
rs937405117 Chr.17: 39758100-39758100 on Build GRCh38
rs1013514645 Chr.17: 39758249-39758249 on Build GRCh38
rs925962427 Chr.17: 39758327-39758327 on Build GRCh38
rs971504691 Chr.17: 39758045-39758045 on Build GRCh38
rs960498105 Chr.17: 39758213-39758213 on Build GRCh38
rs983660170 Chr.17: 39758037-39758037 on Build GRCh38
rs889464889 Chr.17: 39758096-39758096 on Build GRCh38
rs986929948 Chr.17: 39758380-39758380 on Build GRCh38
rs143781570 Chr.17: 39758112-39758112 on Build GRCh38
rs1051253336 Chr.17: 39758072-39758072 on Build GRCh38
rs927378854 Chr.17: 39758097-39758097 on Build GRCh38
rs969328959 Chr.17: 39758299-39758299 on Build GRCh38
rs900272501 Chr.17: 39758354-39758354 on Build GRCh38
rs533647817 Chr.17: 39758106-39758106 on Build GRCh38
rs934121758 Chr.17: 39758378-39758378 on Build GRCh38
rs1005099949 Chr.17: 39758446-39758446 on Build GRCh38
rs946657543 Chr.17: 39758117-39758117 on Build GRCh38
rs1023517737 Chr.17: 39758270-39758270 on Build GRCh38
rs944492632 Chr.17: 39758232-39758232 on Build GRCh38
rs532976878 Chr.17: 39758476-39758476 on Build GRCh38
rs1042287825 Chr.17: 39758135-39758135 on Build GRCh38
rs755309042 Chr.17: 39758231-39758231 on Build GRCh38
rs766980594 Chr.17: 39758051-39758051 on Build GRCh38
rs752874672 Chr.17: 39758386-39758386 on Build GRCh38
rs79951252 Chr.17: 39758388-39758388 on Build GRCh38

View Full Product Details