Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 63532592-63533106 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EFCAB7
Entrez Gene ID
84455
Gene Name
EF-hand calcium binding domain 7
Gene Aliases
-
UniGene
Hs.652324
Gene Chromosome Location
Chr.1: 63523301-63585400 on Build GRCh38
Transcripts
NM_032437.3 XM_005271274.3 XM_006710976.3 XM_006710977.1 XM_011542301.2 XM_017002548.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4009323 c.419G>A p.R140Q Chr.1: 63532689-63532689 on Build GRCh38
4909842 c.435C>T p.A145A Chr.1: 63532705-63532705 on Build GRCh38
6651941 c.456A>G p.V152V Chr.1: 63532726-63532726 on Build GRCh38
6416830 c.475G>C p.D159H Chr.1: 63532745-63532745 on Build GRCh38
6223859 c.420A>G p.R140R Chr.1: 63532690-63532690 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs879095818 Chr.1: 63532665-63532665 on Build GRCh38
rs1036830501 Chr.1: 63532946-63532946 on Build GRCh38
rs894722741 Chr.1: 63532838-63532838 on Build GRCh38
rs529167027 Chr.1: 63532735-63532735 on Build GRCh38
rs781099100 Chr.1: 63532759-63532759 on Build GRCh38
rs745722695 Chr.1: 63532993-63532993 on Build GRCh38
rs770086243 Chr.1: 63532690-63532692 on Build GRCh38
rs1036535454 Chr.1: 63532635-63532635 on Build GRCh38
rs746948782 Chr.1: 63532789-63532789 on Build GRCh38
rs773151222 Chr.1: 63532786-63532786 on Build GRCh38
rs764598238 Chr.1: 63532738-63532738 on Build GRCh38
rs957117394 Chr.1: 63532766-63532766 on Build GRCh38
rs746118162 Chr.1: 63532624-63532624 on Build GRCh38
rs757313366 Chr.1: 63532758-63532758 on Build GRCh38
rs772042625 Chr.1: 63532650-63532650 on Build GRCh38
rs548650185 Chr.1: 63532740-63532740 on Build GRCh38
rs779946842 Chr.1: 63532656-63532656 on Build GRCh38
rs759233815 Chr.1: 63532689-63532689 on Build GRCh38
rs1045692947 Chr.1: 63532923-63532923 on Build GRCh38
rs986766345 Chr.1: 63532853-63532853 on Build GRCh38
rs563807147 Chr.1: 63532639-63532639 on Build GRCh38
rs760173637 Chr.1: 63532680-63532680 on Build GRCh38
rs931434240 Chr.1: 63532992-63532992 on Build GRCh38
rs773572871 Chr.1: 63532725-63532725 on Build GRCh38
rs745993196 Chr.1: 63532765-63532765 on Build GRCh38
rs756311368 Chr.1: 63532772-63532772 on Build GRCh38
rs9803822 Chr.1: 63533073-63533073 on Build GRCh38
rs190070353 Chr.1: 63532692-63532692 on Build GRCh38
rs780317441 Chr.1: 63532773-63532773 on Build GRCh38
rs375797926 Chr.1: 63532780-63532780 on Build GRCh38
rs966873880 Chr.1: 63532677-63532677 on Build GRCh38
rs748335379 Chr.1: 63532677-63532679 on Build GRCh38
rs776384502 Chr.1: 63532688-63532688 on Build GRCh38
rs747133720 Chr.1: 63532655-63532656 on Build GRCh38
rs542383100 Chr.1: 63533067-63533068 on Build GRCh38
rs770943115 Chr.1: 63532790-63532790 on Build GRCh38
rs537407011 Chr.1: 63532855-63532855 on Build GRCh38
rs185089010 Chr.1: 63532619-63532619 on Build GRCh38
rs371843664 Chr.1: 63532618-63532618 on Build GRCh38
rs939836859 Chr.1: 63532907-63532907 on Build GRCh38
rs1033497529 Chr.1: 63533018-63533018 on Build GRCh38
rs765826760 Chr.1: 63532683-63532683 on Build GRCh38
rs767030873 Chr.1: 63532806-63532807 on Build GRCh38
rs912483680 Chr.1: 63532874-63532874 on Build GRCh38
rs753098176 Chr.1: 63532731-63532731 on Build GRCh38
rs202142800 Chr.1: 63532655-63532655 on Build GRCh38
rs1014467282 Chr.1: 63532879-63532879 on Build GRCh38
rs954081984 Chr.1: 63532844-63532844 on Build GRCh38
rs957463443 Chr.1: 63533046-63533046 on Build GRCh38
rs778901115 Chr.1: 63532985-63532985 on Build GRCh38
rs199986363 Chr.1: 63532693-63532693 on Build GRCh38
rs151059690 Chr.1: 63532754-63532754 on Build GRCh38
rs759175391 Chr.1: 63532799-63532799 on Build GRCh38
rs200699317 Chr.1: 63532645-63532645 on Build GRCh38
rs920980033 Chr.1: 63532819-63532819 on Build GRCh38
rs774106386 Chr.1: 63532642-63532642 on Build GRCh38
rs776260524 Chr.1: 63532798-63532798 on Build GRCh38
rs998920713 Chr.1: 63533002-63533002 on Build GRCh38
rs763384780 Chr.1: 63532742-63532742 on Build GRCh38
rs771776511 Chr.1: 63532785-63532785 on Build GRCh38
rs999660225 Chr.1: 63533076-63533076 on Build GRCh38
rs72163225 Chr.1: 63532660-63532661 on Build GRCh38
rs773027833 Chr.1: 63532666-63532666 on Build GRCh38

View Full Product Details