Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 137605264-137605754 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARRDC1
Entrez Gene ID
92714
Gene Name
arrestin domain containing 1
Gene Aliases
-
UniGene
Hs.12999
Gene Chromosome Location
Chr.9: 137605640-137615360 on Build GRCh38
Transcripts
NM_001317968.1 NM_152285.3 XM_005266119.1

SNP Detail

SNP ID Chromosome Location
rs954981672 Chr.9: 137605357-137605357 on Build GRCh38
rs1028326562 Chr.9: 137605689-137605689 on Build GRCh38
rs1049726796 Chr.9: 137605435-137605436 on Build GRCh38
rs775827204 Chr.9: 137605682-137605682 on Build GRCh38
rs771849009 Chr.9: 137605356-137605356 on Build GRCh38
rs866023752 Chr.9: 137605616-137605616 on Build GRCh38
rs953232349 Chr.9: 137605693-137605693 on Build GRCh38
rs960918086 Chr.9: 137605544-137605544 on Build GRCh38
rs959644394 Chr.9: 137605303-137605303 on Build GRCh38
rs773926944 Chr.9: 137605690-137605690 on Build GRCh38
rs905367257 Chr.9: 137605494-137605495 on Build GRCh38
rs979838728 Chr.9: 137605280-137605280 on Build GRCh38
rs990880889 Chr.9: 137605306-137605306 on Build GRCh38
rs1002332173 Chr.9: 137605503-137605503 on Build GRCh38
rs186573104 Chr.9: 137605403-137605403 on Build GRCh38
rs962199647 Chr.9: 137605546-137605546 on Build GRCh38
rs1010011877 Chr.9: 137605604-137605604 on Build GRCh38
rs931095595 Chr.9: 137605640-137605640 on Build GRCh38
rs769022197 Chr.9: 137605688-137605688 on Build GRCh38
rs763365173 Chr.9: 137605685-137605685 on Build GRCh38
rs897594876 Chr.9: 137605384-137605384 on Build GRCh38
rs1057025648 Chr.9: 137605644-137605656 on Build GRCh38
rs779003976 Chr.9: 137605680-137605681 on Build GRCh38
rs972434549 Chr.9: 137605558-137605558 on Build GRCh38
rs760501724 Chr.9: 137605733-137605733 on Build GRCh38
rs918019862 Chr.9: 137605700-137605700 on Build GRCh38
rs750001196 Chr.9: 137605725-137605725 on Build GRCh38
rs919661094 Chr.9: 137605595-137605595 on Build GRCh38
rs865919125 Chr.9: 137605290-137605290 on Build GRCh38
rs766890761 Chr.9: 137605705-137605705 on Build GRCh38
rs1035288703 Chr.9: 137605519-137605519 on Build GRCh38
rs540171161 Chr.9: 137605663-137605663 on Build GRCh38
rs944390769 Chr.9: 137605326-137605326 on Build GRCh38
rs761485602 Chr.9: 137605703-137605703 on Build GRCh38
rs968572158 Chr.9: 137605649-137605649 on Build GRCh38
rs1032377761 Chr.9: 137605296-137605296 on Build GRCh38
rs1015060348 Chr.9: 137605481-137605481 on Build GRCh38
rs1036017654 Chr.9: 137605353-137605353 on Build GRCh38
rs994592172 Chr.9: 137605410-137605410 on Build GRCh38
rs981270529 Chr.9: 137605720-137605720 on Build GRCh38
rs553621923 Chr.9: 137605669-137605669 on Build GRCh38
rs911787940 Chr.9: 137605321-137605321 on Build GRCh38
rs988131812 Chr.9: 137605415-137605415 on Build GRCh38

View Full Product Details