Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 137614871-137615373 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARRDC1
Entrez Gene ID
92714
Gene Name
arrestin domain containing 1
Gene Aliases
-
UniGene
Hs.12999
Gene Chromosome Location
Chr.9: 137605640-137615360 on Build GRCh38
Transcripts
NM_001317968.1 NM_152285.3 XM_005266119.1 XM_006717320.3 XM_006717321.3 XM_006717322.3 XM_017015288.1 NR_122035.1 NR_122036.1

Gene Symbol
ARRDC1-AS1
Entrez Gene ID
85026
Gene Name
ARRDC1 antisense RNA 1
Gene Aliases
C9orf37
UniGene
Hs.522520
Gene Chromosome Location
Chr.9: 137615332-137618895 on Build GRCh38
Transcripts
NM_001317968.1 NM_152285.3 XM_005266119.1 XM_006717320.3 XM_006717321.3 XM_006717322.3 XM_017015288.1 NR_122035.1 NR_122036.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3089574 c.1254T>C p.Y418Y Chr.9: 137615090-137615090 on Build GRCh38
4152207 c.1213T>C p.Y405H Chr.9: 137614976-137614976 on Build GRCh38
3089572 c.1136G>A p.G379D Chr.9: 137614899-137614899 on Build GRCh38
1190817 c.1291_1292delCC p.P431fs*1 Chr.9: 137615127-137615128 on Build GRCh38
7356492 c.1208C>A p.P403Q Chr.9: 137614971-137614971 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs769357115 Chr.9: 137615108-137615108 on Build GRCh38
rs754724073 Chr.9: 137615024-137615024 on Build GRCh38
rs763362421 Chr.9: 137614991-137614991 on Build GRCh38
rs760355513 Chr.9: 137615139-137615139 on Build GRCh38
rs201878624 Chr.9: 137615102-137615102 on Build GRCh38
rs1031701630 Chr.9: 137614964-137614964 on Build GRCh38
rs770885209 Chr.9: 137614979-137614979 on Build GRCh38
rs890243555 Chr.9: 137615344-137615344 on Build GRCh38
rs931681063 Chr.9: 137615289-137615289 on Build GRCh38
rs532705950 Chr.9: 137615008-137615008 on Build GRCh38
rs757365279 Chr.9: 137614930-137614931 on Build GRCh38
rs538409772 Chr.9: 137615143-137615143 on Build GRCh38
rs995887832 Chr.9: 137614907-137614907 on Build GRCh38
rs750019684 Chr.9: 137615016-137615016 on Build GRCh38
rs3177227 Chr.9: 137615347-137615347 on Build GRCh38
rs763785406 Chr.9: 137614928-137614928 on Build GRCh38
rs3174164 Chr.9: 137615263-137615263 on Build GRCh38
rs1032794460 Chr.9: 137614968-137614968 on Build GRCh38
rs776819816 Chr.9: 137614981-137614981 on Build GRCh38
rs766927533 Chr.9: 137615014-137615014 on Build GRCh38
rs141454610 Chr.9: 137615078-137615078 on Build GRCh38
rs919536531 Chr.9: 137615224-137615224 on Build GRCh38
rs765672051 Chr.9: 137614906-137614906 on Build GRCh38
rs770071956 Chr.9: 137614990-137614991 on Build GRCh38
rs755628016 Chr.9: 137615022-137615022 on Build GRCh38
rs745905042 Chr.9: 137615038-137615038 on Build GRCh38
rs952338408 Chr.9: 137615236-137615236 on Build GRCh38
rs771241596 Chr.9: 137615036-137615036 on Build GRCh38
rs770487926 Chr.9: 137614988-137614988 on Build GRCh38
rs781125964 Chr.9: 137615037-137615037 on Build GRCh38
rs760125003 Chr.9: 137614904-137614904 on Build GRCh38
rs373166866 Chr.9: 137614896-137614896 on Build GRCh38
rs553044554 Chr.9: 137615253-137615253 on Build GRCh38
rs1001357903 Chr.9: 137614926-137614926 on Build GRCh38
rs749316708 Chr.9: 137615180-137615180 on Build GRCh38
rs775913729 Chr.9: 137615076-137615092 on Build GRCh38
rs953113345 Chr.9: 137614973-137614973 on Build GRCh38
rs372424462 Chr.9: 137615049-137615049 on Build GRCh38
rs747175743 Chr.9: 137615035-137615035 on Build GRCh38
rs778720078 Chr.9: 137615032-137615032 on Build GRCh38
rs753167592 Chr.9: 137614910-137614910 on Build GRCh38
rs141470293 Chr.9: 137614899-137614899 on Build GRCh38
rs748490636 Chr.9: 137615118-137615118 on Build GRCh38
rs181357513 Chr.9: 137615272-137615272 on Build GRCh38
rs768325901 Chr.9: 137615128-137615129 on Build GRCh38
rs776100932 Chr.9: 137615043-137615043 on Build GRCh38
rs779822289 Chr.9: 137614954-137614954 on Build GRCh38
rs370236867 Chr.9: 137615086-137615086 on Build GRCh38
rs758130796 Chr.9: 137615091-137615091 on Build GRCh38
rs148707333 Chr.9: 137614930-137614930 on Build GRCh38
rs375659457 Chr.9: 137615070-137615070 on Build GRCh38
rs752600066 Chr.9: 137615164-137615164 on Build GRCh38
rs949965454 Chr.9: 137615273-137615273 on Build GRCh38
rs781772629 Chr.9: 137614931-137614931 on Build GRCh38
rs780330268 Chr.9: 137615105-137615105 on Build GRCh38
rs768449695 Chr.9: 137614965-137614965 on Build GRCh38
rs538977695 Chr.9: 137615028-137615028 on Build GRCh38
rs371006761 Chr.9: 137615106-137615106 on Build GRCh38
rs995856621 Chr.9: 137615111-137615111 on Build GRCh38
rs761370288 Chr.9: 137615050-137615050 on Build GRCh38
rs746311327 Chr.9: 137614971-137614972 on Build GRCh38
rs1000134992 Chr.9: 137615336-137615336 on Build GRCh38
rs756232027 Chr.9: 137615103-137615103 on Build GRCh38
rs928067629 Chr.9: 137615192-137615192 on Build GRCh38
rs765153130 Chr.9: 137615148-137615148 on Build GRCh38
rs201013536 Chr.9: 137614946-137614946 on Build GRCh38
rs143543277 Chr.9: 137614963-137614963 on Build GRCh38
rs756136351 Chr.9: 137614947-137614947 on Build GRCh38
rs759945901 Chr.9: 137614985-137614985 on Build GRCh38
rs770016947 Chr.9: 137615041-137615041 on Build GRCh38
rs374533199 Chr.9: 137615099-137615099 on Build GRCh38
rs774728153 Chr.9: 137615109-137615109 on Build GRCh38
rs1475494 Chr.9: 137615329-137615329 on Build GRCh38
rs552792074 Chr.9: 137615047-137615047 on Build GRCh38
rs59628511 Chr.9: 137615202-137615202 on Build GRCh38
rs764585267 Chr.9: 137615007-137615007 on Build GRCh38
rs368733277 Chr.9: 137615184-137615184 on Build GRCh38
rs114986132 Chr.9: 137615271-137615271 on Build GRCh38
rs769591479 Chr.9: 137615322-137615322 on Build GRCh38
rs112136439 Chr.9: 137615223-137615223 on Build GRCh38
rs759464082 Chr.9: 137615146-137615146 on Build GRCh38
rs374293181 Chr.9: 137615142-137615142 on Build GRCh38
rs754957845 Chr.9: 137615088-137615088 on Build GRCh38
rs756466968 Chr.9: 137615177-137615177 on Build GRCh38
rs758174127 Chr.9: 137615165-137615165 on Build GRCh38
rs758603274 Chr.9: 137615083-137615083 on Build GRCh38
rs148038871 Chr.9: 137614978-137614978 on Build GRCh38
rs372586162 Chr.9: 137615171-137615171 on Build GRCh38
rs369356952 Chr.9: 137614989-137614989 on Build GRCh38
rs780280953 Chr.9: 137615179-137615179 on Build GRCh38
rs569374739 Chr.9: 137615090-137615090 on Build GRCh38
rs765901267 Chr.9: 137615077-137615077 on Build GRCh38
rs14357 Chr.9: 137615249-137615249 on Build GRCh38
rs376611695 Chr.9: 137615172-137615172 on Build GRCh38
rs748392202 Chr.9: 137614931-137614931 on Build GRCh38
rs756686281 Chr.9: 137614936-137614936 on Build GRCh38
rs772878243 Chr.9: 137615136-137615136 on Build GRCh38
rs2501563 Chr.9: 137615190-137615190 on Build GRCh38
rs372157985 Chr.9: 137615189-137615189 on Build GRCh38
rs761580021 Chr.9: 137615012-137615012 on Build GRCh38
rs779906135 Chr.9: 137615023-137615023 on Build GRCh38
rs772342951 Chr.9: 137615127-137615127 on Build GRCh38
rs1018691489 Chr.9: 137615214-137615215 on Build GRCh38
rs12554630 Chr.9: 137615201-137615201 on Build GRCh38
rs1049044372 Chr.9: 137615059-137615059 on Build GRCh38
rs777920141 Chr.9: 137614975-137614975 on Build GRCh38
rs767133720 Chr.9: 137615069-137615069 on Build GRCh38
rs780795182 Chr.9: 137614943-137614943 on Build GRCh38
rs530116866 Chr.9: 137614911-137614911 on Build GRCh38
rs912081718 Chr.9: 137615017-137615017 on Build GRCh38
rs748963930 Chr.9: 137615079-137615081 on Build GRCh38
rs773780626 Chr.9: 137615139-137615139 on Build GRCh38
rs966634114 Chr.9: 137615218-137615218 on Build GRCh38

View Full Product Details