Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 50248164-50248701 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ASAH2
Entrez Gene ID
56624
Gene Name
N-acylsphingosine amidohydrolase 2
Gene Aliases
BCDase, HNAC1, LCDase, N-CDase, NCDase
UniGene
Hs.512645
Gene Chromosome Location
Chr.10: 50184861-50251564 on Build GRCh38
Transcripts
NM_001143974.1 NM_019893.2 XM_011539970.2 XM_011539971.2 XM_011539972.2 XM_011539973.2 XM_017016435.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4934176 c.11G>A p.R4H Chr.10: 50248600-50248600 on Build GRCh38
6461201 c.85A>T p.S29C Chr.10: 50248526-50248526 on Build GRCh38
1675230 c.79C>A p.L27I Chr.10: 50248532-50248532 on Build GRCh38
2146188 c.73G>A p.V25M Chr.10: 50248538-50248538 on Build GRCh38
5526990 c.45C>T p.F15F Chr.10: 50248566-50248566 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs751691065 Chr.10: 50248532-50248532 on Build GRCh38
rs959479671 Chr.10: 50248317-50248317 on Build GRCh38
rs996875543 Chr.10: 50248233-50248233 on Build GRCh38
rs776206430 Chr.10: 50248495-50248495 on Build GRCh38
rs746545115 Chr.10: 50248575-50248575 on Build GRCh38
rs763814274 Chr.10: 50248443-50248443 on Build GRCh38
rs758080831 Chr.10: 50248354-50248354 on Build GRCh38
rs775227080 Chr.10: 50248562-50248562 on Build GRCh38
rs961173989 Chr.10: 50248311-50248311 on Build GRCh38
rs922133248 Chr.10: 50248490-50248490 on Build GRCh38
rs748449565 Chr.10: 50248537-50248537 on Build GRCh38
rs760066642 Chr.10: 50248436-50248436 on Build GRCh38
rs762780479 Chr.10: 50248362-50248362 on Build GRCh38
rs7091407 Chr.10: 50248228-50248228 on Build GRCh38
rs1003750985 Chr.10: 50248645-50248645 on Build GRCh38
rs756545409 Chr.10: 50248653-50248653 on Build GRCh38
rs1012313450 Chr.10: 50248660-50248660 on Build GRCh38
rs763187531 Chr.10: 50248605-50248605 on Build GRCh38
rs991344711 Chr.10: 50248323-50248323 on Build GRCh38
rs368446245 Chr.10: 50248578-50248578 on Build GRCh38
rs945516417 Chr.10: 50248584-50248584 on Build GRCh38
rs751951726 Chr.10: 50248626-50248626 on Build GRCh38
rs754944432 Chr.10: 50248534-50248534 on Build GRCh38
rs1049402592 Chr.10: 50248258-50248258 on Build GRCh38
rs1040548949 Chr.10: 50248482-50248482 on Build GRCh38
rs7071083 Chr.10: 50248380-50248380 on Build GRCh38
rs778901441 Chr.10: 50248542-50248542 on Build GRCh38
rs983585360 Chr.10: 50248480-50248480 on Build GRCh38
rs756428088 Chr.10: 50248641-50248641 on Build GRCh38
rs756374986 Chr.10: 50248538-50248538 on Build GRCh38
rs535439765 Chr.10: 50248274-50248274 on Build GRCh38
rs1035483960 Chr.10: 50248316-50248316 on Build GRCh38
rs376205298 Chr.10: 50248600-50248600 on Build GRCh38
rs775059431 Chr.10: 50248391-50248391 on Build GRCh38
rs758953048 Chr.10: 50248518-50248518 on Build GRCh38
rs765653388 Chr.10: 50248508-50248508 on Build GRCh38
rs1027872510 Chr.10: 50248364-50248364 on Build GRCh38
rs567285940 Chr.10: 50248286-50248286 on Build GRCh38
rs180822330 Chr.10: 50248514-50248514 on Build GRCh38
rs946850672 Chr.10: 50248668-50248668 on Build GRCh38
rs569878470 Chr.10: 50248271-50248271 on Build GRCh38
rs766746773 Chr.10: 50248525-50248525 on Build GRCh38
rs115286664 Chr.10: 50248355-50248355 on Build GRCh38
rs1034849885 Chr.10: 50248651-50248651 on Build GRCh38
rs944821649 Chr.10: 50248474-50248474 on Build GRCh38
rs746212421 Chr.10: 50248652-50248652 on Build GRCh38
rs543686006 Chr.10: 50248661-50248661 on Build GRCh38
rs761489791 Chr.10: 50248594-50248594 on Build GRCh38
rs922806271 Chr.10: 50248200-50248200 on Build GRCh38
rs746988753 Chr.10: 50248224-50248224 on Build GRCh38
rs2813305 Chr.10: 50248209-50248209 on Build GRCh38
rs1015490259 Chr.10: 50248303-50248303 on Build GRCh38
rs959494071 Chr.10: 50248656-50248656 on Build GRCh38
rs1041163366 Chr.10: 50248221-50248221 on Build GRCh38
rs760085939 Chr.10: 50248629-50248629 on Build GRCh38
rs772081662 Chr.10: 50248549-50248549 on Build GRCh38
rs201170544 Chr.10: 50248435-50248435 on Build GRCh38
rs2813304 Chr.10: 50248314-50248314 on Build GRCh38
rs367923748 Chr.10: 50248637-50248637 on Build GRCh38
rs745613507 Chr.10: 50248547-50248547 on Build GRCh38
rs557694994 Chr.10: 50248198-50248198 on Build GRCh38
rs768029145 Chr.10: 50248621-50248621 on Build GRCh38
rs202096152 Chr.10: 50248633-50248633 on Build GRCh38
rs781320322 Chr.10: 50248535-50248535 on Build GRCh38
rs752803685 Chr.10: 50248638-50248638 on Build GRCh38
rs776230724 Chr.10: 50248252-50248252 on Build GRCh38
rs1003496381 Chr.10: 50248293-50248293 on Build GRCh38
rs878859007 Chr.10: 50248422-50248422 on Build GRCh38
rs560016121 Chr.10: 50248249-50248249 on Build GRCh38
rs760334150 Chr.10: 50248485-50248485 on Build GRCh38
rs550045925 Chr.10: 50248241-50248241 on Build GRCh38
rs578123995 Chr.10: 50248601-50248601 on Build GRCh38
rs924202430 Chr.10: 50248449-50248449 on Build GRCh38
rs114108618 Chr.10: 50248498-50248498 on Build GRCh38

View Full Product Details