Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 63185474-63185985 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
JMJD1C
Entrez Gene ID
221037
Gene Name
jumonji domain containing 1C
Gene Aliases
TRIP-8, TRIP8
UniGene
Hs.413416
Gene Chromosome Location
Chr.10: 63167221-63522075 on Build GRCh38
Transcripts
NM_001282948.1 NM_001318153.1 NM_001318154.1 NM_001322252.1 NM_001322254.1 NM_001322258.1 NM_032776.2 NR_134512.1 XM_011539503.1 XM_017015897.1 XM_017015898.1 XM_017015899.1 XM_017015900.1 XM_017015901.1 XM_017015902.1 XM_017015903.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
919452 c.6119G>A p.R2040K Chr.10: 63185563-63185563 on Build GRCh38
3935118 c.6086C>T p.S2029L Chr.10: 63185596-63185596 on Build GRCh38
1739652 c.6106A>G p.M2036V Chr.10: 63185576-63185576 on Build GRCh38
3935117 c.6797C>T p.S2266L Chr.10: 63185596-63185596 on Build GRCh38
1739651 c.6817A>G p.M2273V Chr.10: 63185576-63185576 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs922788731 Chr.10: 63185567-63185567 on Build GRCh38
rs759063415 Chr.10: 63185526-63185526 on Build GRCh38
rs758571360 Chr.10: 63185589-63185589 on Build GRCh38
rs895219347 Chr.10: 63185784-63185784 on Build GRCh38
rs1022584114 Chr.10: 63185514-63185514 on Build GRCh38
rs1001156296 Chr.10: 63185808-63185808 on Build GRCh38
rs937137342 Chr.10: 63185726-63185726 on Build GRCh38
rs767965203 Chr.10: 63185531-63185531 on Build GRCh38
rs957105920 Chr.10: 63185530-63185530 on Build GRCh38
rs373934433 Chr.10: 63185576-63185576 on Build GRCh38
rs1017334450 Chr.10: 63185931-63185931 on Build GRCh38
rs545637201 Chr.10: 63185711-63185711 on Build GRCh38
rs190438141 Chr.10: 63185894-63185894 on Build GRCh38
rs761207633 Chr.10: 63185533-63185533 on Build GRCh38
rs776005261 Chr.10: 63185532-63185532 on Build GRCh38
rs930051070 Chr.10: 63185764-63185764 on Build GRCh38
rs773672311 Chr.10: 63185525-63185525 on Build GRCh38
rs920535799 Chr.10: 63185706-63185706 on Build GRCh38
rs561874207 Chr.10: 63185773-63185773 on Build GRCh38
rs573024840 Chr.10: 63185765-63185765 on Build GRCh38
rs3211105 Chr.10: 63185604-63185604 on Build GRCh38
rs545430823 Chr.10: 63185919-63185919 on Build GRCh38
rs955542361 Chr.10: 63185902-63185902 on Build GRCh38
rs770435943 Chr.10: 63185674-63185674 on Build GRCh38
rs1030244837 Chr.10: 63185838-63185838 on Build GRCh38
rs973080294 Chr.10: 63185945-63185945 on Build GRCh38
rs929221469 Chr.10: 63185718-63185718 on Build GRCh38
rs558664422 Chr.10: 63185870-63185870 on Build GRCh38
rs944048072 Chr.10: 63185800-63185800 on Build GRCh38
rs973876556 Chr.10: 63185744-63185744 on Build GRCh38
rs939451782 Chr.10: 63185751-63185751 on Build GRCh38
rs754282649 Chr.10: 63185539-63185539 on Build GRCh38
rs750471016 Chr.10: 63185586-63185586 on Build GRCh38
rs1001040688 Chr.10: 63185508-63185508 on Build GRCh38
rs981353505 Chr.10: 63185622-63185622 on Build GRCh38
rs748109864 Chr.10: 63185506-63185506 on Build GRCh38
rs150646588 Chr.10: 63185793-63185800 on Build GRCh38
rs749117336 Chr.10: 63185660-63185660 on Build GRCh38
rs182335218 Chr.10: 63185727-63185727 on Build GRCh38
rs757910065 Chr.10: 63185807-63185807 on Build GRCh38
rs999749998 Chr.10: 63185814-63185814 on Build GRCh38
rs202076128 Chr.10: 63185523-63185523 on Build GRCh38
rs919796688 Chr.10: 63185754-63185754 on Build GRCh38
rs964410928 Chr.10: 63185942-63185942 on Build GRCh38
rs748077584 Chr.10: 63185618-63185618 on Build GRCh38
rs774018251 Chr.10: 63185693-63185693 on Build GRCh38
rs757416787 Chr.10: 63185551-63185551 on Build GRCh38
rs1039920688 Chr.10: 63185803-63185803 on Build GRCh38
rs775349157 Chr.10: 63185550-63185551 on Build GRCh38
rs762881894 Chr.10: 63185616-63185617 on Build GRCh38
rs542084800 Chr.10: 63185771-63185771 on Build GRCh38
rs1032503472 Chr.10: 63185864-63185864 on Build GRCh38
rs756136220 Chr.10: 63185632-63185632 on Build GRCh38
rs1037315641 Chr.10: 63185682-63185682 on Build GRCh38
rs987638127 Chr.10: 63185909-63185909 on Build GRCh38
rs185526726 Chr.10: 63185868-63185868 on Build GRCh38
rs777426701 Chr.10: 63185649-63185649 on Build GRCh38
rs775962939 Chr.10: 63185730-63185733 on Build GRCh38
rs370959235 Chr.10: 63185535-63185535 on Build GRCh38
rs376798332 Chr.10: 63185513-63185513 on Build GRCh38
rs373552182 Chr.10: 63185803-63185810 on Build GRCh38

View Full Product Details