Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 31817548-31818061 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HSPA1A
Entrez Gene ID
3303
Gene Name
heat shock protein family A (Hsp70) member 1A
Gene Aliases
HEL-S-103, HSP70-1, HSP70-1A, HSP70.1, HSP70I, HSP72, HSPA1
UniGene
Hs.274402
Gene Chromosome Location
Chr.6: 31815514-31817942 on Build GRCh38
Transcripts
NM_005345.5

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3861843 c.1867G>T p.G623W Chr.6: 31817623-31817623 on Build GRCh38
6562511 c.1886G>C p.G629A Chr.6: 31817642-31817642 on Build GRCh38
6682561 c.1834C>A p.Q612K Chr.6: 31817590-31817590 on Build GRCh38
3674660 c.1843G>T p.G615C Chr.6: 31817599-31817599 on Build GRCh38
1077722 c.1876G>T p.G626C Chr.6: 31817632-31817632 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs764202242 Chr.6: 31817605-31817605 on Build GRCh38
rs758541631 Chr.6: 31817869-31817869 on Build GRCh38
rs753771041 Chr.6: 31817611-31817611 on Build GRCh38
rs508633 Chr.6: 31817676-31817676 on Build GRCh38
rs758117058 Chr.6: 31817618-31817618 on Build GRCh38
rs566619820 Chr.6: 31817820-31817820 on Build GRCh38
rs527830727 Chr.6: 31817822-31817822 on Build GRCh38
rs199732319 Chr.6: 31817712-31817712 on Build GRCh38
rs508603 Chr.6: 31817661-31817661 on Build GRCh38
rs902873936 Chr.6: 31817948-31817948 on Build GRCh38
rs962334802 Chr.6: 31817804-31817804 on Build GRCh38
rs763188460 Chr.6: 31817600-31817600 on Build GRCh38
rs562455991 Chr.6: 31817707-31817707 on Build GRCh38
rs765239235 Chr.6: 31817580-31817580 on Build GRCh38
rs551174012 Chr.6: 31817777-31817777 on Build GRCh38
rs1014963830 Chr.6: 31817778-31817778 on Build GRCh38
rs571973573 Chr.6: 31818007-31818007 on Build GRCh38
rs935136269 Chr.6: 31817887-31817891 on Build GRCh38
rs777256711 Chr.6: 31817620-31817620 on Build GRCh38
rs746592204 Chr.6: 31817622-31817622 on Build GRCh38
rs774424456 Chr.6: 31817695-31817695 on Build GRCh38
rs770410665 Chr.6: 31817623-31817623 on Build GRCh38
rs947192690 Chr.6: 31817918-31817918 on Build GRCh38
rs777286374 Chr.6: 31817570-31817570 on Build GRCh38
rs567588232 Chr.6: 31817950-31817950 on Build GRCh38
rs991120720 Chr.6: 31818023-31818023 on Build GRCh38
rs754825210 Chr.6: 31817614-31817614 on Build GRCh38
rs750583276 Chr.6: 31817832-31817832 on Build GRCh38
rs996814855 Chr.6: 31817949-31817949 on Build GRCh38
rs1014920762 Chr.6: 31817970-31817970 on Build GRCh38
rs981743345 Chr.6: 31817807-31817807 on Build GRCh38
rs761795494 Chr.6: 31817609-31817609 on Build GRCh38
rs923688845 Chr.6: 31817858-31817858 on Build GRCh38
rs752068654 Chr.6: 31817686-31817686 on Build GRCh38
rs914332664 Chr.6: 31817894-31817894 on Build GRCh38
rs1046146122 Chr.6: 31817841-31817841 on Build GRCh38
rs771512819 Chr.6: 31817569-31817569 on Build GRCh38
rs778809676 Chr.6: 31817616-31817616 on Build GRCh38
rs529632338 Chr.6: 31817967-31817967 on Build GRCh38
rs768990311 Chr.6: 31817677-31817677 on Build GRCh38
rs751703293 Chr.6: 31817608-31817608 on Build GRCh38
rs561574210 Chr.6: 31817584-31817584 on Build GRCh38
rs34629121 Chr.6: 31817917-31817918 on Build GRCh38
rs952729121 Chr.6: 31817685-31817685 on Build GRCh38
rs761966416 Chr.6: 31817698-31817698 on Build GRCh38
rs775974928 Chr.6: 31817662-31817662 on Build GRCh38
rs549339651 Chr.6: 31817842-31817842 on Build GRCh38
rs529162552 Chr.6: 31817654-31817654 on Build GRCh38
rs781669728 Chr.6: 31817642-31817642 on Build GRCh38
rs771505399 Chr.6: 31817573-31817573 on Build GRCh38
rs538255931 Chr.6: 31817969-31817969 on Build GRCh38
rs994962530 Chr.6: 31818001-31818001 on Build GRCh38
rs41269729 Chr.6: 31817898-31817898 on Build GRCh38
rs757711628 Chr.6: 31817726-31817727 on Build GRCh38
rs991580935 Chr.6: 31817884-31817884 on Build GRCh38
rs766482400 Chr.6: 31817610-31817610 on Build GRCh38
rs777736210 Chr.6: 31817668-31817670 on Build GRCh38
rs746476471 Chr.6: 31817572-31817572 on Build GRCh38
rs775389464 Chr.6: 31817951-31817951 on Build GRCh38
rs899711212 Chr.6: 31818020-31818020 on Build GRCh38
rs775724932 Chr.6: 31817599-31817599 on Build GRCh38
rs759946698 Chr.6: 31817581-31817581 on Build GRCh38
rs376942723 Chr.6: 31817767-31817767 on Build GRCh38
rs28470887 Chr.6: 31818015-31818015 on Build GRCh38
rs777114966 Chr.6: 31817574-31817574 on Build GRCh38
rs1041856466 Chr.6: 31817921-31817921 on Build GRCh38
rs556867428 Chr.6: 31817971-31817971 on Build GRCh38
rs532931636 Chr.6: 31817738-31817738 on Build GRCh38
rs368938912 Chr.6: 31817821-31817821 on Build GRCh38
rs752313605 Chr.6: 31817617-31817617 on Build GRCh38

View Full Product Details