Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 35582785-35583331 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FKBP5
Entrez Gene ID
2289
Gene Name
FK506 binding protein 5
Gene Aliases
AIG6, FKBP51, FKBP54, P54, PPIase, Ptg-10
UniGene
Hs.407190
Gene Chromosome Location
Chr.6: 35573585-35728583 on Build GRCh38
Transcripts
NM_001145775.2 NM_001145776.1 NM_001145777.1 NM_004117.3

SNP Detail

SNP ID Chromosome Location
rs145888809 Chr.6: 35583100-35583100 on Build GRCh38
rs1017933751 Chr.6: 35582942-35582942 on Build GRCh38
rs924385413 Chr.6: 35583026-35583026 on Build GRCh38
rs34583947 Chr.6: 35583198-35583199 on Build GRCh38
rs1007796043 Chr.6: 35583199-35583199 on Build GRCh38
rs965341156 Chr.6: 35583203-35583203 on Build GRCh38
rs972332498 Chr.6: 35583230-35583230 on Build GRCh38
rs74781741 Chr.6: 35582925-35582925 on Build GRCh38
rs931066188 Chr.6: 35582859-35582859 on Build GRCh38
rs891848360 Chr.6: 35583123-35583123 on Build GRCh38
rs761677067 Chr.6: 35582836-35582836 on Build GRCh38
rs916356530 Chr.6: 35583268-35583268 on Build GRCh38
rs1051583937 Chr.6: 35583188-35583188 on Build GRCh38
rs910117211 Chr.6: 35583146-35583146 on Build GRCh38
rs1017548998 Chr.6: 35583210-35583210 on Build GRCh38
rs530647638 Chr.6: 35582882-35582882 on Build GRCh38
rs59320339 Chr.6: 35583138-35583138 on Build GRCh38
rs375917826 Chr.6: 35582898-35582898 on Build GRCh38
rs56889814 Chr.6: 35583089-35583090 on Build GRCh38
rs998286239 Chr.6: 35583177-35583177 on Build GRCh38
rs953993602 Chr.6: 35583161-35583161 on Build GRCh38
rs10631894 Chr.6: 35583079-35583080 on Build GRCh38
rs986075249 Chr.6: 35582940-35582940 on Build GRCh38
rs968586283 Chr.6: 35583227-35583227 on Build GRCh38
rs745656978 Chr.6: 35582821-35582821 on Build GRCh38
rs918311066 Chr.6: 35582854-35582854 on Build GRCh38
rs887939032 Chr.6: 35582915-35582915 on Build GRCh38
rs1037019003 Chr.6: 35583164-35583164 on Build GRCh38
rs765184115 Chr.6: 35583019-35583019 on Build GRCh38
rs954673863 Chr.6: 35582938-35582938 on Build GRCh38
rs978382392 Chr.6: 35583003-35583003 on Build GRCh38
rs11967924 Chr.6: 35583196-35583196 on Build GRCh38
rs917431557 Chr.6: 35583072-35583072 on Build GRCh38
rs376167865 Chr.6: 35583090-35583090 on Build GRCh38
rs1030190464 Chr.6: 35582937-35582937 on Build GRCh38
rs371128385 Chr.6: 35582832-35582832 on Build GRCh38
rs907481586 Chr.6: 35583064-35583064 on Build GRCh38
rs918260175 Chr.6: 35583242-35583242 on Build GRCh38
rs7754640 Chr.6: 35583252-35583252 on Build GRCh38
rs1031697201 Chr.6: 35583074-35583074 on Build GRCh38
rs934428990 Chr.6: 35583042-35583042 on Build GRCh38
rs1049185574 Chr.6: 35582874-35582874 on Build GRCh38
rs993357054 Chr.6: 35582824-35582824 on Build GRCh38
rs553794638 Chr.6: 35582878-35582878 on Build GRCh38
rs779908362 Chr.6: 35583125-35583125 on Build GRCh38
rs758336264 Chr.6: 35583053-35583053 on Build GRCh38
rs1003258937 Chr.6: 35583073-35583073 on Build GRCh38
rs985338243 Chr.6: 35583172-35583172 on Build GRCh38
rs968446129 Chr.6: 35582959-35582959 on Build GRCh38
rs931013840 Chr.6: 35583248-35583248 on Build GRCh38

View Full Product Details