Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 10462427-10462950 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DFFA
Entrez Gene ID
1676
Gene Name
DNA fragmentation factor subunit alpha
Gene Aliases
DFF-45, DFF1, ICAD
UniGene
Hs.484782
Gene Chromosome Location
Chr.1: 10460531-10472556 on Build GRCh38
Transcripts
NM_004401.2 NM_213566.1

SNP Detail

SNP ID Chromosome Location
rs11121573 Chr.1: 10462716-10462716 on Build GRCh38
rs894068820 Chr.1: 10462445-10462445 on Build GRCh38
rs927713950 Chr.1: 10462710-10462710 on Build GRCh38
rs935455432 Chr.1: 10462723-10462723 on Build GRCh38
rs553820785 Chr.1: 10462789-10462789 on Build GRCh38
rs993980865 Chr.1: 10462567-10462567 on Build GRCh38
rs1019017872 Chr.1: 10462544-10462544 on Build GRCh38
rs775333065 Chr.1: 10462820-10462820 on Build GRCh38
rs768671073 Chr.1: 10462867-10462867 on Build GRCh38
rs927966293 Chr.1: 10462747-10462747 on Build GRCh38
rs984483301 Chr.1: 10462625-10462625 on Build GRCh38
rs184768054 Chr.1: 10462631-10462631 on Build GRCh38
rs139956763 Chr.1: 10462505-10462505 on Build GRCh38
rs145882179 Chr.1: 10462682-10462682 on Build GRCh38
rs1051153021 Chr.1: 10462767-10462767 on Build GRCh38
rs946988095 Chr.1: 10462757-10462757 on Build GRCh38
rs140650992 Chr.1: 10462870-10462870 on Build GRCh38
rs897500286 Chr.1: 10462902-10462902 on Build GRCh38
rs915438828 Chr.1: 10462753-10462753 on Build GRCh38
rs993928494 Chr.1: 10462917-10462917 on Build GRCh38
rs368673670 Chr.1: 10462920-10462920 on Build GRCh38
rs952668348 Chr.1: 10462602-10462602 on Build GRCh38
rs1018798749 Chr.1: 10462672-10462672 on Build GRCh38
rs188176909 Chr.1: 10462514-10462514 on Build GRCh38
rs915379587 Chr.1: 10462783-10462783 on Build GRCh38
rs888887720 Chr.1: 10462830-10462830 on Build GRCh38
rs893984606 Chr.1: 10462520-10462520 on Build GRCh38
rs1011288709 Chr.1: 10462452-10462452 on Build GRCh38
rs77891640 Chr.1: 10462812-10462812 on Build GRCh38
rs1025488851 Chr.1: 10462590-10462590 on Build GRCh38
rs149855440 Chr.1: 10462675-10462675 on Build GRCh38
rs969488652 Chr.1: 10462673-10462673 on Build GRCh38
rs1050453877 Chr.1: 10462457-10462457 on Build GRCh38
rs1023291458 Chr.1: 10462629-10462629 on Build GRCh38
rs935574352 Chr.1: 10462750-10462750 on Build GRCh38
rs571534065 Chr.1: 10462701-10462701 on Build GRCh38
rs988313891 Chr.1: 10462735-10462735 on Build GRCh38
rs1008978081 Chr.1: 10462526-10462526 on Build GRCh38
rs538676013 Chr.1: 10462755-10462755 on Build GRCh38
rs776662341 Chr.1: 10462896-10462896 on Build GRCh38
rs571422836 Chr.1: 10462688-10462688 on Build GRCh38
rs951972608 Chr.1: 10462707-10462707 on Build GRCh38
rs1052587238 Chr.1: 10462512-10462512 on Build GRCh38
rs180710967 Chr.1: 10462564-10462564 on Build GRCh38
rs750406747 Chr.1: 10462794-10462794 on Build GRCh38
rs746959107 Chr.1: 10462852-10462852 on Build GRCh38

View Full Product Details