Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 66102790-66103245 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CTNNA3
Entrez Gene ID
29119
Gene Name
catenin alpha 3
Gene Aliases
ARVD13, VR22
UniGene
Hs.21375
Gene Chromosome Location
Chr.10: 65912518-67696217 on Build GRCh38
Transcripts
NM_001127384.2 NM_013266.3 XM_017016151.1 XM_017016152.1 XM_017016153.1 XM_017016154.1 XM_017016155.1 XM_017016156.1 XM_017016157.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
401055 c.1977G>T p.R659S Chr.10: 66103157-66103157 on Build GRCh38
2158755 c.1936C>A p.R646S Chr.10: 66103198-66103198 on Build GRCh38
1249402 c.1896A>G p.E632E Chr.10: 66103238-66103238 on Build GRCh38
6450278 c.1950C>A p.S650R Chr.10: 66103184-66103184 on Build GRCh38
919553 c.1893G>T p.E631D Chr.10: 66103241-66103241 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs766708884 Chr.10: 66103128-66103128 on Build GRCh38
rs556623128 Chr.10: 66103175-66103175 on Build GRCh38
rs768222263 Chr.10: 66103179-66103179 on Build GRCh38
rs186717320 Chr.10: 66103049-66103049 on Build GRCh38
rs998782164 Chr.10: 66103130-66103130 on Build GRCh38
rs757138676 Chr.10: 66103154-66103154 on Build GRCh38
rs948926578 Chr.10: 66102853-66102853 on Build GRCh38
rs1036020377 Chr.10: 66102907-66102907 on Build GRCh38
rs1048706213 Chr.10: 66102927-66102927 on Build GRCh38
rs954219089 Chr.10: 66103220-66103220 on Build GRCh38
rs373519359 Chr.10: 66103129-66103129 on Build GRCh38
rs376285665 Chr.10: 66102976-66102976 on Build GRCh38
rs141141746 Chr.10: 66103207-66103207 on Build GRCh38
rs780701411 Chr.10: 66103189-66103189 on Build GRCh38
rs750351645 Chr.10: 66103142-66103143 on Build GRCh38
rs1040077870 Chr.10: 66102822-66102822 on Build GRCh38
rs755284081 Chr.10: 66102904-66102904 on Build GRCh38
rs910159052 Chr.10: 66102898-66102898 on Build GRCh38
rs376927818 Chr.10: 66103195-66103195 on Build GRCh38
rs747672598 Chr.10: 66103194-66103194 on Build GRCh38
rs752753211 Chr.10: 66103151-66103151 on Build GRCh38
rs200764970 Chr.10: 66103166-66103166 on Build GRCh38
rs897592012 Chr.10: 66102852-66102852 on Build GRCh38
rs534574169 Chr.10: 66103076-66103076 on Build GRCh38
rs779211946 Chr.10: 66102925-66102925 on Build GRCh38
rs773649571 Chr.10: 66103118-66103118 on Build GRCh38
rs372121471 Chr.10: 66103089-66103089 on Build GRCh38
rs565978538 Chr.10: 66103069-66103069 on Build GRCh38
rs370269225 Chr.10: 66103197-66103197 on Build GRCh38
rs1013441188 Chr.10: 66103056-66103056 on Build GRCh38
rs770269331 Chr.10: 66102848-66102849 on Build GRCh38
rs370600208 Chr.10: 66103201-66103201 on Build GRCh38
rs546646934 Chr.10: 66103054-66103054 on Build GRCh38
rs574293198 Chr.10: 66103139-66103139 on Build GRCh38
rs551084909 Chr.10: 66102977-66102977 on Build GRCh38
rs147760277 Chr.10: 66103205-66103205 on Build GRCh38
rs778836534 Chr.10: 66103157-66103157 on Build GRCh38
rs916039278 Chr.10: 66102840-66102840 on Build GRCh38
rs749334816 Chr.10: 66102868-66102868 on Build GRCh38
rs758246563 Chr.10: 66103174-66103174 on Build GRCh38
rs148809676 Chr.10: 66103019-66103019 on Build GRCh38
rs111425421 Chr.10: 66103198-66103198 on Build GRCh38
rs1049107780 Chr.10: 66102865-66102865 on Build GRCh38
rs750151799 Chr.10: 66103172-66103172 on Build GRCh38
rs554754922 Chr.10: 66103127-66103127 on Build GRCh38
rs897503901 Chr.10: 66102923-66102923 on Build GRCh38
rs537371978 Chr.10: 66102945-66102945 on Build GRCh38
rs1034023747 Chr.10: 66103001-66103001 on Build GRCh38
rs941696762 Chr.10: 66102900-66102900 on Build GRCh38

View Full Product Details