Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 33205131-33205635 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HSD17B8
Entrez Gene ID
7923
Gene Name
hydroxysteroid 17-beta dehydrogenase 8
Gene Aliases
D6S2245E, FABG, FABGL, H2-KE6, HKE6, KE6, RING2, SDR30C1, dJ1033B10.9
UniGene
Hs.415058
Gene Chromosome Location
Chr.6: 33204637-33206831 on Build GRCh38
Transcripts
NM_014234.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5634013 c.416C>G p.A139G Chr.6: 33205475-33205475 on Build GRCh38
6682384 c.441T>C p.C147C Chr.6: 33205500-33205500 on Build GRCh38
5979178 c.440G>A p.C147Y Chr.6: 33205499-33205499 on Build GRCh38
32581 c.472G>T p.V158L Chr.6: 33205531-33205531 on Build GRCh38
421373 c.281C>G p.S94C Chr.6: 33205231-33205231 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs764290156 Chr.6: 33205576-33205576 on Build GRCh38
rs759065768 Chr.6: 33205248-33205248 on Build GRCh38
rs191965195 Chr.6: 33205218-33205218 on Build GRCh38
rs779093965 Chr.6: 33205215-33205215 on Build GRCh38
rs574238516 Chr.6: 33205165-33205165 on Build GRCh38
rs1011449385 Chr.6: 33205590-33205590 on Build GRCh38
rs773720520 Chr.6: 33205227-33205227 on Build GRCh38
rs750720359 Chr.6: 33205208-33205209 on Build GRCh38
rs764196969 Chr.6: 33205205-33205205 on Build GRCh38
rs756730982 Chr.6: 33205596-33205596 on Build GRCh38
rs377019063 Chr.6: 33205385-33205385 on Build GRCh38
rs776253757 Chr.6: 33205461-33205461 on Build GRCh38
rs142469266 Chr.6: 33205551-33205551 on Build GRCh38
rs774994670 Chr.6: 33205604-33205604 on Build GRCh38
rs984496894 Chr.6: 33205427-33205427 on Build GRCh38
rs746330387 Chr.6: 33205495-33205495 on Build GRCh38
rs779300617 Chr.6: 33205309-33205309 on Build GRCh38
rs769365208 Chr.6: 33205343-33205343 on Build GRCh38
rs150581613 Chr.6: 33205450-33205450 on Build GRCh38
rs778593327 Chr.6: 33205403-33205403 on Build GRCh38
rs138352449 Chr.6: 33205248-33205250 on Build GRCh38
rs751543629 Chr.6: 33205272-33205272 on Build GRCh38
rs745585312 Chr.6: 33205583-33205585 on Build GRCh38
rs779112327 Chr.6: 33205299-33205299 on Build GRCh38
rs750308032 Chr.6: 33205425-33205425 on Build GRCh38
rs755080187 Chr.6: 33205292-33205292 on Build GRCh38
rs755465645 Chr.6: 33205260-33205263 on Build GRCh38
rs751742249 Chr.6: 33205578-33205578 on Build GRCh38
rs919533134 Chr.6: 33205376-33205376 on Build GRCh38
rs781573716 Chr.6: 33205581-33205581 on Build GRCh38
rs201761806 Chr.6: 33205190-33205190 on Build GRCh38
rs550548519 Chr.6: 33205407-33205407 on Build GRCh38
rs141630682 Chr.6: 33205223-33205223 on Build GRCh38
rs903075140 Chr.6: 33205417-33205417 on Build GRCh38
rs756238441 Chr.6: 33205211-33205211 on Build GRCh38
rs776370953 Chr.6: 33205168-33205168 on Build GRCh38
rs752825641 Chr.6: 33205559-33205559 on Build GRCh38
rs749352910 Chr.6: 33205212-33205212 on Build GRCh38
rs757578803 Chr.6: 33205579-33205579 on Build GRCh38
rs749757690 Chr.6: 33205247-33205248 on Build GRCh38
rs781558343 Chr.6: 33205493-33205493 on Build GRCh38
rs111231091 Chr.6: 33205330-33205330 on Build GRCh38
rs757712746 Chr.6: 33205460-33205460 on Build GRCh38
rs768476738 Chr.6: 33205525-33205525 on Build GRCh38
rs748870542 Chr.6: 33205602-33205602 on Build GRCh38
rs776045109 Chr.6: 33205243-33205243 on Build GRCh38
rs769226572 Chr.6: 33205591-33205591 on Build GRCh38
rs767600140 Chr.6: 33205185-33205185 on Build GRCh38
rs200066054 Chr.6: 33205356-33205356 on Build GRCh38
rs893130078 Chr.6: 33205487-33205487 on Build GRCh38
rs775474408 Chr.6: 33205535-33205535 on Build GRCh38
rs372114440 Chr.6: 33205499-33205499 on Build GRCh38
rs188608072 Chr.6: 33205365-33205365 on Build GRCh38
rs764913157 Chr.6: 33205175-33205175 on Build GRCh38
rs759072636 Chr.6: 33205169-33205169 on Build GRCh38
rs110662 Chr.6: 33205155-33205155 on Build GRCh38
rs748546166 Chr.6: 33205324-33205324 on Build GRCh38
rs145122117 Chr.6: 33205342-33205342 on Build GRCh38
rs774045796 Chr.6: 33205532-33205532 on Build GRCh38
rs757640691 Chr.6: 33205480-33205480 on Build GRCh38
rs996842144 Chr.6: 33205491-33205491 on Build GRCh38
rs920242574 Chr.6: 33205224-33205224 on Build GRCh38
rs772537165 Chr.6: 33205430-33205431 on Build GRCh38
rs137920002 Chr.6: 33205361-33205361 on Build GRCh38
rs763008180 Chr.6: 33205567-33205567 on Build GRCh38
rs760563977 Chr.6: 33205556-33205556 on Build GRCh38
rs763927121 Chr.6: 33205271-33205271 on Build GRCh38
rs144457204 Chr.6: 33205261-33205261 on Build GRCh38
rs748430763 Chr.6: 33205340-33205341 on Build GRCh38
rs556499620 Chr.6: 33205262-33205262 on Build GRCh38
rs771610415 Chr.6: 33205502-33205502 on Build GRCh38
rs757456441 Chr.6: 33205184-33205185 on Build GRCh38
rs758662649 Chr.6: 33205459-33205459 on Build GRCh38
rs759623617 Chr.6: 33205428-33205428 on Build GRCh38
rs116067382 Chr.6: 33205600-33205600 on Build GRCh38
rs758783369 Chr.6: 33205320-33205320 on Build GRCh38
rs748309476 Chr.6: 33205312-33205312 on Build GRCh38
rs751477353 Chr.6: 33205208-33205208 on Build GRCh38
rs369676098 Chr.6: 33205433-33205433 on Build GRCh38
rs376588362 Chr.6: 33205275-33205275 on Build GRCh38
rs375398823 Chr.6: 33205588-33205588 on Build GRCh38
rs753920057 Chr.6: 33205284-33205284 on Build GRCh38
rs183728488 Chr.6: 33205333-33205333 on Build GRCh38
rs758257314 Chr.6: 33205191-33205191 on Build GRCh38
rs771842527 Chr.6: 33205550-33205550 on Build GRCh38
rs770507785 Chr.6: 33205233-33205233 on Build GRCh38
rs748782700 Chr.6: 33205504-33205504 on Build GRCh38
rs777950811 Chr.6: 33205461-33205461 on Build GRCh38
rs920210460 Chr.6: 33205594-33205594 on Build GRCh38
rs756246487 Chr.6: 33205218-33205219 on Build GRCh38
rs778398553 Chr.6: 33205515-33205517 on Build GRCh38
rs751927513 Chr.6: 33205470-33205470 on Build GRCh38
rs767462927 Chr.6: 33205283-33205283 on Build GRCh38

View Full Product Details