Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 220102161-220102703 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IARS2
Entrez Gene ID
55699
Gene Name
isoleucyl-tRNA synthetase 2, mitochondrial
Gene Aliases
CAGSSS, ILERS
UniGene
Hs.262823
Gene Chromosome Location
Chr.1: 220094113-220148041 on Build GRCh38
Transcripts
NM_018060.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5350049 c.602G>T p.R201L Chr.1: 220102180-220102180 on Build GRCh38
6683456 c.713G>A p.R238Q Chr.1: 220102376-220102376 on Build GRCh38
1338968 c.808T>C p.Y270H Chr.1: 220102553-220102553 on Build GRCh38
4953773 c.681T>G p.F227L Chr.1: 220102259-220102259 on Build GRCh38
1210208 c.785C>A p.P262H Chr.1: 220102530-220102530 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs756393373 Chr.1: 220102395-220102395 on Build GRCh38
rs201916678 Chr.1: 220102540-220102540 on Build GRCh38
rs746455426 Chr.1: 220102533-220102533 on Build GRCh38
rs377432411 Chr.1: 220102545-220102545 on Build GRCh38
rs767135644 Chr.1: 220102188-220102188 on Build GRCh38
rs774960759 Chr.1: 220102463-220102463 on Build GRCh38
rs924595832 Chr.1: 220102641-220102641 on Build GRCh38
rs745688807 Chr.1: 220102313-220102313 on Build GRCh38
rs1042026666 Chr.1: 220102226-220102226 on Build GRCh38
rs369155122 Chr.1: 220102364-220102364 on Build GRCh38
rs762202072 Chr.1: 220102465-220102465 on Build GRCh38
rs769112693 Chr.1: 220102553-220102553 on Build GRCh38
rs753250629 Chr.1: 220102257-220102257 on Build GRCh38
rs757319390 Chr.1: 220102370-220102370 on Build GRCh38
rs149353254 Chr.1: 220102216-220102216 on Build GRCh38
rs917298497 Chr.1: 220102544-220102544 on Build GRCh38
rs751095095 Chr.1: 220102512-220102512 on Build GRCh38
rs372618006 Chr.1: 220102429-220102429 on Build GRCh38
rs751587007 Chr.1: 220102271-220102271 on Build GRCh38
rs754889916 Chr.1: 220102409-220102409 on Build GRCh38
rs182226161 Chr.1: 220102352-220102352 on Build GRCh38
rs904984929 Chr.1: 220102455-220102455 on Build GRCh38
rs571001903 Chr.1: 220102288-220102289 on Build GRCh38
rs752366568 Chr.1: 220102669-220102669 on Build GRCh38
rs1032292776 Chr.1: 220102518-220102518 on Build GRCh38
rs372276818 Chr.1: 220102317-220102317 on Build GRCh38
rs201539020 Chr.1: 220102436-220102436 on Build GRCh38
rs546640845 Chr.1: 220102274-220102274 on Build GRCh38
rs751613884 Chr.1: 220102367-220102367 on Build GRCh38
rs766741951 Chr.1: 220102626-220102626 on Build GRCh38
rs777266427 Chr.1: 220102531-220102531 on Build GRCh38
rs754766431 Chr.1: 220102498-220102498 on Build GRCh38
rs539127735 Chr.1: 220102629-220102629 on Build GRCh38
rs764539319 Chr.1: 220102449-220102449 on Build GRCh38
rs141996928 Chr.1: 220102214-220102214 on Build GRCh38
rs199601961 Chr.1: 220102404-220102404 on Build GRCh38
rs767766570 Chr.1: 220102467-220102467 on Build GRCh38
rs199575981 Chr.1: 220102412-220102412 on Build GRCh38
rs189084759 Chr.1: 220102327-220102327 on Build GRCh38
rs758325882 Chr.1: 220102529-220102529 on Build GRCh38
rs777779272 Chr.1: 220102269-220102269 on Build GRCh38
rs373433981 Chr.1: 220102520-220102520 on Build GRCh38
rs374990412 Chr.1: 220102358-220102358 on Build GRCh38
rs770588599 Chr.1: 220102534-220102534 on Build GRCh38
rs976770107 Chr.1: 220102591-220102591 on Build GRCh38
rs775841640 Chr.1: 220102431-220102431 on Build GRCh38
rs368937943 Chr.1: 220102203-220102203 on Build GRCh38
rs773439017 Chr.1: 220102594-220102594 on Build GRCh38
rs202128341 Chr.1: 220102341-220102341 on Build GRCh38
rs775825237 Chr.1: 220102357-220102357 on Build GRCh38
rs958850547 Chr.1: 220102443-220102443 on Build GRCh38
rs749298044 Chr.1: 220102319-220102319 on Build GRCh38
rs766651661 Chr.1: 220102493-220102493 on Build GRCh38
rs767672750 Chr.1: 220102376-220102376 on Build GRCh38
rs759657956 Chr.1: 220102637-220102637 on Build GRCh38
rs756410015 Chr.1: 220102490-220102490 on Build GRCh38
rs762450476 Chr.1: 220102592-220102592 on Build GRCh38
rs774564592 Chr.1: 220102566-220102566 on Build GRCh38
rs376610134 Chr.1: 220102318-220102318 on Build GRCh38
rs370306595 Chr.1: 220102452-220102452 on Build GRCh38
rs903078644 Chr.1: 220102236-220102236 on Build GRCh38
rs988091643 Chr.1: 220102232-220102232 on Build GRCh38
rs187251505 Chr.1: 220102355-220102355 on Build GRCh38
rs760791870 Chr.1: 220102602-220102602 on Build GRCh38
rs776306373 Chr.1: 220102537-220102537 on Build GRCh38
rs778544749 Chr.1: 220102507-220102507 on Build GRCh38
rs761439693 Chr.1: 220102187-220102187 on Build GRCh38
rs771648093 Chr.1: 220102348-220102348 on Build GRCh38
rs773738046 Chr.1: 220102332-220102332 on Build GRCh38
rs772640835 Chr.1: 220102593-220102593 on Build GRCh38
rs914016212 Chr.1: 220102242-220102242 on Build GRCh38
rs750528193 Chr.1: 220102478-220102478 on Build GRCh38
rs781426318 Chr.1: 220102294-220102294 on Build GRCh38
rs1054248277 Chr.1: 220102312-220102312 on Build GRCh38
rs758922128 Chr.1: 220102267-220102267 on Build GRCh38
rs1028913458 Chr.1: 220102609-220102609 on Build GRCh38
rs1009545961 Chr.1: 220102536-220102536 on Build GRCh38
rs772692964 Chr.1: 220102423-220102423 on Build GRCh38
rs747645857 Chr.1: 220102420-220102420 on Build GRCh38
rs749790428 Chr.1: 220102298-220102299 on Build GRCh38
rs374904870 Chr.1: 220102430-220102430 on Build GRCh38
rs370222815 Chr.1: 220102438-220102438 on Build GRCh38
rs533806944 Chr.1: 220102657-220102657 on Build GRCh38
rs753601921 Chr.1: 220102497-220102497 on Build GRCh38
rs868785797 Chr.1: 220102375-220102375 on Build GRCh38
rs917287197 Chr.1: 220102190-220102190 on Build GRCh38
rs750724636 Chr.1: 220102390-220102390 on Build GRCh38
rs780062054 Chr.1: 220102403-220102403 on Build GRCh38

View Full Product Details