Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 80197387-80197955 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DIO2
Entrez Gene ID
1734
Gene Name
deiodinase, iodothyronine, type II
Gene Aliases
5DII, D2, DIOII, SelY, TXDI2
UniGene
Hs.202354
Gene Chromosome Location
Chr.14: 80197525-80231054 on Build GRCh38
Transcripts
NM_000793.5 NM_001007023.4 NM_001242502.2 NM_001242503.2 NM_001324462.1 NM_013989.4

SNP Detail

SNP ID Chromosome Location
rs552954648 Chr.14: 80197685-80197685 on Build GRCh38
rs1033263051 Chr.14: 80197894-80197894 on Build GRCh38
rs890165173 Chr.14: 80197875-80197875 on Build GRCh38
rs950323004 Chr.14: 80197855-80197855 on Build GRCh38
rs916140468 Chr.14: 80197594-80197594 on Build GRCh38
rs747944845 Chr.14: 80197684-80197684 on Build GRCh38
rs557547315 Chr.14: 80197462-80197462 on Build GRCh38
rs1020074788 Chr.14: 80197883-80197883 on Build GRCh38
rs1054432340 Chr.14: 80197718-80197718 on Build GRCh38
rs967133975 Chr.14: 80197884-80197884 on Build GRCh38
rs984575689 Chr.14: 80197476-80197476 on Build GRCh38
rs1046002625 Chr.14: 80197876-80197876 on Build GRCh38
rs967227529 Chr.14: 80197542-80197542 on Build GRCh38
rs916093535 Chr.14: 80197837-80197837 on Build GRCh38
rs754600710 Chr.14: 80197611-80197611 on Build GRCh38
rs568962031 Chr.14: 80197888-80197888 on Build GRCh38
rs386779204 Chr.14: 80197680-80197681 on Build GRCh38
rs541713349 Chr.14: 80197730-80197730 on Build GRCh38
rs950437654 Chr.14: 80197440-80197440 on Build GRCh38
rs533344208 Chr.14: 80197745-80197745 on Build GRCh38
rs879282333 Chr.14: 80197836-80197836 on Build GRCh38
rs975754555 Chr.14: 80197409-80197409 on Build GRCh38
rs780994629 Chr.14: 80197639-80197639 on Build GRCh38
rs777823251 Chr.14: 80197794-80197794 on Build GRCh38
rs548968884 Chr.14: 80197820-80197820 on Build GRCh38
rs572801391 Chr.14: 80197702-80197702 on Build GRCh38
rs908931955 Chr.14: 80197819-80197819 on Build GRCh38
rs192334437 Chr.14: 80197593-80197593 on Build GRCh38
rs965975246 Chr.14: 80197743-80197743 on Build GRCh38
rs564738665 Chr.14: 80197739-80197739 on Build GRCh38
rs997874577 Chr.14: 80197754-80197754 on Build GRCh38
rs184719105 Chr.14: 80197880-80197880 on Build GRCh38
rs1025980349 Chr.14: 80197788-80197788 on Build GRCh38
rs12891403 Chr.14: 80197508-80197508 on Build GRCh38
rs1056813 Chr.14: 80197776-80197776 on Build GRCh38
rs1005654654 Chr.14: 80197414-80197414 on Build GRCh38
rs1056815 Chr.14: 80197713-80197713 on Build GRCh38
rs1056817 Chr.14: 80197659-80197659 on Build GRCh38
rs925781573 Chr.14: 80197603-80197605 on Build GRCh38
rs1056379866 Chr.14: 80197561-80197561 on Build GRCh38
rs563622669 Chr.14: 80197806-80197806 on Build GRCh38
rs565434788 Chr.14: 80197798-80197798 on Build GRCh38
rs113059592 Chr.14: 80197682-80197682 on Build GRCh38
rs537176855 Chr.14: 80197552-80197552 on Build GRCh38
rs918953404 Chr.14: 80197428-80197428 on Build GRCh38
rs1029078245 Chr.14: 80197468-80197468 on Build GRCh38
rs964656375 Chr.14: 80197437-80197437 on Build GRCh38
rs144870997 Chr.14: 80197805-80197805 on Build GRCh38
rs770799855 Chr.14: 80197879-80197879 on Build GRCh38
rs941423668 Chr.14: 80197779-80197779 on Build GRCh38
rs769761484 Chr.14: 80197723-80197723 on Build GRCh38
rs1007365916 Chr.14: 80197734-80197734 on Build GRCh38
rs149059509 Chr.14: 80197808-80197808 on Build GRCh38
rs941474876 Chr.14: 80197524-80197524 on Build GRCh38
rs893231592 Chr.14: 80197732-80197732 on Build GRCh38
rs76977648 Chr.14: 80197485-80197485 on Build GRCh38
rs954326662 Chr.14: 80197484-80197484 on Build GRCh38
rs1020043654 Chr.14: 80197512-80197512 on Build GRCh38
rs542651371 Chr.14: 80197667-80197667 on Build GRCh38
rs1017213039 Chr.14: 80197434-80197434 on Build GRCh38
rs796352810 Chr.14: 80197419-80197419 on Build GRCh38

View Full Product Details