Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 145804854-145805452 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FBXO30
Entrez Gene ID
84085
Gene Name
F-box protein 30
Gene Aliases
Fbx30
UniGene
Hs.421095
Gene Chromosome Location
Chr.6: 145798135-145864409 on Build GRCh38
Transcripts
NM_032145.4 XM_005267159.3 XM_011536177.2 XM_017011353.1 NR_038246.1

Gene Symbol
LOC100507557
Entrez Gene ID
100507557
Gene Name
uncharacterized LOC100507557
Gene Aliases
-
UniGene
Hs.124537
Gene Chromosome Location
Chr.6: 145734869-145886585 on Build GRCh38
Transcripts
NM_032145.4 XM_005267159.3 XM_011536177.2 XM_017011353.1 NR_038246.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6662165 c.1525G>A p.A509T Chr.6: 145804881-145804881 on Build GRCh38
3734962 c.1544A>T p.Q515L Chr.6: 145804862-145804862 on Build GRCh38
5869468 c.1481G>A p.S494N Chr.6: 145804925-145804925 on Build GRCh38
3928137 c.1194T>C p.S398S Chr.6: 145805212-145805212 on Build GRCh38
1206824 c.1282G>A p.A428T Chr.6: 145805124-145805124 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs779882606 Chr.6: 145805363-145805363 on Build GRCh38
rs745784772 Chr.6: 145805341-145805341 on Build GRCh38
rs144190690 Chr.6: 145805237-145805237 on Build GRCh38
rs141023654 Chr.6: 145805419-145805419 on Build GRCh38
rs746247911 Chr.6: 145804978-145804978 on Build GRCh38
rs777896008 Chr.6: 145805333-145805333 on Build GRCh38
rs767896439 Chr.6: 145805310-145805310 on Build GRCh38
rs374826187 Chr.6: 145805066-145805066 on Build GRCh38
rs768826721 Chr.6: 145804989-145804989 on Build GRCh38
rs377254337 Chr.6: 145805109-145805109 on Build GRCh38
rs749193729 Chr.6: 145805100-145805100 on Build GRCh38
rs769732095 Chr.6: 145805000-145805000 on Build GRCh38
rs9373475 Chr.6: 145805283-145805283 on Build GRCh38
rs147206535 Chr.6: 145805154-145805154 on Build GRCh38
rs141905579 Chr.6: 145804993-145804993 on Build GRCh38
rs766907294 Chr.6: 145804937-145804937 on Build GRCh38
rs964307129 Chr.6: 145805033-145805033 on Build GRCh38
rs757311577 Chr.6: 145805336-145805336 on Build GRCh38
rs762762551 Chr.6: 145805013-145805013 on Build GRCh38
rs200906244 Chr.6: 145805057-145805057 on Build GRCh38
rs769311176 Chr.6: 145805398-145805398 on Build GRCh38
rs765724282 Chr.6: 145805325-145805330 on Build GRCh38
rs143078736 Chr.6: 145805378-145805378 on Build GRCh38
rs754549761 Chr.6: 145804931-145804931 on Build GRCh38
rs759299720 Chr.6: 145805121-145805121 on Build GRCh38
rs202071659 Chr.6: 145804881-145804881 on Build GRCh38
rs150961850 Chr.6: 145805297-145805297 on Build GRCh38
rs1029594513 Chr.6: 145805046-145805046 on Build GRCh38
rs993458104 Chr.6: 145805198-145805198 on Build GRCh38
rs777720532 Chr.6: 145805411-145805411 on Build GRCh38
rs766830976 Chr.6: 145805156-145805156 on Build GRCh38
rs762186782 Chr.6: 145805143-145805143 on Build GRCh38
rs765070363 Chr.6: 145804905-145804905 on Build GRCh38
rs752054093 Chr.6: 145805113-145805113 on Build GRCh38
rs150645956 Chr.6: 145805027-145805027 on Build GRCh38
rs779495298 Chr.6: 145804972-145804972 on Build GRCh38
rs758785938 Chr.6: 145804984-145804984 on Build GRCh38
rs757691576 Chr.6: 145804968-145804968 on Build GRCh38
rs370994230 Chr.6: 145804995-145804995 on Build GRCh38
rs754018447 Chr.6: 145805427-145805427 on Build GRCh38
rs563279700 Chr.6: 145805410-145805410 on Build GRCh38
rs776272889 Chr.6: 145805293-145805293 on Build GRCh38
rs759090831 Chr.6: 145805039-145805039 on Build GRCh38
rs766036145 Chr.6: 145804922-145804922 on Build GRCh38
rs769371575 Chr.6: 145805007-145805007 on Build GRCh38
rs780452906 Chr.6: 145804985-145804985 on Build GRCh38
rs776361732 Chr.6: 145805386-145805386 on Build GRCh38
rs903725222 Chr.6: 145804952-145804952 on Build GRCh38
rs974315287 Chr.6: 145805090-145805090 on Build GRCh38
rs762672729 Chr.6: 145804921-145804921 on Build GRCh38
rs760291062 Chr.6: 145805150-145805150 on Build GRCh38
rs771860707 Chr.6: 145805272-145805272 on Build GRCh38
rs926888924 Chr.6: 145805139-145805139 on Build GRCh38
rs753921063 Chr.6: 145805334-145805334 on Build GRCh38
rs748193584 Chr.6: 145805081-145805081 on Build GRCh38
rs775043012 Chr.6: 145805274-145805274 on Build GRCh38
rs762439175 Chr.6: 145805035-145805035 on Build GRCh38
rs752144204 Chr.6: 145805041-145805041 on Build GRCh38
rs781296462 Chr.6: 145805168-145805168 on Build GRCh38
rs149933372 Chr.6: 145805146-145805146 on Build GRCh38
rs760378775 Chr.6: 145805279-145805279 on Build GRCh38
rs771635809 Chr.6: 145805128-145805128 on Build GRCh38
rs773080045 Chr.6: 145805004-145805004 on Build GRCh38
rs774003792 Chr.6: 145805119-145805119 on Build GRCh38
rs755315370 Chr.6: 145805167-145805167 on Build GRCh38
rs770850397 Chr.6: 145805256-145805256 on Build GRCh38
rs778747166 Chr.6: 145805340-145805340 on Build GRCh38
rs759270409 Chr.6: 145804882-145804882 on Build GRCh38
rs771811128 Chr.6: 145805344-145805344 on Build GRCh38
rs3811103 Chr.6: 145805418-145805418 on Build GRCh38
rs766813107 Chr.6: 145805299-145805299 on Build GRCh38
rs751870347 Chr.6: 145805158-145805158 on Build GRCh38
rs756229457 Chr.6: 145805183-145805183 on Build GRCh38
rs759852038 Chr.6: 145805304-145805304 on Build GRCh38
rs774762944 Chr.6: 145805406-145805406 on Build GRCh38
rs747686549 Chr.6: 145805390-145805390 on Build GRCh38
rs917524677 Chr.6: 145805199-145805199 on Build GRCh38
rs757320923 Chr.6: 145805431-145805431 on Build GRCh38
rs756317455 Chr.6: 145805329-145805329 on Build GRCh38
rs768423485 Chr.6: 145805367-145805367 on Build GRCh38
rs147780756 Chr.6: 145805258-145805258 on Build GRCh38
rs140656607 Chr.6: 145805155-145805155 on Build GRCh38
rs765866118 Chr.6: 145805054-145805054 on Build GRCh38
rs367842440 Chr.6: 145805072-145805072 on Build GRCh38
rs982940203 Chr.6: 145805249-145805249 on Build GRCh38
rs752966838 Chr.6: 145805317-145805317 on Build GRCh38
rs755050488 Chr.6: 145805348-145805348 on Build GRCh38
rs139127963 Chr.6: 145805045-145805045 on Build GRCh38
rs200343922 Chr.6: 145805174-145805174 on Build GRCh38
rs777809508 Chr.6: 145805191-145805191 on Build GRCh38
rs769819787 Chr.6: 145805094-145805094 on Build GRCh38
rs747165499 Chr.6: 145804988-145804988 on Build GRCh38
rs767113391 Chr.6: 145805040-145805040 on Build GRCh38
rs1042900776 Chr.6: 145805197-145805197 on Build GRCh38
rs926807830 Chr.6: 145805421-145805421 on Build GRCh38
rs371783420 Chr.6: 145805063-145805063 on Build GRCh38
rs567622272 Chr.6: 145804876-145804876 on Build GRCh38
rs750576313 Chr.6: 145805190-145805190 on Build GRCh38
rs35603594 Chr.6: 145805431-145805432 on Build GRCh38
rs368211050 Chr.6: 145805366-145805366 on Build GRCh38
rs772992095 Chr.6: 145804915-145804915 on Build GRCh38
rs760083960 Chr.6: 145805051-145805051 on Build GRCh38
rs755146602 Chr.6: 145805067-145805067 on Build GRCh38
rs17075385 Chr.6: 145805261-145805261 on Build GRCh38
rs778843908 Chr.6: 145805257-145805257 on Build GRCh38
rs549794776 Chr.6: 145804945-145804945 on Build GRCh38
rs372576394 Chr.6: 145805096-145805096 on Build GRCh38
rs892412739 Chr.6: 145805372-145805372 on Build GRCh38
rs780307520 Chr.6: 145805263-145805263 on Build GRCh38
rs752060783 Chr.6: 145805301-145805301 on Build GRCh38

View Full Product Details