Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 6502384-6502977 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ANGPT2
Entrez Gene ID
285
Gene Name
angiopoietin 2
Gene Aliases
AGPT2, ANG2
UniGene
Hs.583870
Gene Chromosome Location
Chr.8: 6499651-6563420 on Build GRCh38
Transcripts
NM_001118887.1 NM_001118888.1 NM_001147.2 XM_017013318.1 NM_001322042.1 NM_024596.4 XM_011534755.2 XM_011534756.2 XM_011534757.2 XM_011534758.2 XM_011534759.2 XM_011534760.2 XM_017013829.1 XM_017013830.1 XM_017013831.1 XM_017013832.1 XM_017013833.1 XM_017013834.1

Gene Symbol
MCPH1
Entrez Gene ID
79648
Gene Name
microcephalin 1
Gene Aliases
BRIT1, MCT
UniGene
Hs.593807
Gene Chromosome Location
Chr.8: 6406592-6648505 on Build GRCh38
Transcripts
NM_001118887.1 NM_001118888.1 NM_001147.2 XM_017013318.1 NM_001322042.1 NM_024596.4 XM_011534755.2 XM_011534756.2 XM_011534757.2 XM_011534758.2 XM_011534759.2 XM_011534760.2 XM_017013829.1 XM_017013830.1 XM_017013831.1 XM_017013832.1 XM_017013833.1 XM_017013834.1

SNP Detail

SNP ID Chromosome Location
rs528800338 Chr.8: 6502650-6502650 on Build GRCh38
rs555070227 Chr.8: 6502860-6502860 on Build GRCh38
rs542673330 Chr.8: 6502623-6502623 on Build GRCh38
rs886792562 Chr.8: 6502687-6502687 on Build GRCh38
rs1048125192 Chr.8: 6502790-6502790 on Build GRCh38
rs557677761 Chr.8: 6502902-6502902 on Build GRCh38
rs1032417113 Chr.8: 6502901-6502901 on Build GRCh38
rs1015175920 Chr.8: 6502827-6502827 on Build GRCh38
rs949294761 Chr.8: 6502923-6502923 on Build GRCh38
rs528636943 Chr.8: 6502849-6502849 on Build GRCh38
rs1017067161 Chr.8: 6502535-6502535 on Build GRCh38
rs1055309335 Chr.8: 6502451-6502451 on Build GRCh38
rs982001167 Chr.8: 6502941-6502941 on Build GRCh38
rs7006660 Chr.8: 6502447-6502447 on Build GRCh38
rs564026405 Chr.8: 6502845-6502845 on Build GRCh38
rs953602567 Chr.8: 6502712-6502712 on Build GRCh38
rs556137797 Chr.8: 6502518-6502518 on Build GRCh38
rs375469076 Chr.8: 6502885-6502885 on Build GRCh38
rs975203844 Chr.8: 6502538-6502538 on Build GRCh38
rs1004252836 Chr.8: 6502711-6502711 on Build GRCh38
rs923357014 Chr.8: 6502430-6502430 on Build GRCh38
rs936605554 Chr.8: 6502450-6502450 on Build GRCh38
rs962545070 Chr.8: 6502536-6502536 on Build GRCh38
rs531154723 Chr.8: 6502690-6502690 on Build GRCh38
rs925323415 Chr.8: 6502620-6502620 on Build GRCh38
rs534595782 Chr.8: 6502897-6502897 on Build GRCh38
rs750442962 Chr.8: 6502433-6502433 on Build GRCh38
rs56075253 Chr.8: 6502606-6502606 on Build GRCh38
rs760954034 Chr.8: 6502887-6502887 on Build GRCh38
rs543507839 Chr.8: 6502481-6502481 on Build GRCh38
rs949242887 Chr.8: 6502757-6502757 on Build GRCh38
rs1030424041 Chr.8: 6502685-6502685 on Build GRCh38
rs558822563 Chr.8: 6502816-6502816 on Build GRCh38
rs1048422916 Chr.8: 6502686-6502686 on Build GRCh38
rs1053977 Chr.8: 6502643-6502643 on Build GRCh38
rs986211113 Chr.8: 6502717-6502717 on Build GRCh38
rs997695939 Chr.8: 6502859-6502859 on Build GRCh38
rs932046798 Chr.8: 6502432-6502432 on Build GRCh38
rs904944709 Chr.8: 6502580-6502580 on Build GRCh38
rs758276021 Chr.8: 6502449-6502449 on Build GRCh38
rs1028157784 Chr.8: 6502564-6502564 on Build GRCh38
rs958315872 Chr.8: 6502912-6502912 on Build GRCh38
rs896783743 Chr.8: 6502499-6502499 on Build GRCh38
rs1052335475 Chr.8: 6502498-6502498 on Build GRCh38
rs746318167 Chr.8: 6502916-6502916 on Build GRCh38
rs904996258 Chr.8: 6502784-6502784 on Build GRCh38
rs938128811 Chr.8: 6502835-6502835 on Build GRCh38
rs76020419 Chr.8: 6502768-6502768 on Build GRCh38
rs571863742 Chr.8: 6502667-6502667 on Build GRCh38
rs761518738 Chr.8: 6502555-6502555 on Build GRCh38
rs953556067 Chr.8: 6502881-6502881 on Build GRCh38
rs936670474 Chr.8: 6502740-6502740 on Build GRCh38
rs937776526 Chr.8: 6502787-6502787 on Build GRCh38
rs754589076 Chr.8: 6502694-6502694 on Build GRCh38
rs369776646 Chr.8: 6502414-6502414 on Build GRCh38
rs1053973 Chr.8: 6502665-6502665 on Build GRCh38
rs569665273 Chr.8: 6502749-6502749 on Build GRCh38
rs913925252 Chr.8: 6502587-6502587 on Build GRCh38
rs988140473 Chr.8: 6502914-6502914 on Build GRCh38
rs898091946 Chr.8: 6502844-6502844 on Build GRCh38
rs1001925457 Chr.8: 6502585-6502585 on Build GRCh38
rs1046217843 Chr.8: 6502565-6502565 on Build GRCh38
rs986702341 Chr.8: 6502581-6502581 on Build GRCh38
rs559971446 Chr.8: 6502628-6502628 on Build GRCh38
rs557539275 Chr.8: 6502442-6502442 on Build GRCh38
rs80324828 Chr.8: 6502955-6502955 on Build GRCh38
rs925281161 Chr.8: 6502818-6502818 on Build GRCh38
rs1053978 Chr.8: 6502642-6502642 on Build GRCh38
rs1013414994 Chr.8: 6502527-6502527 on Build GRCh38
rs1053976 Chr.8: 6502653-6502653 on Build GRCh38
rs925302107 Chr.8: 6502719-6502719 on Build GRCh38
rs990847847 Chr.8: 6502848-6502848 on Build GRCh38
rs1024004684 Chr.8: 6502522-6502522 on Build GRCh38
rs986751893 Chr.8: 6502788-6502788 on Build GRCh38
rs955307816 Chr.8: 6502570-6502570 on Build GRCh38
rs142453549 Chr.8: 6502698-6502698 on Build GRCh38
rs996591002 Chr.8: 6502755-6502755 on Build GRCh38
rs966741746 Chr.8: 6502805-6502805 on Build GRCh38
rs549830317 Chr.8: 6502739-6502739 on Build GRCh38
rs748656591 Chr.8: 6502551-6502577 on Build GRCh38
rs185050796 Chr.8: 6502658-6502658 on Build GRCh38
rs577395197 Chr.8: 6502915-6502915 on Build GRCh38
rs373673677 Chr.8: 6502883-6502883 on Build GRCh38
rs988088180 Chr.8: 6502742-6502742 on Build GRCh38
rs949663232 Chr.8: 6502679-6502679 on Build GRCh38
rs888144344 Chr.8: 6502796-6502796 on Build GRCh38
rs976171508 Chr.8: 6502420-6502420 on Build GRCh38
rs536883872 Chr.8: 6502950-6502950 on Build GRCh38
rs888095074 Chr.8: 6502617-6502617 on Build GRCh38
rs776327652 Chr.8: 6502532-6502532 on Build GRCh38

View Full Product Details