Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 232768066-232768615 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GIGYF2
Entrez Gene ID
26058
Gene Name
GRB10 interacting GYF protein 2
Gene Aliases
GYF2, PARK11, PERQ2, PERQ3, TNRC15
UniGene
Hs.565319
Gene Chromosome Location
Chr.2: 232697305-232860577 on Build GRCh38
Transcripts
NM_001103146.1 NM_001103147.1 NM_001103148.1 NM_015575.3 NR_103492.1 NM_001172416.1 NM_001172417.1 NM_002242.4

Gene Symbol
KCNJ13
Entrez Gene ID
3769
Gene Name
potassium voltage-gated channel subfamily J member 13
Gene Aliases
KIR1.4, KIR7.1, LCA16, SVD
UniGene
Hs.467338
Gene Chromosome Location
Chr.2: 232765802-232776565 on Build GRCh38
Transcripts
NM_001103146.1 NM_001103147.1 NM_001103148.1 NM_015575.3 NR_103492.1 NM_001172416.1 NM_001172417.1 NM_002242.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6691128 c.913C>T p.R305* Chr.2: 232768361-232768361 on Build GRCh38
3991143 c.756T>A p.P252P Chr.2: 232768518-232768518 on Build GRCh38
4092419 c.811C>A p.L271I Chr.2: 232768463-232768463 on Build GRCh38
4133550 c.930A>G p.E310E Chr.2: 232768344-232768344 on Build GRCh38
3972086 c.844T>C p.C282R Chr.2: 232768430-232768430 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs370613677 Chr.2: 232768436-232768436 on Build GRCh38
rs759022075 Chr.2: 232768161-232768161 on Build GRCh38
rs757634383 Chr.2: 232768274-232768274 on Build GRCh38
rs17853727 Chr.2: 232768405-232768405 on Build GRCh38
rs570138880 Chr.2: 232768097-232768097 on Build GRCh38
rs1051985295 Chr.2: 232768500-232768500 on Build GRCh38
rs750953704 Chr.2: 232768582-232768582 on Build GRCh38
rs555860663 Chr.2: 232768429-232768429 on Build GRCh38
rs982027064 Chr.2: 232768563-232768563 on Build GRCh38
rs765534142 Chr.2: 232768253-232768253 on Build GRCh38
rs768232798 Chr.2: 232768466-232768466 on Build GRCh38
rs778392451 Chr.2: 232768168-232768168 on Build GRCh38
rs766235801 Chr.2: 232768377-232768377 on Build GRCh38
rs763207758 Chr.2: 232768388-232768388 on Build GRCh38
rs139746604 Chr.2: 232768294-232768294 on Build GRCh38
rs570363677 Chr.2: 232768242-232768242 on Build GRCh38
rs750120231 Chr.2: 232768233-232768233 on Build GRCh38
rs374411396 Chr.2: 232768447-232768447 on Build GRCh38
rs888593629 Chr.2: 232768156-232768156 on Build GRCh38
rs748224832 Chr.2: 232768159-232768159 on Build GRCh38
rs201555149 Chr.2: 232768529-232768529 on Build GRCh38
rs760515492 Chr.2: 232768375-232768375 on Build GRCh38
rs757048919 Chr.2: 232768142-232768142 on Build GRCh38
rs377146231 Chr.2: 232768376-232768376 on Build GRCh38
rs929557852 Chr.2: 232768472-232768472 on Build GRCh38
rs370182405 Chr.2: 232768360-232768360 on Build GRCh38
rs576752433 Chr.2: 232768195-232768195 on Build GRCh38
rs760640227 Chr.2: 232768505-232768505 on Build GRCh38
rs375157738 Chr.2: 232768453-232768453 on Build GRCh38
rs373596360 Chr.2: 232768146-232768146 on Build GRCh38
rs758581367 Chr.2: 232768263-232768263 on Build GRCh38
rs980892735 Chr.2: 232768368-232768368 on Build GRCh38
rs1048157859 Chr.2: 232768123-232768123 on Build GRCh38
rs747432883 Chr.2: 232768171-232768171 on Build GRCh38
rs771346059 Chr.2: 232768174-232768174 on Build GRCh38
rs528587960 Chr.2: 232768548-232768548 on Build GRCh38
rs747562741 Chr.2: 232768269-232768269 on Build GRCh38
rs761770948 Chr.2: 232768577-232768577 on Build GRCh38
rs545594184 Chr.2: 232768231-232768231 on Build GRCh38
rs539013622 Chr.2: 232768404-232768404 on Build GRCh38
rs957867680 Chr.2: 232768440-232768440 on Build GRCh38
rs764204353 Chr.2: 232768562-232768562 on Build GRCh38
rs748955297 Chr.2: 232768465-232768465 on Build GRCh38
rs773108284 Chr.2: 232768373-232768373 on Build GRCh38
rs942173837 Chr.2: 232768105-232768105 on Build GRCh38
rs762432264 Chr.2: 232768209-232768209 on Build GRCh38
rs769488519 Chr.2: 232768305-232768305 on Build GRCh38
rs769427290 Chr.2: 232768189-232768189 on Build GRCh38
rs752863162 Chr.2: 232768260-232768260 on Build GRCh38
rs779652772 Chr.2: 232768308-232768308 on Build GRCh38
rs767179140 Chr.2: 232768144-232768144 on Build GRCh38
rs746208996 Chr.2: 232768298-232768298 on Build GRCh38
rs755099409 Chr.2: 232768154-232768154 on Build GRCh38
rs377736321 Chr.2: 232768344-232768344 on Build GRCh38
rs147191772 Chr.2: 232768467-232768467 on Build GRCh38
rs368221882 Chr.2: 232768471-232768471 on Build GRCh38
rs770861822 Chr.2: 232768544-232768544 on Build GRCh38
rs777663064 Chr.2: 232768267-232768267 on Build GRCh38
rs773815103 Chr.2: 232768489-232768489 on Build GRCh38
rs374301010 Chr.2: 232768184-232768184 on Build GRCh38
rs779138733 Chr.2: 232768157-232768157 on Build GRCh38
rs954056272 Chr.2: 232768369-232768369 on Build GRCh38
rs535054686 Chr.2: 232768198-232768198 on Build GRCh38
rs748902194 Chr.2: 232768313-232768313 on Build GRCh38
rs771899391 Chr.2: 232768370-232768370 on Build GRCh38
rs1004804255 Chr.2: 232768173-232768173 on Build GRCh38
rs772086040 Chr.2: 232768493-232768493 on Build GRCh38
rs761231820 Chr.2: 232768542-232768542 on Build GRCh38
rs926688430 Chr.2: 232768372-232768372 on Build GRCh38
rs750831125 Chr.2: 232768446-232768446 on Build GRCh38
rs376837580 Chr.2: 232768153-232768153 on Build GRCh38
rs17857137 Chr.2: 232768349-232768349 on Build GRCh38
rs956131677 Chr.2: 232768326-232768326 on Build GRCh38
rs774380473 Chr.2: 232768218-232768218 on Build GRCh38
rs143607153 Chr.2: 232768552-232768552 on Build GRCh38
rs374945617 Chr.2: 232768367-232768367 on Build GRCh38
rs369373214 Chr.2: 232768143-232768143 on Build GRCh38
rs764061215 Chr.2: 232768210-232768210 on Build GRCh38
rs781665941 Chr.2: 232768276-232768276 on Build GRCh38
rs146797648 Chr.2: 232768585-232768585 on Build GRCh38
rs776500156 Chr.2: 232768549-232768549 on Build GRCh38
rs566257347 Chr.2: 232768180-232768180 on Build GRCh38
rs752987924 Chr.2: 232768382-232768382 on Build GRCh38
rs751713849 Chr.2: 232768569-232768569 on Build GRCh38
rs370090481 Chr.2: 232768177-232768177 on Build GRCh38
rs374463684 Chr.2: 232768578-232768578 on Build GRCh38
rs371946844 Chr.2: 232768496-232768496 on Build GRCh38

View Full Product Details