Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 154349269-154349849 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FLNA
Entrez Gene ID
2316
Gene Name
filamin A
Gene Aliases
ABP-280, ABPX, CSBS, CVD1, FLN, FLN-A, FLN1, FMD, MNS, NHBP, OPD, OPD1, OPD2, XLVD, XMVD
UniGene
Hs.195464
Gene Chromosome Location
Chr.X: 154348529-154374638 on Build GRCh38
Transcripts
NM_001110556.1 NM_001456.3 XM_011531127.1 XM_011531128.1 XM_011531129.1 XM_011531130.1 XM_011531131.1

SNP Detail

SNP ID Chromosome Location
rs886039104 Chr.X: 154349514-154349514 on Build GRCh38
rs200694807 Chr.X: 154349522-154349522 on Build GRCh38
rs2070815 Chr.X: 154349441-154349441 on Build GRCh38
rs782784577 Chr.X: 154349450-154349450 on Build GRCh38
rs782291033 Chr.X: 154349404-154349404 on Build GRCh38
rs34328289 Chr.X: 154349326-154349327 on Build GRCh38
rs782502145 Chr.X: 154349689-154349689 on Build GRCh38
rs781941043 Chr.X: 154349584-154349584 on Build GRCh38
rs782594128 Chr.X: 154349355-154349355 on Build GRCh38
rs781867343 Chr.X: 154349343-154349343 on Build GRCh38
rs782338659 Chr.X: 154349750-154349750 on Build GRCh38
rs782190081 Chr.X: 154349748-154349748 on Build GRCh38
rs886039148 Chr.X: 154349510-154349510 on Build GRCh38
rs111243479 Chr.X: 154349360-154349360 on Build GRCh38
rs782263210 Chr.X: 154349504-154349504 on Build GRCh38
rs781909155 Chr.X: 154349656-154349656 on Build GRCh38
rs781924831 Chr.X: 154349778-154349779 on Build GRCh38
rs375433852 Chr.X: 154349609-154349609 on Build GRCh38
rs782091553 Chr.X: 154349828-154349828 on Build GRCh38
rs373103712 Chr.X: 154349812-154349812 on Build GRCh38
rs76337075 Chr.X: 154349432-154349432 on Build GRCh38
rs878854464 Chr.X: 154349807-154349807 on Build GRCh38
rs112682871 Chr.X: 154349583-154349583 on Build GRCh38
rs187774579 Chr.X: 154349554-154349554 on Build GRCh38
rs782156724 Chr.X: 154349575-154349575 on Build GRCh38
rs909387820 Chr.X: 154349738-154349738 on Build GRCh38
rs368529342 Chr.X: 154349314-154349314 on Build GRCh38
rs201883808 Chr.X: 154349534-154349534 on Build GRCh38
rs886039087 Chr.X: 154349800-154349800 on Build GRCh38
rs782535006 Chr.X: 154349345-154349345 on Build GRCh38
rs148571693 Chr.X: 154349613-154349613 on Build GRCh38
rs782747934 Chr.X: 154349830-154349830 on Build GRCh38
rs782442409 Chr.X: 154349657-154349657 on Build GRCh38
rs201663443 Chr.X: 154349354-154349354 on Build GRCh38
rs985402675 Chr.X: 154349713-154349713 on Build GRCh38
rs200195310 Chr.X: 154349695-154349695 on Build GRCh38
rs782308141 Chr.X: 154349405-154349405 on Build GRCh38
rs981689322 Chr.X: 154349692-154349692 on Build GRCh38
rs1022884327 Chr.X: 154349661-154349661 on Build GRCh38
rs782611801 Chr.X: 154349675-154349675 on Build GRCh38
rs886044883 Chr.X: 154349506-154349506 on Build GRCh38
rs781959191 Chr.X: 154349559-154349559 on Build GRCh38
rs781905890 Chr.X: 154349468-154349468 on Build GRCh38
rs74667788 Chr.X: 154349767-154349767 on Build GRCh38
rs368746337 Chr.X: 154349791-154349791 on Build GRCh38
rs782125615 Chr.X: 154349449-154349449 on Build GRCh38
rs782714612 Chr.X: 154349488-154349488 on Build GRCh38
rs782709737 Chr.X: 154349408-154349408 on Build GRCh38
rs782006629 Chr.X: 154349403-154349403 on Build GRCh38
rs991486151 Chr.X: 154349307-154349307 on Build GRCh38
rs998077945 Chr.X: 154349506-154349506 on Build GRCh38
rs782276654 Chr.X: 154349697-154349697 on Build GRCh38
rs371724771 Chr.X: 154349469-154349469 on Build GRCh38
rs782375435 Chr.X: 154349393-154349393 on Build GRCh38
rs377518545 Chr.X: 154349380-154349380 on Build GRCh38
rs909656307 Chr.X: 154349459-154349459 on Build GRCh38
rs782335342 Chr.X: 154349555-154349555 on Build GRCh38
rs372491500 Chr.X: 154349608-154349608 on Build GRCh38
rs782046703 Chr.X: 154349311-154349311 on Build GRCh38
rs782013219 Chr.X: 154349571-154349571 on Build GRCh38
rs782084663 Chr.X: 154349606-154349606 on Build GRCh38
rs782400342 Chr.X: 154349771-154349771 on Build GRCh38
rs7063300 Chr.X: 154349351-154349351 on Build GRCh38
rs781930608 Chr.X: 154349406-154349406 on Build GRCh38
rs782496288 Chr.X: 154349491-154349491 on Build GRCh38
rs782431987 Chr.X: 154349732-154349732 on Build GRCh38
rs782383611 Chr.X: 154349563-154349563 on Build GRCh38
rs367666924 Chr.X: 154349462-154349462 on Build GRCh38
rs782790817 Chr.X: 154349617-154349617 on Build GRCh38
rs782557713 Chr.X: 154349751-154349751 on Build GRCh38
rs782651197 Chr.X: 154349496-154349496 on Build GRCh38
rs782563616 Chr.X: 154349733-154349733 on Build GRCh38
rs201762017 Chr.X: 154349490-154349490 on Build GRCh38
rs398123624 Chr.X: 154349447-154349447 on Build GRCh38
rs113471366 Chr.X: 154349359-154349359 on Build GRCh38
rs781983475 Chr.X: 154349435-154349435 on Build GRCh38
rs782686474 Chr.X: 154349350-154349350 on Build GRCh38
rs782244918 Chr.X: 154349560-154349560 on Build GRCh38
rs373872030 Chr.X: 154349451-154349451 on Build GRCh38
rs782555930 Chr.X: 154349550-154349550 on Build GRCh38
rs782602947 Chr.X: 154349671-154349671 on Build GRCh38
rs971390304 Chr.X: 154349682-154349682 on Build GRCh38
rs398123623 Chr.X: 154349643-154349650 on Build GRCh38
rs863223626 Chr.X: 154349681-154349681 on Build GRCh38
rs782320498 Chr.X: 154349749-154349749 on Build GRCh38
rs781813065 Chr.X: 154349321-154349321 on Build GRCh38

View Full Product Details