Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 118907429-118907959 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ASF1A
Entrez Gene ID
25842
Gene Name
anti-silencing function 1A histone chaperone
Gene Aliases
CGI-98, CIA, HSPC146
UniGene
Hs.292316
Gene Chromosome Location
Chr.6: 118894077-118909171 on Build GRCh38
Transcripts
NM_014034.2 NM_017696.2

Gene Symbol
MCM9
Entrez Gene ID
254394
Gene Name
minichromosome maintenance 9 homologous recombination repair factor
Gene Aliases
C6orf61, MCMDC1, ODG4, dJ329L24.1, dJ329L24.3
UniGene
Hs.279008
Gene Chromosome Location
Chr.6: 118813449-118935162 on Build GRCh38
Transcripts
NM_014034.2 NM_017696.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5209612 c.473delA p.K159fs*35 Chr.6: 118907472-118907472 on Build GRCh38
3170807 c.516A>C p.S172S Chr.6: 118907515-118907515 on Build GRCh38
4625655 c.490G>T p.E164* Chr.6: 118907489-118907489 on Build GRCh38
3170806 c.501T>C p.N167N Chr.6: 118907500-118907500 on Build GRCh38
3170804 c.468C>T p.N156N Chr.6: 118907467-118907467 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs991207956 Chr.6: 118907814-118907815 on Build GRCh38
rs775710424 Chr.6: 118907590-118907590 on Build GRCh38
rs779525420 Chr.6: 118907518-118907518 on Build GRCh38
rs549775585 Chr.6: 118907853-118907853 on Build GRCh38
rs750133147 Chr.6: 118907627-118907627 on Build GRCh38
rs79242554 Chr.6: 118907584-118907584 on Build GRCh38
rs749980809 Chr.6: 118907511-118907511 on Build GRCh38
rs117626273 Chr.6: 118907682-118907682 on Build GRCh38
rs193193876 Chr.6: 118907878-118907878 on Build GRCh38
rs764589675 Chr.6: 118907479-118907479 on Build GRCh38
rs557053322 Chr.6: 118907507-118907507 on Build GRCh38
rs781052600 Chr.6: 118907534-118907534 on Build GRCh38
rs915623142 Chr.6: 118907818-118907822 on Build GRCh38
rs184693166 Chr.6: 118907462-118907462 on Build GRCh38
rs925727394 Chr.6: 118907553-118907553 on Build GRCh38
rs977686765 Chr.6: 118907774-118907774 on Build GRCh38
rs576955613 Chr.6: 118907548-118907548 on Build GRCh38
rs771966125 Chr.6: 118907763-118907763 on Build GRCh38
rs7765016 Chr.6: 118907908-118907908 on Build GRCh38
rs1046499115 Chr.6: 118907702-118907702 on Build GRCh38
rs935808699 Chr.6: 118907785-118907785 on Build GRCh38
rs925789969 Chr.6: 118907778-118907778 on Build GRCh38
rs780947972 Chr.6: 118907663-118907663 on Build GRCh38
rs190021689 Chr.6: 118907659-118907659 on Build GRCh38
rs929558720 Chr.6: 118907935-118907935 on Build GRCh38
rs1007662436 Chr.6: 118907694-118907694 on Build GRCh38
rs542163773 Chr.6: 118907709-118907709 on Build GRCh38
rs957293290 Chr.6: 118907496-118907496 on Build GRCh38
rs1045965800 Chr.6: 118907621-118907621 on Build GRCh38
rs371152823 Chr.6: 118907605-118907605 on Build GRCh38
rs935751073 Chr.6: 118907576-118907576 on Build GRCh38
rs772339793 Chr.6: 118907586-118907586 on Build GRCh38
rs990407258 Chr.6: 118907499-118907499 on Build GRCh38
rs754840793 Chr.6: 118907531-118907531 on Build GRCh38
rs754929337 Chr.6: 118907660-118907660 on Build GRCh38
rs765876829 Chr.6: 118907658-118907658 on Build GRCh38
rs377743166 Chr.6: 118907537-118907537 on Build GRCh38
rs765606134 Chr.6: 118907608-118907608 on Build GRCh38
rs923010143 Chr.6: 118907599-118907599 on Build GRCh38
rs748037517 Chr.6: 118907743-118907743 on Build GRCh38
rs562329367 Chr.6: 118907753-118907753 on Build GRCh38
rs752645451 Chr.6: 118907664-118907664 on Build GRCh38
rs768988715 Chr.6: 118907596-118907596 on Build GRCh38
rs545883830 Chr.6: 118907603-118907603 on Build GRCh38
rs928988254 Chr.6: 118907616-118907616 on Build GRCh38
rs116091423 Chr.6: 118907770-118907770 on Build GRCh38
rs1027812017 Chr.6: 118907684-118907684 on Build GRCh38
rs1045367391 Chr.6: 118907861-118907861 on Build GRCh38
rs562829563 Chr.6: 118907681-118907681 on Build GRCh38
rs1047376154 Chr.6: 118907622-118907622 on Build GRCh38
rs552245656 Chr.6: 118907925-118907925 on Build GRCh38
rs761270719 Chr.6: 118907594-118907594 on Build GRCh38
rs966040767 Chr.6: 118907745-118907745 on Build GRCh38
rs115697045 Chr.6: 118907691-118907691 on Build GRCh38
rs376047323 Chr.6: 118907645-118907645 on Build GRCh38
rs964578794 Chr.6: 118907898-118907898 on Build GRCh38
rs773192408 Chr.6: 118907840-118907840 on Build GRCh38
rs746069451 Chr.6: 118907582-118907582 on Build GRCh38
rs754086680 Chr.6: 118907487-118907487 on Build GRCh38
rs757952076 Chr.6: 118907514-118907514 on Build GRCh38
rs762588943 Chr.6: 118907629-118907629 on Build GRCh38
rs747014984 Chr.6: 118907529-118907529 on Build GRCh38
rs1055891472 Chr.6: 118907587-118907587 on Build GRCh38

View Full Product Details