Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 175159280-175159874 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KIAA0040
Entrez Gene ID
9674
Gene Name
KIAA0040
Gene Aliases
-
UniGene
Hs.518138
Gene Chromosome Location
Chr.1: 175156986-175193300 on Build GRCh38
Transcripts
NM_001162893.1 NM_001162894.1 NM_001162895.1 NM_001319230.1 NM_001319231.1 NM_014656.2 XM_017002909.1 XM_017002910.1 XM_017002911.1

SNP Detail

SNP ID Chromosome Location
rs964878139 Chr.1: 175159528-175159528 on Build GRCh38
rs4650716 Chr.1: 175159575-175159575 on Build GRCh38
rs896347029 Chr.1: 175159323-175159323 on Build GRCh38
rs923998424 Chr.1: 175159511-175159511 on Build GRCh38
rs1052822148 Chr.1: 175159750-175159750 on Build GRCh38
rs548685143 Chr.1: 175159853-175159853 on Build GRCh38
rs1012150519 Chr.1: 175159474-175159474 on Build GRCh38
rs974892985 Chr.1: 175159451-175159451 on Build GRCh38
rs552303337 Chr.1: 175159663-175159663 on Build GRCh38
rs963770714 Chr.1: 175159758-175159758 on Build GRCh38
rs142967416 Chr.1: 175159627-175159627 on Build GRCh38
rs575225564 Chr.1: 175159317-175159317 on Build GRCh38
rs537774723 Chr.1: 175159325-175159325 on Build GRCh38
rs892800433 Chr.1: 175159759-175159759 on Build GRCh38
rs1045639624 Chr.1: 175159309-175159309 on Build GRCh38
rs991313728 Chr.1: 175159648-175159648 on Build GRCh38
rs371951820 Chr.1: 175159400-175159401 on Build GRCh38
rs564836781 Chr.1: 175159402-175159402 on Build GRCh38
rs909286996 Chr.1: 175159604-175159604 on Build GRCh38
rs542557734 Chr.1: 175159711-175159711 on Build GRCh38
rs879021902 Chr.1: 175159685-175159685 on Build GRCh38
rs71573011 Chr.1: 175159400-175159407 on Build GRCh38
rs747478937 Chr.1: 175159844-175159844 on Build GRCh38
rs534277991 Chr.1: 175159475-175159475 on Build GRCh38
rs11809587 Chr.1: 175159308-175159308 on Build GRCh38
rs998560819 Chr.1: 175159338-175159338 on Build GRCh38
rs67332874 Chr.1: 175159401-175159401 on Build GRCh38
rs35639410 Chr.1: 175159401-175159401 on Build GRCh38
rs1053262378 Chr.1: 175159344-175159344 on Build GRCh38
rs532216336 Chr.1: 175159661-175159661 on Build GRCh38
rs78454573 Chr.1: 175159494-175159494 on Build GRCh38
rs556049290 Chr.1: 175159739-175159739 on Build GRCh38
rs966212277 Chr.1: 175159439-175159439 on Build GRCh38
rs997745547 Chr.1: 175159542-175159542 on Build GRCh38
rs1023535448 Chr.1: 175159370-175159370 on Build GRCh38
rs750320403 Chr.1: 175159534-175159534 on Build GRCh38
rs995137846 Chr.1: 175159398-175159398 on Build GRCh38
rs1008376550 Chr.1: 175159737-175159737 on Build GRCh38
rs1038821224 Chr.1: 175159821-175159821 on Build GRCh38
rs577293349 Chr.1: 175159537-175159537 on Build GRCh38
rs892925547 Chr.1: 175159471-175159471 on Build GRCh38
rs188384767 Chr.1: 175159741-175159741 on Build GRCh38
rs557445112 Chr.1: 175159536-175159536 on Build GRCh38
rs560587170 Chr.1: 175159570-175159570 on Build GRCh38
rs76040830 Chr.1: 175159777-175159777 on Build GRCh38
rs769082071 Chr.1: 175159394-175159394 on Build GRCh38
rs34270511 Chr.1: 175159407-175159407 on Build GRCh38
rs922152309 Chr.1: 175159452-175159452 on Build GRCh38
rs1025725782 Chr.1: 175159664-175159664 on Build GRCh38
rs557664738 Chr.1: 175159399-175159399 on Build GRCh38
rs72719370 Chr.1: 175159400-175159400 on Build GRCh38
rs6664840 Chr.1: 175159590-175159590 on Build GRCh38
rs962632711 Chr.1: 175159379-175159379 on Build GRCh38
rs935400300 Chr.1: 175159521-175159521 on Build GRCh38

View Full Product Details