Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 107526115-107526537 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BBX
Entrez Gene ID
56987
Gene Name
BBX, HMG-box containing
Gene Aliases
ARTC1, HBP2, HSPC339, MDS001
UniGene
Hs.124366
Gene Chromosome Location
Chr.3: 107522936-107811329 on Build GRCh38
Transcripts
NM_001142568.2 NM_001276286.1 NM_020235.6 XM_005247642.3 XM_005247643.3 XM_005247644.3 XM_011512999.2 XM_011513000.1 XM_011513001.1 XM_011513002.2 XM_011513003.2 XM_011513004.2 XM_011513005.2 XM_011513006.2 XM_011513007.2 XM_011513010.2 XM_017006881.1 XM_017006882.1 XM_017006883.1

SNP Detail

SNP ID Chromosome Location
rs924879794 Chr.3: 107526392-107526392 on Build GRCh38
rs1023114120 Chr.3: 107526354-107526354 on Build GRCh38
rs9876650 Chr.3: 107526204-107526204 on Build GRCh38
rs934997751 Chr.3: 107526174-107526174 on Build GRCh38
rs180968570 Chr.3: 107526397-107526397 on Build GRCh38
rs923836112 Chr.3: 107526378-107526378 on Build GRCh38
rs550848241 Chr.3: 107526336-107526336 on Build GRCh38
rs950251067 Chr.3: 107526135-107526135 on Build GRCh38
rs607878 Chr.3: 107526179-107526179 on Build GRCh38
rs758140192 Chr.3: 107526185-107526185 on Build GRCh38
rs991583099 Chr.3: 107526338-107526338 on Build GRCh38
rs561683442 Chr.3: 107526182-107526182 on Build GRCh38
rs982044577 Chr.3: 107526374-107526374 on Build GRCh38
rs917724223 Chr.3: 107526410-107526410 on Build GRCh38
rs563624189 Chr.3: 107526275-107526275 on Build GRCh38
rs774945546 Chr.3: 107526487-107526487 on Build GRCh38
rs1036002760 Chr.3: 107526497-107526497 on Build GRCh38
rs1029754243 Chr.3: 107526271-107526271 on Build GRCh38
rs768103384 Chr.3: 107526371-107526371 on Build GRCh38
rs1017588936 Chr.3: 107526217-107526217 on Build GRCh38
rs533005144 Chr.3: 107526487-107526487 on Build GRCh38
rs771478778 Chr.3: 107526435-107526435 on Build GRCh38
rs956586966 Chr.3: 107526383-107526383 on Build GRCh38
rs574814438 Chr.3: 107526251-107526251 on Build GRCh38
rs897423792 Chr.3: 107526513-107526513 on Build GRCh38
rs547094604 Chr.3: 107526466-107526466 on Build GRCh38
rs955463053 Chr.3: 107526294-107526294 on Build GRCh38
rs543733305 Chr.3: 107526252-107526252 on Build GRCh38
rs1029563614 Chr.3: 107526286-107526286 on Build GRCh38
rs546373207 Chr.3: 107526362-107526362 on Build GRCh38
rs887054685 Chr.3: 107526291-107526291 on Build GRCh38
rs1016986074 Chr.3: 107526323-107526323 on Build GRCh38
rs912717396 Chr.3: 107526455-107526467 on Build GRCh38
rs528163458 Chr.3: 107526146-107526146 on Build GRCh38
rs924880221 Chr.3: 107526144-107526144 on Build GRCh38
rs998488247 Chr.3: 107526226-107526226 on Build GRCh38
rs746733916 Chr.3: 107526219-107526219 on Build GRCh38
rs577060524 Chr.3: 107526276-107526276 on Build GRCh38
rs981734872 Chr.3: 107526137-107526137 on Build GRCh38
rs1004210099 Chr.3: 107526301-107526301 on Build GRCh38
rs897350533 Chr.3: 107526223-107526223 on Build GRCh38
rs947789969 Chr.3: 107526443-107526443 on Build GRCh38
rs560842282 Chr.3: 107526493-107526493 on Build GRCh38
rs527338058 Chr.3: 107526418-107526418 on Build GRCh38
rs554913812 Chr.3: 107526220-107526220 on Build GRCh38
rs1005843493 Chr.3: 107526205-107526205 on Build GRCh38

View Full Product Details