Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 40924326-40924844 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CNTN1
Entrez Gene ID
1272
Gene Name
contactin 1
Gene Aliases
F3, GP135, MYPCN
UniGene
Hs.143434
Gene Chromosome Location
Chr.12: 40692439-41072412 on Build GRCh38
Transcripts
NM_001256063.1 NM_001256064.1 NM_001843.3 NM_175038.2 XM_005268651.2 XM_006719241.2 XM_011537926.2 XM_011537927.2 XM_017018821.1 XM_017018822.1 XM_017018823.1 XM_017018824.1 XM_017018825.1 XM_017018826.1 XM_017018827.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7361783 c.452G>T p.G151V Chr.12: 40924608-40924608 on Build GRCh38
400153 c.462G>T p.M154I Chr.12: 40924618-40924618 on Build GRCh38
1676868 c.494C>T p.P165L Chr.12: 40924650-40924650 on Build GRCh38
3871551 c.471C>T p.L157L Chr.12: 40924627-40924627 on Build GRCh38
1676867 c.457G>A p.G153R Chr.12: 40924613-40924613 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs540959289 Chr.12: 40924732-40924732 on Build GRCh38
rs753302581 Chr.12: 40924583-40924583 on Build GRCh38
rs1025558923 Chr.12: 40924530-40924530 on Build GRCh38
rs769899922 Chr.12: 40924633-40924633 on Build GRCh38
rs779534185 Chr.12: 40924578-40924578 on Build GRCh38
rs7299744 Chr.12: 40924463-40924463 on Build GRCh38
rs956672528 Chr.12: 40924404-40924404 on Build GRCh38
rs751830078 Chr.12: 40924651-40924651 on Build GRCh38
rs563442407 Chr.12: 40924387-40924387 on Build GRCh38
rs184703177 Chr.12: 40924566-40924566 on Build GRCh38
rs928145355 Chr.12: 40924494-40924494 on Build GRCh38
rs146099643 Chr.12: 40924551-40924551 on Build GRCh38
rs374932678 Chr.12: 40924667-40924667 on Build GRCh38
rs952921668 Chr.12: 40924819-40924819 on Build GRCh38
rs777939151 Chr.12: 40924702-40924702 on Build GRCh38
rs760608825 Chr.12: 40924383-40924383 on Build GRCh38
rs570939203 Chr.12: 40924485-40924485 on Build GRCh38
rs768380048 Chr.12: 40924402-40924402 on Build GRCh38
rs778263271 Chr.12: 40924592-40924592 on Build GRCh38
rs1030965830 Chr.12: 40924378-40924378 on Build GRCh38
rs540267446 Chr.12: 40924359-40924359 on Build GRCh38
rs944116841 Chr.12: 40924585-40924585 on Build GRCh38
rs1022239090 Chr.12: 40924675-40924675 on Build GRCh38
rs932519800 Chr.12: 40924666-40924666 on Build GRCh38
rs1021500295 Chr.12: 40924424-40924424 on Build GRCh38
rs745976709 Chr.12: 40924621-40924621 on Build GRCh38
rs755503956 Chr.12: 40924577-40924577 on Build GRCh38
rs969593879 Chr.12: 40924539-40924539 on Build GRCh38
rs969909359 Chr.12: 40924680-40924680 on Build GRCh38
rs375966126 Chr.12: 40924686-40924686 on Build GRCh38
rs575754931 Chr.12: 40924549-40924549 on Build GRCh38
rs781083008 Chr.12: 40924616-40924616 on Build GRCh38
rs1014389210 Chr.12: 40924756-40924756 on Build GRCh38
rs761808311 Chr.12: 40924634-40924634 on Build GRCh38
rs980947063 Chr.12: 40924804-40924804 on Build GRCh38
rs992632730 Chr.12: 40924510-40924510 on Build GRCh38
rs753587977 Chr.12: 40924668-40924668 on Build GRCh38
rs181708847 Chr.12: 40924409-40924409 on Build GRCh38
rs3816251 Chr.12: 40924367-40924367 on Build GRCh38
rs377324964 Chr.12: 40924799-40924799 on Build GRCh38
rs57340925 Chr.12: 40924548-40924548 on Build GRCh38
rs981375297 Chr.12: 40924487-40924487 on Build GRCh38
rs904436514 Chr.12: 40924692-40924692 on Build GRCh38
rs1022713854 Chr.12: 40924757-40924757 on Build GRCh38
rs372134933 Chr.12: 40924764-40924764 on Build GRCh38
rs956725052 Chr.12: 40924646-40924646 on Build GRCh38
rs566391368 Chr.12: 40924795-40924795 on Build GRCh38
rs766559055 Chr.12: 40924464-40924464 on Build GRCh38
rs538318732 Chr.12: 40924535-40924535 on Build GRCh38
rs886038489 Chr.12: 40924543-40924543 on Build GRCh38
rs908567017 Chr.12: 40924572-40924572 on Build GRCh38
rs764895806 Chr.12: 40924681-40924681 on Build GRCh38
rs773073688 Chr.12: 40924648-40924648 on Build GRCh38
rs764040709 Chr.12: 40924544-40924544 on Build GRCh38
rs565322294 Chr.12: 40924360-40924360 on Build GRCh38
rs760190868 Chr.12: 40924518-40924519 on Build GRCh38
rs762818004 Chr.12: 40924542-40924542 on Build GRCh38
rs148387796 Chr.12: 40924537-40924537 on Build GRCh38
rs1048795475 Chr.12: 40924812-40924812 on Build GRCh38
rs200290654 Chr.12: 40924700-40924700 on Build GRCh38
rs532632745 Chr.12: 40924366-40924366 on Build GRCh38
rs765752838 Chr.12: 40924565-40924565 on Build GRCh38
rs1036575014 Chr.12: 40924500-40924500 on Build GRCh38
rs2897101 Chr.12: 40924818-40924818 on Build GRCh38
rs1031297103 Chr.12: 40924740-40924740 on Build GRCh38
rs543140437 Chr.12: 40924349-40924349 on Build GRCh38
rs747130472 Chr.12: 40924597-40924597 on Build GRCh38
rs776772859 Chr.12: 40924540-40924540 on Build GRCh38
rs758820625 Chr.12: 40924584-40924584 on Build GRCh38
rs907650664 Chr.12: 40924531-40924531 on Build GRCh38
rs895519412 Chr.12: 40924743-40924743 on Build GRCh38
rs766090898 Chr.12: 40924665-40924665 on Build GRCh38
rs534964247 Chr.12: 40924456-40924456 on Build GRCh38
rs549218952 Chr.12: 40924410-40924410 on Build GRCh38
rs983070278 Chr.12: 40924821-40924821 on Build GRCh38
rs565254543 Chr.12: 40924760-40924760 on Build GRCh38
rs767405486 Chr.12: 40924550-40924550 on Build GRCh38
rs61508958 Chr.12: 40924429-40924429 on Build GRCh38
rs776347034 Chr.12: 40924442-40924442 on Build GRCh38
rs753157393 Chr.12: 40924536-40924537 on Build GRCh38
rs1018768776 Chr.12: 40924364-40924364 on Build GRCh38
rs771697105 Chr.12: 40924636-40924636 on Build GRCh38
rs759097821 Chr.12: 40924678-40924678 on Build GRCh38
rs372959410 Chr.12: 40924598-40924598 on Build GRCh38

View Full Product Details