Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 157748073-157748664 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FCRL2
Entrez Gene ID
79368
Gene Name
Fc receptor like 2
Gene Aliases
CD307b, FCRH2, IFGP4, IRTA4, SPAP1, SPAP1A, SPAP1B, SPAP1C
UniGene
Hs.437393
Gene Chromosome Location
Chr.1: 157745733-157777472 on Build GRCh38
Transcripts
NM_001159488.1 NM_030764.3 NR_125358.1 XM_011509974.2 XM_011509975.2 XM_011509976.2 XM_017002316.1 XM_017002317.1 XM_017002318.1 XM_017002319.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1747833 c.1450G>C p.E484Q Chr.1: 157748562-157748562 on Build GRCh38
463188 c.1394-1G>T p.? Chr.1: 157748619-157748619 on Build GRCh38
5911325 c.1447C>T p.P483S Chr.1: 157748565-157748565 on Build GRCh38
3369346 c.1438A>G p.M480V Chr.1: 157748574-157748574 on Build GRCh38
676269 c.1410G>A p.V470V Chr.1: 157748602-157748602 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs374404136 Chr.1: 157748622-157748622 on Build GRCh38
rs1027048239 Chr.1: 157748294-157748294 on Build GRCh38
rs955482055 Chr.1: 157748279-157748279 on Build GRCh38
rs575866781 Chr.1: 157748633-157748633 on Build GRCh38
rs962760680 Chr.1: 157748255-157748255 on Build GRCh38
rs531793494 Chr.1: 157748487-157748487 on Build GRCh38
rs186690452 Chr.1: 157748376-157748376 on Build GRCh38
rs372871876 Chr.1: 157748588-157748588 on Build GRCh38
rs1008267213 Chr.1: 157748483-157748483 on Build GRCh38
rs201754069 Chr.1: 157748609-157748609 on Build GRCh38
rs759487385 Chr.1: 157748563-157748563 on Build GRCh38
rs746167760 Chr.1: 157748143-157748143 on Build GRCh38
rs777459755 Chr.1: 157748628-157748628 on Build GRCh38
rs774828933 Chr.1: 157748522-157748522 on Build GRCh38
rs951193790 Chr.1: 157748299-157748299 on Build GRCh38
rs58239785 Chr.1: 157748473-157748480 on Build GRCh38
rs758981255 Chr.1: 157748505-157748505 on Build GRCh38
rs3818813 Chr.1: 157748535-157748535 on Build GRCh38
rs377678994 Chr.1: 157748600-157748600 on Build GRCh38
rs915586822 Chr.1: 157748532-157748532 on Build GRCh38
rs935665898 Chr.1: 157748357-157748357 on Build GRCh38
rs536097252 Chr.1: 157748497-157748497 on Build GRCh38
rs1055559283 Chr.1: 157748166-157748166 on Build GRCh38
rs916139738 Chr.1: 157748431-157748431 on Build GRCh38
rs747540646 Chr.1: 157748521-157748521 on Build GRCh38
rs967324133 Chr.1: 157748297-157748297 on Build GRCh38
rs997310965 Chr.1: 157748271-157748271 on Build GRCh38
rs373103479 Chr.1: 157748272-157748272 on Build GRCh38
rs370047704 Chr.1: 157748570-157748570 on Build GRCh38
rs925578460 Chr.1: 157748352-157748352 on Build GRCh38
rs755460380 Chr.1: 157748611-157748611 on Build GRCh38
rs991739182 Chr.1: 157748097-157748097 on Build GRCh38
rs200770449 Chr.1: 157748533-157748533 on Build GRCh38
rs908348404 Chr.1: 157748578-157748578 on Build GRCh38
rs914107723 Chr.1: 157748292-157748292 on Build GRCh38
rs374583664 Chr.1: 157748441-157748442 on Build GRCh38
rs560479081 Chr.1: 157748358-157748358 on Build GRCh38
rs71084251 Chr.1: 157748480-157748481 on Build GRCh38
rs1004141408 Chr.1: 157748209-157748209 on Build GRCh38
rs2275232 Chr.1: 157748640-157748640 on Build GRCh38
rs781437817 Chr.1: 157748616-157748616 on Build GRCh38
rs6696643 Chr.1: 157748493-157748493 on Build GRCh38
rs1017310533 Chr.1: 157748247-157748247 on Build GRCh38
rs35515182 Chr.1: 157748510-157748511 on Build GRCh38
rs894257777 Chr.1: 157748182-157748182 on Build GRCh38
rs138580358 Chr.1: 157748412-157748412 on Build GRCh38
rs996841749 Chr.1: 157748435-157748435 on Build GRCh38
rs767265232 Chr.1: 157748582-157748582 on Build GRCh38
rs543727628 Chr.1: 157748237-157748237 on Build GRCh38
rs72221192 Chr.1: 157748442-157748443 on Build GRCh38
rs561866124 Chr.1: 157748477-157748480 on Build GRCh38
rs561817880 Chr.1: 157748238-157748238 on Build GRCh38
rs1014077637 Chr.1: 157748186-157748186 on Build GRCh38
rs925539452 Chr.1: 157748500-157748500 on Build GRCh38
rs774221665 Chr.1: 157748557-157748557 on Build GRCh38
rs200327020 Chr.1: 157748571-157748571 on Build GRCh38
rs67699507 Chr.1: 157748449-157748450 on Build GRCh38
rs1038636761 Chr.1: 157748618-157748618 on Build GRCh38
rs770412973 Chr.1: 157748553-157748553 on Build GRCh38
rs915660199 Chr.1: 157748360-157748360 on Build GRCh38
rs890950295 Chr.1: 157748226-157748226 on Build GRCh38
rs370742052 Chr.1: 157748598-157748598 on Build GRCh38
rs932596689 Chr.1: 157748630-157748630 on Build GRCh38
rs77280832 Chr.1: 157748433-157748433 on Build GRCh38
rs780281130 Chr.1: 157748607-157748607 on Build GRCh38
rs573844417 Chr.1: 157748253-157748253 on Build GRCh38
rs769447659 Chr.1: 157748626-157748626 on Build GRCh38
rs960157081 Chr.1: 157748382-157748382 on Build GRCh38
rs776090678 Chr.1: 157748280-157748280 on Build GRCh38
rs1004514109 Chr.1: 157748384-157748384 on Build GRCh38
rs887015242 Chr.1: 157748195-157748195 on Build GRCh38
rs539191796 Chr.1: 157748601-157748601 on Build GRCh38
rs1043179562 Chr.1: 157748161-157748161 on Build GRCh38
rs183538725 Chr.1: 157748105-157748105 on Build GRCh38
rs747283044 Chr.1: 157748643-157748643 on Build GRCh38
rs915920441 Chr.1: 157748099-157748099 on Build GRCh38
rs979724566 Chr.1: 157748339-157748339 on Build GRCh38
rs749679897 Chr.1: 157748301-157748301 on Build GRCh38
rs949815705 Chr.1: 157748370-157748370 on Build GRCh38
rs955451405 Chr.1: 157748464-157748464 on Build GRCh38
rs200153368 Chr.1: 157748441-157748441 on Build GRCh38
rs553371725 Chr.1: 157748288-157748288 on Build GRCh38
rs903288501 Chr.1: 157748174-157748174 on Build GRCh38
rs898410770 Chr.1: 157748620-157748620 on Build GRCh38
rs550486762 Chr.1: 157748491-157748491 on Build GRCh38
rs935723996 Chr.1: 157748148-157748148 on Build GRCh38
rs1045555368 Chr.1: 157748374-157748374 on Build GRCh38
rs1008100125 Chr.1: 157748227-157748227 on Build GRCh38
rs747775807 Chr.1: 157748523-157748523 on Build GRCh38
rs1041741268 Chr.1: 157748118-157748118 on Build GRCh38
rs761974943 Chr.1: 157748583-157748583 on Build GRCh38
rs762389750 Chr.1: 157748548-157748548 on Build GRCh38
rs1018115367 Chr.1: 157748228-157748228 on Build GRCh38
rs758612307 Chr.1: 157748606-157748606 on Build GRCh38
rs143605468 Chr.1: 157748442-157748445 on Build GRCh38
rs16839030 Chr.1: 157748104-157748104 on Build GRCh38
rs150151373 Chr.1: 157748189-157748189 on Build GRCh38

View Full Product Details