Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 63819598-63820141 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FTSJ3
Entrez Gene ID
117246
Gene Name
FtsJ homolog 3
Gene Aliases
EPCS3, SPB1
UniGene
Hs.463785
Gene Chromosome Location
Chr.17: 63819433-63827671 on Build GRCh38
Transcripts
NM_017647.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
982703 c.2434C>T p.R812C Chr.17: 63819912-63819912 on Build GRCh38
3421781 c.2347C>T p.R783* Chr.17: 63820083-63820083 on Build GRCh38
6886101 c.2467G>A p.V823M Chr.17: 63819879-63819879 on Build GRCh38
4068625 c.2406A>T p.V802V Chr.17: 63819940-63819940 on Build GRCh38
1385187 c.2447G>A p.G816E Chr.17: 63819899-63819899 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs751496618 Chr.17: 63819954-63819954 on Build GRCh38
rs1018998343 Chr.17: 63820108-63820108 on Build GRCh38
rs185973898 Chr.17: 63820099-63820099 on Build GRCh38
rs755095380 Chr.17: 63819791-63819791 on Build GRCh38
rs563310643 Chr.17: 63819765-63819765 on Build GRCh38
rs370714262 Chr.17: 63819955-63819955 on Build GRCh38
rs747129028 Chr.17: 63820066-63820066 on Build GRCh38
rs767034414 Chr.17: 63820025-63820025 on Build GRCh38
rs114763743 Chr.17: 63819810-63819810 on Build GRCh38
rs759584076 Chr.17: 63819920-63819920 on Build GRCh38
rs747257896 Chr.17: 63820001-63820001 on Build GRCh38
rs532745165 Chr.17: 63819710-63819710 on Build GRCh38
rs1045436188 Chr.17: 63819721-63819721 on Build GRCh38
rs764002549 Chr.17: 63819798-63819798 on Build GRCh38
rs757994274 Chr.17: 63819814-63819814 on Build GRCh38
rs779366840 Chr.17: 63819823-63819823 on Build GRCh38
rs377705927 Chr.17: 63819963-63819963 on Build GRCh38
rs750779578 Chr.17: 63820117-63820117 on Build GRCh38
rs768148368 Chr.17: 63819828-63819828 on Build GRCh38
rs762710245 Chr.17: 63820022-63820022 on Build GRCh38
rs757087696 Chr.17: 63819808-63819808 on Build GRCh38
rs948527574 Chr.17: 63820009-63820009 on Build GRCh38
rs765544044 Chr.17: 63820116-63820116 on Build GRCh38
rs898084442 Chr.17: 63820100-63820100 on Build GRCh38
rs763147438 Chr.17: 63819766-63819766 on Build GRCh38
rs759832303 Chr.17: 63819890-63819890 on Build GRCh38
rs771371440 Chr.17: 63819813-63819814 on Build GRCh38
rs925094316 Chr.17: 63820048-63820048 on Build GRCh38
rs755880819 Chr.17: 63819702-63819702 on Build GRCh38
rs144046237 Chr.17: 63819836-63819836 on Build GRCh38
rs986457818 Chr.17: 63819758-63819758 on Build GRCh38
rs758808221 Chr.17: 63820121-63820121 on Build GRCh38
rs779042374 Chr.17: 63819809-63819809 on Build GRCh38
rs754026039 Chr.17: 63820062-63820065 on Build GRCh38
rs767798509 Chr.17: 63819908-63819910 on Build GRCh38
rs775922784 Chr.17: 63819928-63819928 on Build GRCh38
rs775762608 Chr.17: 63819997-63819998 on Build GRCh38
rs768723069 Chr.17: 63820004-63820004 on Build GRCh38
rs200242137 Chr.17: 63819983-63819983 on Build GRCh38
rs771302233 Chr.17: 63819912-63819912 on Build GRCh38
rs760842390 Chr.17: 63819999-63819999 on Build GRCh38
rs912294356 Chr.17: 63819780-63819780 on Build GRCh38
rs752159429 Chr.17: 63820037-63820037 on Build GRCh38
rs780313886 Chr.17: 63820063-63820063 on Build GRCh38
rs767480122 Chr.17: 63820101-63820101 on Build GRCh38
rs771053578 Chr.17: 63819881-63819881 on Build GRCh38
rs977279078 Chr.17: 63819925-63819925 on Build GRCh38
rs774079721 Chr.17: 63820095-63820095 on Build GRCh38
rs754457858 Chr.17: 63819903-63819903 on Build GRCh38
rs201755066 Chr.17: 63819930-63819930 on Build GRCh38
rs1046696935 Chr.17: 63820118-63820118 on Build GRCh38
rs370085151 Chr.17: 63819929-63819929 on Build GRCh38
rs762982336 Chr.17: 63819945-63819945 on Build GRCh38
rs750138661 Chr.17: 63819944-63819944 on Build GRCh38
rs953711835 Chr.17: 63819720-63819720 on Build GRCh38
rs749652084 Chr.17: 63819968-63819968 on Build GRCh38
rs750995958 Chr.17: 63819902-63819902 on Build GRCh38
rs771304846 Chr.17: 63819972-63819972 on Build GRCh38
rs776883705 Chr.17: 63820005-63820005 on Build GRCh38
rs534387064 Chr.17: 63819624-63819625 on Build GRCh38
rs770642137 Chr.17: 63820093-63820093 on Build GRCh38
rs201190672 Chr.17: 63819984-63819984 on Build GRCh38
rs895432096 Chr.17: 63819891-63819891 on Build GRCh38
rs749783868 Chr.17: 63819808-63819808 on Build GRCh38
rs774847539 Chr.17: 63819834-63819835 on Build GRCh38
rs895106443 Chr.17: 63819642-63819642 on Build GRCh38
rs774889086 Chr.17: 63819915-63819915 on Build GRCh38
rs753437920 Chr.17: 63820104-63820104 on Build GRCh38
rs768761341 Chr.17: 63820069-63820069 on Build GRCh38
rs1013629273 Chr.17: 63819649-63819649 on Build GRCh38
rs750381400 Chr.17: 63819899-63819899 on Build GRCh38
rs772264343 Chr.17: 63819921-63819921 on Build GRCh38
rs148678282 Chr.17: 63819895-63819895 on Build GRCh38
rs368258895 Chr.17: 63820040-63820040 on Build GRCh38
rs542369102 Chr.17: 63820047-63820047 on Build GRCh38
rs372869515 Chr.17: 63819908-63819908 on Build GRCh38
rs117079025 Chr.17: 63819685-63819685 on Build GRCh38
rs991030022 Chr.17: 63820055-63820055 on Build GRCh38
rs921194348 Chr.17: 63819708-63819708 on Build GRCh38
rs201254119 Chr.17: 63819753-63819753 on Build GRCh38
rs199627207 Chr.17: 63819752-63819752 on Build GRCh38
rs925406853 Chr.17: 63819832-63819832 on Build GRCh38
rs950808944 Chr.17: 63819657-63819657 on Build GRCh38
rs753644776 Chr.17: 63819804-63819804 on Build GRCh38
rs746689894 Chr.17: 63819826-63819826 on Build GRCh38
rs541317216 Chr.17: 63819674-63819674 on Build GRCh38
rs760234926 Chr.17: 63820103-63820103 on Build GRCh38
rs542987056 Chr.17: 63819760-63819760 on Build GRCh38
rs748401152 Chr.17: 63820083-63820083 on Build GRCh38
rs780609940 Chr.17: 63819956-63819956 on Build GRCh38
rs753515910 Chr.17: 63820044-63820044 on Build GRCh38
rs565732602 Chr.17: 63820024-63820024 on Build GRCh38
rs776780004 Chr.17: 63820082-63820082 on Build GRCh38
rs769071697 Chr.17: 63819762-63819762 on Build GRCh38
rs746391206 Chr.17: 63820051-63820051 on Build GRCh38
rs776926799 Chr.17: 63819763-63819763 on Build GRCh38
rs766270101 Chr.17: 63819946-63819946 on Build GRCh38
rs528333562 Chr.17: 63820038-63820038 on Build GRCh38
rs572648132 Chr.17: 63819664-63819664 on Build GRCh38
rs375957187 Chr.17: 63819764-63819764 on Build GRCh38
rs749821889 Chr.17: 63819911-63819911 on Build GRCh38
rs199564934 Chr.17: 63819898-63819898 on Build GRCh38
rs752355649 Chr.17: 63819959-63819959 on Build GRCh38
rs775655190 Chr.17: 63820000-63820000 on Build GRCh38
rs763299454 Chr.17: 63819876-63819876 on Build GRCh38
rs758927243 Chr.17: 63820061-63820061 on Build GRCh38
rs190641756 Chr.17: 63820102-63820102 on Build GRCh38
rs756877415 Chr.17: 63820112-63820112 on Build GRCh38
rs779346380 Chr.17: 63819982-63819982 on Build GRCh38
rs766187244 Chr.17: 63820023-63820023 on Build GRCh38
rs201676074 Chr.17: 63819858-63819858 on Build GRCh38
rs757424604 Chr.17: 63820117-63820118 on Build GRCh38
rs974363261 Chr.17: 63819693-63819693 on Build GRCh38
rs779641176 Chr.17: 63819900-63819900 on Build GRCh38
rs759856668 Chr.17: 63819856-63819858 on Build GRCh38
rs376996404 Chr.17: 63819909-63819909 on Build GRCh38
rs966356420 Chr.17: 63820045-63820045 on Build GRCh38
rs774244917 Chr.17: 63819887-63819887 on Build GRCh38
rs766360236 Chr.17: 63819769-63819769 on Build GRCh38
rs948388115 Chr.17: 63819719-63819719 on Build GRCh38
rs370920404 Chr.17: 63820043-63820043 on Build GRCh38
rs751808023 Chr.17: 63819789-63819789 on Build GRCh38
rs747791647 Chr.17: 63819904-63819904 on Build GRCh38
rs73992400 Chr.17: 63819656-63819656 on Build GRCh38
rs764226116 Chr.17: 63819999-63820000 on Build GRCh38
rs767892003 Chr.17: 63819893-63819893 on Build GRCh38
rs770124798 Chr.17: 63819837-63819837 on Build GRCh38

View Full Product Details