Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 1227851-1228437 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CBARP
Entrez Gene ID
255057
Gene Name
CACN beta subunit associated regulatory protein
Gene Aliases
BARP, C19orf26, DOS
UniGene
Hs.346575
Gene Chromosome Location
Chr.19: 1228287-1238512 on Build GRCh38
Transcripts
XM_017026555.1 XM_017026556.1 XM_017026557.1 XM_017026558.1 NM_000455.4 XM_005259617.2 XM_011528209.1

Gene Symbol
STK11
Entrez Gene ID
6794
Gene Name
serine/threonine kinase 11
Gene Aliases
LKB1, PJS, hLKB1
UniGene
Hs.515005
Gene Chromosome Location
Chr.19: 1205799-1228435 on Build GRCh38
Transcripts
XM_017026555.1 XM_017026556.1 XM_017026557.1 XM_017026558.1 NM_000455.4 XM_005259617.2 XM_011528209.1

SNP Detail

SNP ID Chromosome Location
rs1054374747 Chr.19: 1228238-1228238 on Build GRCh38
rs866183878 Chr.19: 1227929-1227929 on Build GRCh38
rs922149435 Chr.19: 1227934-1227934 on Build GRCh38
rs950829868 Chr.19: 1227923-1227923 on Build GRCh38
rs561432739 Chr.19: 1228190-1228190 on Build GRCh38
rs369942515 Chr.19: 1228032-1228034 on Build GRCh38
rs1032897789 Chr.19: 1228076-1228080 on Build GRCh38
rs1027644340 Chr.19: 1228141-1228141 on Build GRCh38
rs10415095 Chr.19: 1228192-1228192 on Build GRCh38
rs1019412809 Chr.19: 1228166-1228166 on Build GRCh38
rs543187681 Chr.19: 1228189-1228189 on Build GRCh38
rs944831682 Chr.19: 1227930-1227930 on Build GRCh38
rs575695993 Chr.19: 1228177-1228177 on Build GRCh38
rs886450107 Chr.19: 1228358-1228358 on Build GRCh38
rs1009973064 Chr.19: 1227888-1227888 on Build GRCh38
rs1029201339 Chr.19: 1228393-1228393 on Build GRCh38
rs759351935 Chr.19: 1227889-1227889 on Build GRCh38
rs573000220 Chr.19: 1227972-1227972 on Build GRCh38
rs991265414 Chr.19: 1227942-1227942 on Build GRCh38
rs545375742 Chr.19: 1228058-1228059 on Build GRCh38
rs142990629 Chr.19: 1228414-1228422 on Build GRCh38
rs891166347 Chr.19: 1228052-1228053 on Build GRCh38
rs971590191 Chr.19: 1227974-1227974 on Build GRCh38
rs748335753 Chr.19: 1228050-1228051 on Build GRCh38
rs368014506 Chr.19: 1228205-1228205 on Build GRCh38
rs565456677 Chr.19: 1228207-1228207 on Build GRCh38
rs1041566760 Chr.19: 1228056-1228059 on Build GRCh38
rs958414259 Chr.19: 1228105-1228105 on Build GRCh38
rs1008740983 Chr.19: 1228052-1228055 on Build GRCh38
rs532639857 Chr.19: 1228254-1228254 on Build GRCh38
rs535975204 Chr.19: 1227941-1227941 on Build GRCh38
rs912942792 Chr.19: 1228215-1228215 on Build GRCh38
rs1045495924 Chr.19: 1228244-1228244 on Build GRCh38
rs779428892 Chr.19: 1228236-1228236 on Build GRCh38
rs916960919 Chr.19: 1227954-1227954 on Build GRCh38
rs746772394 Chr.19: 1228015-1228015 on Build GRCh38
rs992108209 Chr.19: 1227879-1227879 on Build GRCh38
rs886054223 Chr.19: 1227964-1227964 on Build GRCh38
rs181506760 Chr.19: 1228283-1228283 on Build GRCh38
rs547982238 Chr.19: 1228363-1228363 on Build GRCh38
rs1051208491 Chr.19: 1228047-1228054 on Build GRCh38
rs922105616 Chr.19: 1228183-1228183 on Build GRCh38
rs903142806 Chr.19: 1228060-1228060 on Build GRCh38
rs935826095 Chr.19: 1228394-1228394 on Build GRCh38
rs1046924303 Chr.19: 1228353-1228353 on Build GRCh38
rs577030468 Chr.19: 1228070-1228070 on Build GRCh38
rs886054225 Chr.19: 1228333-1228333 on Build GRCh38
rs941613349 Chr.19: 1228024-1228024 on Build GRCh38
rs950717101 Chr.19: 1228164-1228164 on Build GRCh38
rs891528351 Chr.19: 1227883-1227883 on Build GRCh38
rs945474065 Chr.19: 1228373-1228373 on Build GRCh38
rs959411051 Chr.19: 1227909-1227909 on Build GRCh38
rs1024064212 Chr.19: 1228111-1228111 on Build GRCh38
rs545437931 Chr.19: 1228109-1228109 on Build GRCh38
rs986860354 Chr.19: 1228344-1228344 on Build GRCh38
rs1000097156 Chr.19: 1228060-1228067 on Build GRCh38
rs190680390 Chr.19: 1227890-1227890 on Build GRCh38
rs1003904516 Chr.19: 1228274-1228274 on Build GRCh38
rs966551306 Chr.19: 1228378-1228378 on Build GRCh38
rs898438996 Chr.19: 1228122-1228122 on Build GRCh38
rs973351159 Chr.19: 1227916-1227916 on Build GRCh38
rs529638447 Chr.19: 1228310-1228310 on Build GRCh38
rs907040228 Chr.19: 1228041-1228041 on Build GRCh38
rs917408778 Chr.19: 1227881-1227881 on Build GRCh38
rs932616183 Chr.19: 1228044-1228044 on Build GRCh38
rs554638416 Chr.19: 1227947-1227947 on Build GRCh38
rs995369658 Chr.19: 1228139-1228139 on Build GRCh38
rs990393937 Chr.19: 1228206-1228206 on Build GRCh38
rs1015269208 Chr.19: 1228277-1228277 on Build GRCh38
rs111773256 Chr.19: 1228261-1228261 on Build GRCh38
rs894347305 Chr.19: 1228242-1228242 on Build GRCh38
rs962869758 Chr.19: 1228298-1228301 on Build GRCh38
rs983491595 Chr.19: 1227924-1227924 on Build GRCh38
rs550127419 Chr.19: 1227893-1227893 on Build GRCh38
rs948625531 Chr.19: 1228416-1228416 on Build GRCh38
rs903030190 Chr.19: 1228210-1228210 on Build GRCh38
rs1001742962 Chr.19: 1228067-1228067 on Build GRCh38
rs971546289 Chr.19: 1228120-1228120 on Build GRCh38
rs1027592319 Chr.19: 1227919-1227919 on Build GRCh38
rs1056060858 Chr.19: 1228288-1228288 on Build GRCh38
rs935671399 Chr.19: 1227938-1227938 on Build GRCh38
rs983556879 Chr.19: 1228165-1228165 on Build GRCh38
rs886054222 Chr.19: 1227948-1227948 on Build GRCh38
rs558582150 Chr.19: 1228036-1228036 on Build GRCh38
rs921252025 Chr.19: 1228037-1228037 on Build GRCh38
rs370265572 Chr.19: 1228182-1228182 on Build GRCh38
rs563703083 Chr.19: 1228152-1228152 on Build GRCh38
rs898324352 Chr.19: 1228333-1228333 on Build GRCh38
rs945815282 Chr.19: 1227982-1227982 on Build GRCh38
rs1054524262 Chr.19: 1228413-1228425 on Build GRCh38
rs886054224 Chr.19: 1228059-1228060 on Build GRCh38
rs568634564 Chr.19: 1227901-1227901 on Build GRCh38
rs1034644018 Chr.19: 1227908-1227908 on Build GRCh38
rs996263292 Chr.19: 1228382-1228382 on Build GRCh38
rs533940465 Chr.19: 1227989-1227989 on Build GRCh38
rs975029640 Chr.19: 1227932-1227932 on Build GRCh38
rs765275672 Chr.19: 1228014-1228014 on Build GRCh38
rs1019047988 Chr.19: 1228364-1228364 on Build GRCh38
rs974173856 Chr.19: 1228327-1228327 on Build GRCh38
rs995422130 Chr.19: 1228342-1228342 on Build GRCh38

View Full Product Details