Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 86466701-86467243 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RMDN1
Entrez Gene ID
51115
Gene Name
regulator of microtubule dynamics 1
Gene Aliases
CGI-90, FAM82B, RMD-1, RMD1
UniGene
Hs.145386
Gene Chromosome Location
Chr.8: 86467116-86514348 on Build GRCh38
Transcripts
XM_011517090.2 XM_017013518.1 XM_017013525.1 NM_007013.3 XM_005250760.3 XM_005250761.3 XM_011516802.2 XM_017012993.1 XM_017012994.1 XM_017012995.1 XM_017012996.1 XM_017012997.1

Gene Symbol
WWP1
Entrez Gene ID
11059
Gene Name
WW domain containing E3 ubiquitin protein ligase 1
Gene Aliases
AIP5, Tiul1, hSDRP1
UniGene
Hs.655189
Gene Chromosome Location
Chr.8: 86342711-86468357 on Build GRCh38
Transcripts
XM_011517090.2 XM_017013518.1 XM_017013525.1 NM_007013.3 XM_005250760.3 XM_005250761.3 XM_011516802.2 XM_017012993.1 XM_017012994.1 XM_017012995.1 XM_017012996.1 XM_017012997.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6388764 c.2766A>T p.E922D Chr.8: 86466890-86466890 on Build GRCh38
6395102 c.2764G>T p.E922* Chr.8: 86466888-86466888 on Build GRCh38
4602566 c.2701A>G p.S901G Chr.8: 86466825-86466825 on Build GRCh38
6365450 c.2705A>G p.Y902C Chr.8: 86466829-86466829 on Build GRCh38
5024215 c.2678G>C p.R893P Chr.8: 86466802-86466802 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs747496330 Chr.8: 86466858-86466858 on Build GRCh38
rs908777039 Chr.8: 86466892-86466892 on Build GRCh38
rs773424150 Chr.8: 86466912-86466912 on Build GRCh38
rs1020637504 Chr.8: 86466987-86466987 on Build GRCh38
rs148979073 Chr.8: 86466872-86466872 on Build GRCh38
rs779942241 Chr.8: 86467133-86467133 on Build GRCh38
rs878969545 Chr.8: 86466908-86466908 on Build GRCh38
rs1055459488 Chr.8: 86466956-86466959 on Build GRCh38
rs768709416 Chr.8: 86466900-86466902 on Build GRCh38
rs745853034 Chr.8: 86466851-86466852 on Build GRCh38
rs912663224 Chr.8: 86466768-86466768 on Build GRCh38
rs560459890 Chr.8: 86466764-86466764 on Build GRCh38
rs560475132 Chr.8: 86467204-86467204 on Build GRCh38
rs150259568 Chr.8: 86466830-86466830 on Build GRCh38
rs540627047 Chr.8: 86467170-86467170 on Build GRCh38
rs920011469 Chr.8: 86467165-86467165 on Build GRCh38
rs112688537 Chr.8: 86467154-86467154 on Build GRCh38
rs767049925 Chr.8: 86466935-86466935 on Build GRCh38
rs769942103 Chr.8: 86466907-86466907 on Build GRCh38
rs775198805 Chr.8: 86466939-86466939 on Build GRCh38
rs752555908 Chr.8: 86466820-86466820 on Build GRCh38
rs568084293 Chr.8: 86466949-86466949 on Build GRCh38
rs751327320 Chr.8: 86466743-86466744 on Build GRCh38
rs533524876 Chr.8: 86466977-86466977 on Build GRCh38
rs573192257 Chr.8: 86466999-86466999 on Build GRCh38
rs766984847 Chr.8: 86466752-86466752 on Build GRCh38
rs138898649 Chr.8: 86466866-86466866 on Build GRCh38
rs757172016 Chr.8: 86466773-86466776 on Build GRCh38
rs561208452 Chr.8: 86467127-86467127 on Build GRCh38
rs774323203 Chr.8: 86466933-86466933 on Build GRCh38
rs975529368 Chr.8: 86467153-86467153 on Build GRCh38
rs559110007 Chr.8: 86467080-86467080 on Build GRCh38
rs777557253 Chr.8: 86466744-86466744 on Build GRCh38
rs986774007 Chr.8: 86466742-86466742 on Build GRCh38
rs56037084 Chr.8: 86466849-86466849 on Build GRCh38
rs185629286 Chr.8: 86467108-86467108 on Build GRCh38
rs892986874 Chr.8: 86466966-86466966 on Build GRCh38
rs1008239741 Chr.8: 86467091-86467091 on Build GRCh38
rs113657203 Chr.8: 86466818-86466818 on Build GRCh38
rs534097464 Chr.8: 86466852-86466852 on Build GRCh38
rs879064876 Chr.8: 86467176-86467176 on Build GRCh38
rs146502552 Chr.8: 86466904-86466904 on Build GRCh38
rs756151238 Chr.8: 86467025-86467025 on Build GRCh38
rs781320893 Chr.8: 86466944-86466944 on Build GRCh38
rs759431081 Chr.8: 86466934-86466934 on Build GRCh38
rs532455262 Chr.8: 86467206-86467206 on Build GRCh38
rs200553123 Chr.8: 86466931-86466931 on Build GRCh38
rs544707617 Chr.8: 86467118-86467118 on Build GRCh38
rs755897749 Chr.8: 86466802-86466802 on Build GRCh38
rs759037149 Chr.8: 86466744-86466744 on Build GRCh38
rs553300847 Chr.8: 86466984-86466984 on Build GRCh38
rs369655213 Chr.8: 86466774-86466774 on Build GRCh38
rs780958519 Chr.8: 86466779-86466782 on Build GRCh38
rs750611365 Chr.8: 86466973-86466973 on Build GRCh38
rs749039371 Chr.8: 86466927-86466927 on Build GRCh38
rs1001202977 Chr.8: 86466952-86466952 on Build GRCh38
rs187543173 Chr.8: 86466900-86466900 on Build GRCh38
rs193278361 Chr.8: 86467021-86467021 on Build GRCh38
rs1033260134 Chr.8: 86466772-86466772 on Build GRCh38

View Full Product Details