Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 36930460-36931001 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CSF2RB
Entrez Gene ID
1439
Gene Name
colony stimulating factor 2 receptor beta common subunit
Gene Aliases
CD131, CDw131, IL3RB, IL5RB, SMDP5
UniGene
Hs.592192
Gene Chromosome Location
Chr.22: 36913574-36940449 on Build GRCh38
Transcripts
NM_000395.2 XM_005261340.3 XM_011529903.2 XM_011529904.2 XM_011529905.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6526520 c.870C>A p.S290S Chr.22: 36930688-36930688 on Build GRCh38
478930 c.965C>G p.S322C Chr.22: 36930783-36930783 on Build GRCh38
1033900 c.916T>C p.C306R Chr.22: 36930734-36930734 on Build GRCh38
6539640 c.810C>G p.S270R Chr.22: 36930466-36930466 on Build GRCh38
1033901 c.987G>T p.E329D Chr.22: 36930805-36930805 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs776142666 Chr.22: 36930771-36930771 on Build GRCh38
rs150251048 Chr.22: 36930759-36930759 on Build GRCh38
rs1021865587 Chr.22: 36930746-36930746 on Build GRCh38
rs116739204 Chr.22: 36930636-36930636 on Build GRCh38
rs144034620 Chr.22: 36930627-36930627 on Build GRCh38
rs781530019 Chr.22: 36930715-36930715 on Build GRCh38
rs201954835 Chr.22: 36930674-36930674 on Build GRCh38
rs576486802 Chr.22: 36930710-36930710 on Build GRCh38
rs376146978 Chr.22: 36930519-36930519 on Build GRCh38
rs896179285 Chr.22: 36930938-36930938 on Build GRCh38
rs748497720 Chr.22: 36930523-36930523 on Build GRCh38
rs189973117 Chr.22: 36930587-36930587 on Build GRCh38
rs756696682 Chr.22: 36930492-36930492 on Build GRCh38
rs752076635 Chr.22: 36930795-36930795 on Build GRCh38
rs777828160 Chr.22: 36930525-36930525 on Build GRCh38
rs938725526 Chr.22: 36930542-36930542 on Build GRCh38
rs376898001 Chr.22: 36930561-36930561 on Build GRCh38
rs200447061 Chr.22: 36930857-36930857 on Build GRCh38
rs758984857 Chr.22: 36930778-36930778 on Build GRCh38
rs201262017 Chr.22: 36930643-36930643 on Build GRCh38
rs151112655 Chr.22: 36930707-36930707 on Build GRCh38
rs571343472 Chr.22: 36930703-36930703 on Build GRCh38
rs745934088 Chr.22: 36930660-36930660 on Build GRCh38
rs373556286 Chr.22: 36930751-36930751 on Build GRCh38
rs759668933 Chr.22: 36930695-36930695 on Build GRCh38
rs375569377 Chr.22: 36930850-36930850 on Build GRCh38
rs749754665 Chr.22: 36930482-36930482 on Build GRCh38
rs747307683 Chr.22: 36930526-36930526 on Build GRCh38
rs372288492 Chr.22: 36930799-36930799 on Build GRCh38
rs928518258 Chr.22: 36930756-36930756 on Build GRCh38
rs755409736 Chr.22: 36930513-36930513 on Build GRCh38
rs760148608 Chr.22: 36930699-36930699 on Build GRCh38
rs903442749 Chr.22: 36930884-36930884 on Build GRCh38
rs79755961 Chr.22: 36930933-36930933 on Build GRCh38
rs370426702 Chr.22: 36930658-36930658 on Build GRCh38
rs761485907 Chr.22: 36930613-36930613 on Build GRCh38
rs767940994 Chr.22: 36930812-36930812 on Build GRCh38
rs1037817162 Chr.22: 36930879-36930879 on Build GRCh38
rs934984707 Chr.22: 36930892-36930892 on Build GRCh38
rs780151719 Chr.22: 36930823-36930823 on Build GRCh38
rs768505622 Chr.22: 36930671-36930671 on Build GRCh38
rs757623965 Chr.22: 36930713-36930713 on Build GRCh38
rs778151079 Chr.22: 36930638-36930638 on Build GRCh38
rs1013276023 Chr.22: 36930964-36930964 on Build GRCh38
rs890683285 Chr.22: 36930975-36930975 on Build GRCh38
rs761525371 Chr.22: 36930685-36930685 on Build GRCh38
rs373990658 Chr.22: 36930550-36930550 on Build GRCh38
rs117805308 Chr.22: 36930752-36930752 on Build GRCh38
rs762241530 Chr.22: 36930800-36930800 on Build GRCh38
rs746137268 Chr.22: 36930721-36930721 on Build GRCh38
rs539051663 Chr.22: 36930661-36930661 on Build GRCh38
rs746813276 Chr.22: 36930868-36930868 on Build GRCh38
rs530551352 Chr.22: 36930571-36930571 on Build GRCh38
rs764516861 Chr.22: 36930709-36930709 on Build GRCh38
rs868859666 Chr.22: 36930895-36930895 on Build GRCh38
rs267606239 Chr.22: 36930672-36930672 on Build GRCh38
rs1043276087 Chr.22: 36930871-36930871 on Build GRCh38
rs776815049 Chr.22: 36930530-36930530 on Build GRCh38
rs749331765 Chr.22: 36930745-36930745 on Build GRCh38
rs749965216 Chr.22: 36930509-36930509 on Build GRCh38
rs767079000 Chr.22: 36930614-36930614 on Build GRCh38
rs1050599256 Chr.22: 36930557-36930557 on Build GRCh38
rs368230425 Chr.22: 36930779-36930779 on Build GRCh38
rs759697603 Chr.22: 36930532-36930532 on Build GRCh38
rs753939956 Chr.22: 36930829-36930829 on Build GRCh38
rs138918081 Chr.22: 36930760-36930760 on Build GRCh38
rs986241313 Chr.22: 36930804-36930804 on Build GRCh38
rs375783135 Chr.22: 36930758-36930758 on Build GRCh38
rs778878463 Chr.22: 36930849-36930849 on Build GRCh38
rs1038250280 Chr.22: 36930612-36930612 on Build GRCh38
rs772326447 Chr.22: 36930519-36930519 on Build GRCh38
rs143834108 Chr.22: 36930496-36930496 on Build GRCh38
rs750845271 Chr.22: 36930813-36930813 on Build GRCh38
rs772019495 Chr.22: 36930853-36930853 on Build GRCh38
rs756424237 Chr.22: 36930727-36930727 on Build GRCh38
rs751261357 Chr.22: 36930488-36930488 on Build GRCh38
rs547780142 Chr.22: 36930609-36930609 on Build GRCh38
rs771081852 Chr.22: 36930527-36930527 on Build GRCh38
rs749198467 Chr.22: 36930664-36930664 on Build GRCh38
rs747969899 Chr.22: 36930757-36930757 on Build GRCh38
rs757436084 Chr.22: 36930654-36930654 on Build GRCh38
rs775632483 Chr.22: 36930558-36930558 on Build GRCh38
rs555140256 Chr.22: 36930766-36930766 on Build GRCh38
rs775542545 Chr.22: 36930662-36930662 on Build GRCh38
rs371290321 Chr.22: 36930484-36930484 on Build GRCh38
rs375358994 Chr.22: 36930628-36930628 on Build GRCh38
rs1030675779 Chr.22: 36930692-36930692 on Build GRCh38
rs755079110 Chr.22: 36930840-36930840 on Build GRCh38
rs984721671 Chr.22: 36930750-36930750 on Build GRCh38
rs765923544 Chr.22: 36930705-36930705 on Build GRCh38
rs1011820968 Chr.22: 36930588-36930588 on Build GRCh38
rs774127718 Chr.22: 36930677-36930677 on Build GRCh38
rs767143075 Chr.22: 36930687-36930687 on Build GRCh38
rs756336062 Chr.22: 36930820-36930820 on Build GRCh38
rs374545094 Chr.22: 36930859-36930859 on Build GRCh38
rs770515128 Chr.22: 36930878-36930878 on Build GRCh38

View Full Product Details