Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 207470912-207471473 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CR2
Entrez Gene ID
1380
Gene Name
complement component 3d receptor 2
Gene Aliases
C3DR, CD21, CR, CVID7, SLEB9
UniGene
Hs.445757
Gene Chromosome Location
Chr.1: 207454300-207489895 on Build GRCh38
Transcripts
NM_001006658.2 NM_001877.4 XM_011509206.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7078556 c.1405G>T p.A469S Chr.1: 207470999-207470999 on Build GRCh38
3864320 c.1532G>A p.G511D Chr.1: 207471461-207471461 on Build GRCh38
5794378 c.1470T>A p.D490E Chr.1: 207471064-207471064 on Build GRCh38
5529180 c.1452C>T p.H484H Chr.1: 207471046-207471046 on Build GRCh38
3482644 c.1544G>A p.W515* Chr.1: 207471473-207471473 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs761478463 Chr.1: 207471108-207471108 on Build GRCh38
rs371884343 Chr.1: 207471247-207471247 on Build GRCh38
rs753628062 Chr.1: 207471019-207471019 on Build GRCh38
rs770418433 Chr.1: 207471070-207471070 on Build GRCh38
rs745928267 Chr.1: 207471428-207471428 on Build GRCh38
rs150388945 Chr.1: 207471406-207471406 on Build GRCh38
rs983686928 Chr.1: 207471238-207471238 on Build GRCh38
rs761376121 Chr.1: 207470983-207470983 on Build GRCh38
rs770988364 Chr.1: 207470958-207470958 on Build GRCh38
rs776195185 Chr.1: 207471084-207471084 on Build GRCh38
rs908994968 Chr.1: 207471242-207471242 on Build GRCh38
rs766201638 Chr.1: 207471343-207471343 on Build GRCh38
rs754918247 Chr.1: 207471385-207471385 on Build GRCh38
rs201463658 Chr.1: 207471441-207471441 on Build GRCh38
rs370217966 Chr.1: 207470959-207470959 on Build GRCh38
rs932915622 Chr.1: 207471298-207471298 on Build GRCh38
rs773232784 Chr.1: 207471329-207471329 on Build GRCh38
rs1037310416 Chr.1: 207471222-207471222 on Build GRCh38
rs771075787 Chr.1: 207471064-207471064 on Build GRCh38
rs529916567 Chr.1: 207471227-207471227 on Build GRCh38
rs770469733 Chr.1: 207470974-207470974 on Build GRCh38
rs781333289 Chr.1: 207471407-207471407 on Build GRCh38
rs545091003 Chr.1: 207471003-207471003 on Build GRCh38
rs1051949332 Chr.1: 207471334-207471334 on Build GRCh38
rs751165557 Chr.1: 207470981-207470981 on Build GRCh38
rs866712862 Chr.1: 207471202-207471202 on Build GRCh38
rs916035659 Chr.1: 207471129-207471129 on Build GRCh38
rs778039241 Chr.1: 207470949-207470949 on Build GRCh38
rs143872741 Chr.1: 207471062-207471062 on Build GRCh38
rs753721175 Chr.1: 207471379-207471379 on Build GRCh38
rs763216546 Chr.1: 207471440-207471440 on Build GRCh38
rs766293897 Chr.1: 207471376-207471376 on Build GRCh38
rs145359041 Chr.1: 207471144-207471144 on Build GRCh38
rs760248782 Chr.1: 207471375-207471375 on Build GRCh38
rs904218818 Chr.1: 207471429-207471429 on Build GRCh38
rs567049053 Chr.1: 207471271-207471271 on Build GRCh38
rs771942333 Chr.1: 207471199-207471199 on Build GRCh38
rs372251272 Chr.1: 207471403-207471403 on Build GRCh38
rs780027591 Chr.1: 207470950-207470950 on Build GRCh38
rs921570935 Chr.1: 207471297-207471297 on Build GRCh38
rs182176991 Chr.1: 207471338-207471338 on Build GRCh38
rs142212472 Chr.1: 207471069-207471069 on Build GRCh38
rs749132096 Chr.1: 207470933-207470933 on Build GRCh38
rs376278193 Chr.1: 207470975-207470975 on Build GRCh38
rs368563787 Chr.1: 207470976-207470976 on Build GRCh38
rs371167100 Chr.1: 207470984-207470984 on Build GRCh38
rs763823010 Chr.1: 207470980-207470980 on Build GRCh38
rs774611422 Chr.1: 207471105-207471105 on Build GRCh38
rs749924559 Chr.1: 207470997-207470997 on Build GRCh38
rs549973713 Chr.1: 207471248-207471248 on Build GRCh38
rs375667838 Chr.1: 207471098-207471098 on Build GRCh38
rs909899786 Chr.1: 207471173-207471173 on Build GRCh38
rs565317532 Chr.1: 207471004-207471004 on Build GRCh38
rs377088084 Chr.1: 207470953-207470953 on Build GRCh38
rs147378770 Chr.1: 207471052-207471052 on Build GRCh38
rs767126107 Chr.1: 207471121-207471121 on Build GRCh38
rs959926708 Chr.1: 207471122-207471122 on Build GRCh38
rs368256473 Chr.1: 207471386-207471386 on Build GRCh38
rs781307709 Chr.1: 207471066-207471066 on Build GRCh38
rs752479354 Chr.1: 207471396-207471396 on Build GRCh38
rs551964587 Chr.1: 207471341-207471341 on Build GRCh38
rs754830128 Chr.1: 207471026-207471026 on Build GRCh38
rs756203331 Chr.1: 207471437-207471437 on Build GRCh38
rs760035052 Chr.1: 207470970-207470970 on Build GRCh38
rs200000368 Chr.1: 207470956-207470956 on Build GRCh38
rs749825341 Chr.1: 207471097-207471097 on Build GRCh38
rs763451084 Chr.1: 207471280-207471280 on Build GRCh38
rs146833647 Chr.1: 207471149-207471149 on Build GRCh38
rs768417104 Chr.1: 207470940-207470940 on Build GRCh38
rs891263800 Chr.1: 207471072-207471072 on Build GRCh38
rs772749374 Chr.1: 207471123-207471123 on Build GRCh38
rs940233901 Chr.1: 207471174-207471174 on Build GRCh38
rs950554454 Chr.1: 207471231-207471231 on Build GRCh38

View Full Product Details