Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 157787389-157787951 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CLINT1
Entrez Gene ID
9685
Gene Name
clathrin interactor 1
Gene Aliases
CLINT, ENTH, EPN4, EPNR
UniGene
Hs.644000
Gene Chromosome Location
Chr.5: 157785743-157859175 on Build GRCh38
Transcripts
NM_001195555.1 NM_001195556.1 NM_014666.3 XM_011534701.2 XM_017010087.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6226245 c.1613G>A p.R538Q Chr.5: 157787911-157787911 on Build GRCh38
4946021 c.1666G>A p.V556M Chr.5: 157787858-157787858 on Build GRCh38
4781147 c.1762G>A p.A588T Chr.5: 157787762-157787762 on Build GRCh38
3669438 c.1865A>G p.N622S Chr.5: 157787659-157787659 on Build GRCh38
5152198 c.1819T>A p.S607T Chr.5: 157787705-157787705 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs34428070 Chr.5: 157787517-157787517 on Build GRCh38
rs748730282 Chr.5: 157787912-157787912 on Build GRCh38
rs377094963 Chr.5: 157787814-157787814 on Build GRCh38
rs746112729 Chr.5: 157787710-157787710 on Build GRCh38
rs750728181 Chr.5: 157787779-157787780 on Build GRCh38
rs184795188 Chr.5: 157787664-157787664 on Build GRCh38
rs755250645 Chr.5: 157787885-157787885 on Build GRCh38
rs756609846 Chr.5: 157787910-157787910 on Build GRCh38
rs766183796 Chr.5: 157787659-157787659 on Build GRCh38
rs753045326 Chr.5: 157787899-157787899 on Build GRCh38
rs781598188 Chr.5: 157787891-157787891 on Build GRCh38
rs180918874 Chr.5: 157787801-157787801 on Build GRCh38
rs185181472 Chr.5: 157787812-157787812 on Build GRCh38
rs202225711 Chr.5: 157787860-157787860 on Build GRCh38
rs777235612 Chr.5: 157787818-157787818 on Build GRCh38
rs1046493613 Chr.5: 157787915-157787915 on Build GRCh38
rs780532366 Chr.5: 157787714-157787714 on Build GRCh38
rs769326861 Chr.5: 157787746-157787746 on Build GRCh38
rs201102111 Chr.5: 157787754-157787754 on Build GRCh38
rs759210327 Chr.5: 157787834-157787834 on Build GRCh38
rs765382730 Chr.5: 157787789-157787789 on Build GRCh38
rs751842472 Chr.5: 157787882-157787882 on Build GRCh38
rs758692113 Chr.5: 157787798-157787798 on Build GRCh38
rs201922408 Chr.5: 157787815-157787815 on Build GRCh38
rs767548618 Chr.5: 157787663-157787663 on Build GRCh38
rs200566884 Chr.5: 157787662-157787662 on Build GRCh38
rs907933281 Chr.5: 157787490-157787490 on Build GRCh38
rs751406081 Chr.5: 157787661-157787661 on Build GRCh38
rs747477230 Chr.5: 157787718-157787718 on Build GRCh38
rs958216942 Chr.5: 157787421-157787421 on Build GRCh38
rs757362102 Chr.5: 157787693-157787693 on Build GRCh38
rs959123305 Chr.5: 157787864-157787864 on Build GRCh38
rs12283 Chr.5: 157787426-157787426 on Build GRCh38
rs749542673 Chr.5: 157787414-157787414 on Build GRCh38
rs928806451 Chr.5: 157787569-157787569 on Build GRCh38
rs779234554 Chr.5: 157787701-157787701 on Build GRCh38
rs962106806 Chr.5: 157787500-157787500 on Build GRCh38
rs377044151 Chr.5: 157787831-157787831 on Build GRCh38
rs751816685 Chr.5: 157787805-157787805 on Build GRCh38
rs372720105 Chr.5: 157787865-157787865 on Build GRCh38
rs781730711 Chr.5: 157787731-157787731 on Build GRCh38
rs762699033 Chr.5: 157787640-157787640 on Build GRCh38
rs769249684 Chr.5: 157787620-157787620 on Build GRCh38
rs367625780 Chr.5: 157787669-157787669 on Build GRCh38
rs771920075 Chr.5: 157787839-157787839 on Build GRCh38
rs190185289 Chr.5: 157787427-157787427 on Build GRCh38
rs760632370 Chr.5: 157787705-157787705 on Build GRCh38
rs190825502 Chr.5: 157787820-157787820 on Build GRCh38
rs778264324 Chr.5: 157787911-157787911 on Build GRCh38
rs989567985 Chr.5: 157787424-157787424 on Build GRCh38
rs573560187 Chr.5: 157787437-157787437 on Build GRCh38
rs566288936 Chr.5: 157787449-157787449 on Build GRCh38
rs752708518 Chr.5: 157787783-157787783 on Build GRCh38
rs750520152 Chr.5: 157787863-157787863 on Build GRCh38
rs369396378 Chr.5: 157787784-157787784 on Build GRCh38
rs773009783 Chr.5: 157787637-157787637 on Build GRCh38
rs769033835 Chr.5: 157787727-157787727 on Build GRCh38
rs759387255 Chr.5: 157787765-157787765 on Build GRCh38
rs774003462 Chr.5: 157787764-157787764 on Build GRCh38
rs1019461475 Chr.5: 157787429-157787429 on Build GRCh38
rs756791030 Chr.5: 157787913-157787913 on Build GRCh38
rs188009135 Chr.5: 157787758-157787758 on Build GRCh38
rs376089257 Chr.5: 157787766-157787766 on Build GRCh38
rs991705574 Chr.5: 157787867-157787867 on Build GRCh38
rs563718460 Chr.5: 157787844-157787844 on Build GRCh38
rs766645747 Chr.5: 157787879-157787879 on Build GRCh38
rs1026928396 Chr.5: 157787464-157787464 on Build GRCh38
rs750568950 Chr.5: 157787795-157787795 on Build GRCh38
rs377104989 Chr.5: 157787922-157787922 on Build GRCh38
rs908360023 Chr.5: 157787904-157787904 on Build GRCh38
rs80062703 Chr.5: 157787460-157787460 on Build GRCh38
rs760654973 Chr.5: 157787845-157787845 on Build GRCh38
rs772805192 Chr.5: 157787747-157787747 on Build GRCh38
rs181091881 Chr.5: 157787479-157787479 on Build GRCh38
rs540805308 Chr.5: 157787467-157787467 on Build GRCh38
rs770337032 Chr.5: 157787816-157787816 on Build GRCh38
rs887228735 Chr.5: 157787522-157787522 on Build GRCh38
rs1007994432 Chr.5: 157787757-157787757 on Build GRCh38
rs752025740 Chr.5: 157787920-157787920 on Build GRCh38
rs764741309 Chr.5: 157787633-157787633 on Build GRCh38
rs747447457 Chr.5: 157787619-157787619 on Build GRCh38
rs369227922 Chr.5: 157787455-157787455 on Build GRCh38
rs971410587 Chr.5: 157787412-157787412 on Build GRCh38
rs146832675 Chr.5: 157787419-157787419 on Build GRCh38
rs556938195 Chr.5: 157787622-157787622 on Build GRCh38
rs747547797 Chr.5: 157787738-157787738 on Build GRCh38
rs553026541 Chr.5: 157787546-157787546 on Build GRCh38
rs762572549 Chr.5: 157787750-157787750 on Build GRCh38
rs914332652 Chr.5: 157787425-157787425 on Build GRCh38
rs778377249 Chr.5: 157787919-157787919 on Build GRCh38
rs758738942 Chr.5: 157787713-157787713 on Build GRCh38
rs1048249436 Chr.5: 157787511-157787511 on Build GRCh38
rs761447123 Chr.5: 157787624-157787624 on Build GRCh38
rs931140606 Chr.5: 157787488-157787488 on Build GRCh38

View Full Product Details