Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 72228427-72228995 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
INPPL1
Entrez Gene ID
3636
Gene Name
inositol polyphosphate phosphatase like 1
Gene Aliases
OPSMD, SHIP2
UniGene
Hs.523875
Gene Chromosome Location
Chr.11: 72223861-72239147 on Build GRCh38
Transcripts
NM_001567.3 XM_005273978.3 XM_005273979.4 XM_011544999.2 XM_011545000.2 XM_017017673.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6480051 c.414G>A p.K138K Chr.11: 72228743-72228743 on Build GRCh38
5601636 c.456C>T p.A152A Chr.11: 72228785-72228785 on Build GRCh38
931660 c.491A>C p.E164A Chr.11: 72228820-72228820 on Build GRCh38
195545 c.429G>A p.P143P Chr.11: 72228758-72228758 on Build GRCh38
7282713 c.378G>A p.P126P Chr.11: 72228479-72228479 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs773393276 Chr.11: 72228456-72228456 on Build GRCh38
rs768035194 Chr.11: 72228780-72228780 on Build GRCh38
rs536046056 Chr.11: 72228617-72228617 on Build GRCh38
rs1048595584 Chr.11: 72228641-72228641 on Build GRCh38
rs760937587 Chr.11: 72228458-72228458 on Build GRCh38
rs778834790 Chr.11: 72228749-72228749 on Build GRCh38
rs776894351 Chr.11: 72228523-72228523 on Build GRCh38
rs762267711 Chr.11: 72228779-72228779 on Build GRCh38
rs756549919 Chr.11: 72228541-72228541 on Build GRCh38
rs920419704 Chr.11: 72228594-72228594 on Build GRCh38
rs56353029 Chr.11: 72228861-72228861 on Build GRCh38
rs767268219 Chr.11: 72228691-72228691 on Build GRCh38
rs370550151 Chr.11: 72228857-72228857 on Build GRCh38
rs778816094 Chr.11: 72228807-72228807 on Build GRCh38
rs984278353 Chr.11: 72228929-72228929 on Build GRCh38
rs753208760 Chr.11: 72228468-72228468 on Build GRCh38
rs1032175507 Chr.11: 72228854-72228854 on Build GRCh38
rs528612645 Chr.11: 72228478-72228478 on Build GRCh38
rs572825150 Chr.11: 72228773-72228773 on Build GRCh38
rs774083889 Chr.11: 72228507-72228507 on Build GRCh38
rs969186382 Chr.11: 72228951-72228951 on Build GRCh38
rs139756712 Chr.11: 72228469-72228469 on Build GRCh38
rs745317721 Chr.11: 72228486-72228486 on Build GRCh38
rs777492773 Chr.11: 72228827-72228827 on Build GRCh38
rs1031114646 Chr.11: 72228510-72228512 on Build GRCh38
rs376366065 Chr.11: 72228696-72228696 on Build GRCh38
rs942176300 Chr.11: 72228850-72228850 on Build GRCh38
rs768403979 Chr.11: 72228447-72228447 on Build GRCh38
rs759655328 Chr.11: 72228532-72228532 on Build GRCh38
rs538502034 Chr.11: 72228548-72228548 on Build GRCh38
rs749317647 Chr.11: 72228855-72228855 on Build GRCh38
rs752670331 Chr.11: 72228811-72228811 on Build GRCh38
rs1023315114 Chr.11: 72228947-72228947 on Build GRCh38
rs930093088 Chr.11: 72228618-72228618 on Build GRCh38
rs1041030521 Chr.11: 72228748-72228748 on Build GRCh38
rs200615694 Chr.11: 72228536-72228536 on Build GRCh38
rs1030220603 Chr.11: 72228793-72228793 on Build GRCh38
rs764732670 Chr.11: 72228547-72228547 on Build GRCh38
rs779479923 Chr.11: 72228706-72228706 on Build GRCh38
rs774359053 Chr.11: 72228895-72228895 on Build GRCh38
rs368460863 Chr.11: 72228810-72228810 on Build GRCh38
rs769788336 Chr.11: 72228928-72228928 on Build GRCh38
rs771919378 Chr.11: 72228755-72228755 on Build GRCh38
rs763316860 Chr.11: 72228546-72228546 on Build GRCh38
rs746849326 Chr.11: 72228828-72228828 on Build GRCh38
rs916336133 Chr.11: 72228525-72228525 on Build GRCh38
rs201045134 Chr.11: 72228772-72228772 on Build GRCh38
rs1048473891 Chr.11: 72228644-72228644 on Build GRCh38
rs754540210 Chr.11: 72228802-72228802 on Build GRCh38
rs768475256 Chr.11: 72228505-72228505 on Build GRCh38
rs199991260 Chr.11: 72228833-72228833 on Build GRCh38
rs749225658 Chr.11: 72228495-72228495 on Build GRCh38
rs770179572 Chr.11: 72228715-72228715 on Build GRCh38
rs999772869 Chr.11: 72228829-72228829 on Build GRCh38
rs918801512 Chr.11: 72228878-72228878 on Build GRCh38
rs758164532 Chr.11: 72228816-72228816 on Build GRCh38
rs753229253 Chr.11: 72228798-72228798 on Build GRCh38
rs61735633 Chr.11: 72228830-72228830 on Build GRCh38
rs902447558 Chr.11: 72228823-72228823 on Build GRCh38
rs200094851 Chr.11: 72228461-72228461 on Build GRCh38
rs760197203 Chr.11: 72228794-72228794 on Build GRCh38
rs202044589 Chr.11: 72228484-72228484 on Build GRCh38
rs773663316 Chr.11: 72228760-72228760 on Build GRCh38
rs371726889 Chr.11: 72228513-72228513 on Build GRCh38
rs1000207551 Chr.11: 72228509-72228509 on Build GRCh38
rs966495240 Chr.11: 72228531-72228531 on Build GRCh38
rs916614738 Chr.11: 72228552-72228552 on Build GRCh38
rs201929168 Chr.11: 72228834-72228834 on Build GRCh38
rs999276105 Chr.11: 72228825-72228825 on Build GRCh38
rs145795148 Chr.11: 72228479-72228479 on Build GRCh38
rs753411829 Chr.11: 72228729-72228729 on Build GRCh38
rs887520792 Chr.11: 72228667-72228667 on Build GRCh38
rs949650901 Chr.11: 72228560-72228560 on Build GRCh38
rs765269255 Chr.11: 72228534-72228534 on Build GRCh38
rs970723867 Chr.11: 72228866-72228866 on Build GRCh38
rs549746015 Chr.11: 72228674-72228674 on Build GRCh38
rs755620537 Chr.11: 72228700-72228700 on Build GRCh38
rs146603018 Chr.11: 72228737-72228737 on Build GRCh38
rs778377922 Chr.11: 72228853-72228854 on Build GRCh38
rs772728657 Chr.11: 72228867-72228867 on Build GRCh38
rs765934266 Chr.11: 72228882-72228882 on Build GRCh38
rs201835641 Chr.11: 72228856-72228856 on Build GRCh38
rs930248479 Chr.11: 72228913-72228913 on Build GRCh38
rs748448649 Chr.11: 72228567-72228567 on Build GRCh38
rs568856228 Chr.11: 72228579-72228579 on Build GRCh38
rs918846796 Chr.11: 72228601-72228601 on Build GRCh38
rs766088509 Chr.11: 72228796-72228796 on Build GRCh38
rs113871062 Chr.11: 72228788-72228788 on Build GRCh38
rs773408975 Chr.11: 72228874-72228874 on Build GRCh38
rs761113986 Chr.11: 72228876-72228876 on Build GRCh38
rs376049368 Chr.11: 72228753-72228753 on Build GRCh38
rs17847221 Chr.11: 72228551-72228551 on Build GRCh38
rs781357595 Chr.11: 72228757-72228757 on Build GRCh38
rs769232228 Chr.11: 72228768-72228768 on Build GRCh38
rs373971775 Chr.11: 72228463-72228463 on Build GRCh38
rs141059764 Chr.11: 72228765-72228765 on Build GRCh38
rs764410685 Chr.11: 72228475-72228475 on Build GRCh38
rs771083237 Chr.11: 72228521-72228521 on Build GRCh38
rs755641145 Chr.11: 72228487-72228487 on Build GRCh38
rs563962020 Chr.11: 72228891-72228891 on Build GRCh38

View Full Product Details