Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 37388546-37388833 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ACACA
Entrez Gene ID
31
Gene Name
acetyl-CoA carboxylase alpha
Gene Aliases
ACAC, ACACAD, ACC, ACC1, ACCA
UniGene
Hs.160556
Gene Chromosome Location
Chr.17: 37084992-37406822 on Build GRCh38
Transcripts
NM_198834.2 NM_198839.2 XM_006721853.1 XM_011524701.1 XM_017024553.1 XM_017024554.1 NM_001321399.1 NM_173625.4 XM_011524647.2 XM_011524648.2 XM_011524649.2

Gene Symbol
C17orf78
Entrez Gene ID
284099
Gene Name
chromosome 17 open reading frame 78
Gene Aliases
-
UniGene
Hs.439154
Gene Chromosome Location
Chr.17: 37375710-37392708 on Build GRCh38
Transcripts
NM_198834.2 NM_198839.2 XM_006721853.1 XM_011524701.1 XM_017024553.1 XM_017024554.1 NM_001321399.1 NM_173625.4 XM_011524647.2 XM_011524648.2 XM_011524649.2

SNP Detail

SNP ID Chromosome Location
rs755385153 Chr.17: 37388696-37388696 on Build GRCh38
rs775920808 Chr.17: 37388771-37388771 on Build GRCh38
rs765417408 Chr.17: 37388656-37388656 on Build GRCh38
rs752801792 Chr.17: 37388658-37388658 on Build GRCh38
rs999796149 Chr.17: 37388756-37388756 on Build GRCh38
rs967106722 Chr.17: 37388677-37388677 on Build GRCh38
rs543104882 Chr.17: 37388734-37388734 on Build GRCh38
rs778491785 Chr.17: 37388709-37388709 on Build GRCh38
rs927599404 Chr.17: 37388577-37388577 on Build GRCh38
rs777024499 Chr.17: 37388759-37388759 on Build GRCh38
rs527957387 Chr.17: 37388789-37388789 on Build GRCh38
rs778196965 Chr.17: 37388717-37388717 on Build GRCh38
rs763058503 Chr.17: 37388772-37388772 on Build GRCh38
rs1040054663 Chr.17: 37388597-37388597 on Build GRCh38
rs991070877 Chr.17: 37388788-37388788 on Build GRCh38
rs370782155 Chr.17: 37388794-37388794 on Build GRCh38
rs750389019 Chr.17: 37388690-37388690 on Build GRCh38
rs960382689 Chr.17: 37388629-37388629 on Build GRCh38
rs767556417 Chr.17: 37388673-37388673 on Build GRCh38
rs776669509 Chr.17: 37388643-37388643 on Build GRCh38
rs759834542 Chr.17: 37388644-37388644 on Build GRCh38
rs748559497 Chr.17: 37388807-37388807 on Build GRCh38
rs367629966 Chr.17: 37388712-37388712 on Build GRCh38
rs753388785 Chr.17: 37388702-37388702 on Build GRCh38
rs779917611 Chr.17: 37388701-37388701 on Build GRCh38
rs755804849 Chr.17: 37388698-37388698 on Build GRCh38
rs900985469 Chr.17: 37388604-37388604 on Build GRCh38
rs746267957 Chr.17: 37388764-37388764 on Build GRCh38
rs998013080 Chr.17: 37388652-37388652 on Build GRCh38
rs770043925 Chr.17: 37388770-37388770 on Build GRCh38
rs763182987 Chr.17: 37388664-37388664 on Build GRCh38
rs747538887 Chr.17: 37388721-37388721 on Build GRCh38
rs373981919 Chr.17: 37388786-37388786 on Build GRCh38
rs771577934 Chr.17: 37388716-37388716 on Build GRCh38
rs754623510 Chr.17: 37388703-37388703 on Build GRCh38
rs562245823 Chr.17: 37388623-37388623 on Build GRCh38

View Full Product Details