Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 1472706-1473295 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
APC2
Entrez Gene ID
10297
Gene Name
APC2, WNT signaling pathway regulator
Gene Aliases
APCL
UniGene
Hs.446376
Gene Chromosome Location
Chr.19: 1446268-1473244 on Build GRCh38
Transcripts
NM_005883.2 XM_005259475.2 XM_006722607.2 XM_006722608.3 XM_006722609.3 XM_006722610.3 NM_152482.2 XM_005259506.3 XM_006722653.3 XM_017026374.1

Gene Symbol
C19orf25
Entrez Gene ID
148223
Gene Name
chromosome 19 open reading frame 25
Gene Aliases
-
UniGene
Hs.532840
Gene Chromosome Location
Chr.19: 1473201-1479552 on Build GRCh38
Transcripts
NM_005883.2 XM_005259475.2 XM_006722607.2 XM_006722608.3 XM_006722609.3 XM_006722610.3 NM_152482.2 XM_005259506.3 XM_006722653.3 XM_017026374.1

SNP Detail

SNP ID Chromosome Location
rs536144836 Chr.19: 1472766-1472766 on Build GRCh38
rs1010495395 Chr.19: 1473140-1473140 on Build GRCh38
rs922586346 Chr.19: 1472727-1472727 on Build GRCh38
rs1006157634 Chr.19: 1473139-1473139 on Build GRCh38
rs993291178 Chr.19: 1473186-1473186 on Build GRCh38
rs896316894 Chr.19: 1472865-1472865 on Build GRCh38
rs148353673 Chr.19: 1472920-1472920 on Build GRCh38
rs911510093 Chr.19: 1472937-1472937 on Build GRCh38
rs1056342894 Chr.19: 1472859-1472859 on Build GRCh38
rs962161000 Chr.19: 1473027-1473027 on Build GRCh38
rs756722055 Chr.19: 1472991-1472991 on Build GRCh38
rs560045772 Chr.19: 1472737-1472737 on Build GRCh38
rs902667330 Chr.19: 1472957-1472957 on Build GRCh38
rs897663039 Chr.19: 1473157-1473157 on Build GRCh38
rs781049140 Chr.19: 1473249-1473249 on Build GRCh38
rs111865101 Chr.19: 1472949-1472949 on Build GRCh38
rs777593390 Chr.19: 1473212-1473212 on Build GRCh38
rs979716262 Chr.19: 1473205-1473205 on Build GRCh38
rs1018288267 Chr.19: 1473195-1473195 on Build GRCh38
rs1038854358 Chr.19: 1473216-1473216 on Build GRCh38
rs1002549337 Chr.19: 1472853-1472853 on Build GRCh38
rs1044740472 Chr.19: 1473119-1473119 on Build GRCh38
rs576369193 Chr.19: 1473130-1473130 on Build GRCh38
rs933938954 Chr.19: 1472915-1472915 on Build GRCh38
rs778287141 Chr.19: 1473084-1473084 on Build GRCh38
rs1014775909 Chr.19: 1473012-1473012 on Build GRCh38
rs753063254 Chr.19: 1472969-1472969 on Build GRCh38
rs924136063 Chr.19: 1473089-1473089 on Build GRCh38
rs564105043 Chr.19: 1473114-1473114 on Build GRCh38
rs999662767 Chr.19: 1472793-1472793 on Build GRCh38
rs935115078 Chr.19: 1472958-1472958 on Build GRCh38
rs947466003 Chr.19: 1472951-1472951 on Build GRCh38
rs1005900390 Chr.19: 1472892-1472892 on Build GRCh38
rs188144078 Chr.19: 1473067-1473067 on Build GRCh38
rs1056550625 Chr.19: 1473106-1473106 on Build GRCh38
rs938719567 Chr.19: 1473073-1473073 on Build GRCh38
rs1043923564 Chr.19: 1472950-1472950 on Build GRCh38
rs992311796 Chr.19: 1473250-1473250 on Build GRCh38
rs548398316 Chr.19: 1472884-1472884 on Build GRCh38
rs985529273 Chr.19: 1473194-1473194 on Build GRCh38
rs11084894 Chr.19: 1473153-1473153 on Build GRCh38
rs1051471557 Chr.19: 1473253-1473253 on Build GRCh38
rs1801822 Chr.19: 1473078-1473078 on Build GRCh38
rs1031025932 Chr.19: 1473017-1473017 on Build GRCh38
rs561879589 Chr.19: 1472837-1472837 on Build GRCh38
rs755176875 Chr.19: 1473009-1473009 on Build GRCh38
rs750126241 Chr.19: 1473150-1473150 on Build GRCh38
rs561050518 Chr.19: 1473053-1473056 on Build GRCh38
rs759672268 Chr.19: 1472953-1472953 on Build GRCh38
rs887413681 Chr.19: 1473136-1473136 on Build GRCh38
rs147542086 Chr.19: 1473045-1473045 on Build GRCh38
rs947017619 Chr.19: 1473083-1473083 on Build GRCh38
rs57455650 Chr.19: 1472736-1472736 on Build GRCh38
rs192689657 Chr.19: 1473210-1473210 on Build GRCh38
rs561819141 Chr.19: 1473151-1473151 on Build GRCh38
rs192627835 Chr.19: 1473178-1473178 on Build GRCh38
rs923541147 Chr.19: 1472930-1472930 on Build GRCh38
rs140154959 Chr.19: 1473094-1473094 on Build GRCh38
rs2656882 Chr.19: 1473099-1473099 on Build GRCh38
rs143869901 Chr.19: 1473228-1473228 on Build GRCh38
rs1017708380 Chr.19: 1472895-1472895 on Build GRCh38
rs538183560 Chr.19: 1472981-1472981 on Build GRCh38
rs922614212 Chr.19: 1472909-1472909 on Build GRCh38
rs768043307 Chr.19: 1472966-1472966 on Build GRCh38
rs887725849 Chr.19: 1472993-1472993 on Build GRCh38
rs265269 Chr.19: 1473197-1473197 on Build GRCh38
rs530182039 Chr.19: 1472869-1472869 on Build GRCh38
rs192549299 Chr.19: 1472829-1472829 on Build GRCh38
rs113611480 Chr.19: 1472753-1472753 on Build GRCh38
rs779844601 Chr.19: 1473177-1473177 on Build GRCh38
rs956232662 Chr.19: 1472943-1472945 on Build GRCh38
rs1056818228 Chr.19: 1472961-1472961 on Build GRCh38
rs570790970 Chr.19: 1472964-1472964 on Build GRCh38
rs985498021 Chr.19: 1473074-1473074 on Build GRCh38
rs973201614 Chr.19: 1472899-1472899 on Build GRCh38
rs773309011 Chr.19: 1472729-1472729 on Build GRCh38
rs983367472 Chr.19: 1473206-1473206 on Build GRCh38
rs964373185 Chr.19: 1472911-1472911 on Build GRCh38
rs572149897 Chr.19: 1473051-1473051 on Build GRCh38
rs557619787 Chr.19: 1473008-1473008 on Build GRCh38
rs1030011890 Chr.19: 1473128-1473128 on Build GRCh38
rs907684898 Chr.19: 1472870-1472870 on Build GRCh38
rs143164872 Chr.19: 1472960-1472960 on Build GRCh38
rs183053282 Chr.19: 1473047-1473047 on Build GRCh38
rs1044275776 Chr.19: 1472764-1472764 on Build GRCh38
rs1011039349 Chr.19: 1472761-1472761 on Build GRCh38
rs58135531 Chr.19: 1472816-1472816 on Build GRCh38
rs549984286 Chr.19: 1472826-1472826 on Build GRCh38
rs564368011 Chr.19: 1472765-1472765 on Build GRCh38
rs918073903 Chr.19: 1473256-1473256 on Build GRCh38
rs947394063 Chr.19: 1472819-1472819 on Build GRCh38
rs764632240 Chr.19: 1472997-1472997 on Build GRCh38
rs1053228794 Chr.19: 1472805-1472805 on Build GRCh38
rs188901242 Chr.19: 1473188-1473188 on Build GRCh38
rs1039235587 Chr.19: 1473118-1473118 on Build GRCh38
rs972842602 Chr.19: 1473043-1473043 on Build GRCh38
rs556305940 Chr.19: 1472983-1472983 on Build GRCh38
rs968834264 Chr.19: 1473048-1473048 on Build GRCh38
rs1011902584 Chr.19: 1472984-1472984 on Build GRCh38
rs955463430 Chr.19: 1473189-1473189 on Build GRCh38
rs947790434 Chr.19: 1473115-1473115 on Build GRCh38
rs997621350 Chr.19: 1473121-1473121 on Build GRCh38
rs1044461742 Chr.19: 1472952-1472952 on Build GRCh38
rs896399189 Chr.19: 1472970-1472970 on Build GRCh38
rs891475946 Chr.19: 1473259-1473259 on Build GRCh38
rs553573991 Chr.19: 1473050-1473050 on Build GRCh38
rs980600974 Chr.19: 1473049-1473050 on Build GRCh38
rs765082312 Chr.19: 1473026-1473026 on Build GRCh38
rs184453408 Chr.19: 1473180-1473180 on Build GRCh38
rs545822390 Chr.19: 1472762-1472762 on Build GRCh38
rs543760656 Chr.19: 1473132-1473132 on Build GRCh38
rs1034720073 Chr.19: 1472798-1472798 on Build GRCh38
rs527816888 Chr.19: 1473181-1473181 on Build GRCh38

View Full Product Details