Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 48039022-48039456 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CABP5
Entrez Gene ID
56344
Gene Name
calcium binding protein 5
Gene Aliases
CABP3
UniGene
Hs.117694
Gene Chromosome Location
Chr.19: 48029383-48048342 on Build GRCh38
Transcripts
NM_019855.4 XM_017026992.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5902049 c.330G>A p.M110I Chr.19: 48039226-48039226 on Build GRCh38
3106885 c.319G>T p.V107F Chr.19: 48039237-48039237 on Build GRCh38
7138303 c.272A>G p.E91G Chr.19: 48039284-48039284 on Build GRCh38
4542137 c.312G>A p.M104I Chr.19: 48039244-48039244 on Build GRCh38
4488402 c.331C>T p.R111W Chr.19: 48039225-48039225 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs952915138 Chr.19: 48039292-48039292 on Build GRCh38
rs878987478 Chr.19: 48039214-48039214 on Build GRCh38
rs753616832 Chr.19: 48039173-48039173 on Build GRCh38
rs756172317 Chr.19: 48039342-48039342 on Build GRCh38
rs1013544219 Chr.19: 48039387-48039387 on Build GRCh38
rs200158927 Chr.19: 48039240-48039240 on Build GRCh38
rs141869209 Chr.19: 48039182-48039182 on Build GRCh38
rs371388887 Chr.19: 48039358-48039358 on Build GRCh38
rs147685459 Chr.19: 48039277-48039277 on Build GRCh38
rs190809812 Chr.19: 48039141-48039141 on Build GRCh38
rs5828329 Chr.19: 48039093-48039093 on Build GRCh38
rs891015 Chr.19: 48039157-48039157 on Build GRCh38
rs374198345 Chr.19: 48039339-48039339 on Build GRCh38
rs889641621 Chr.19: 48039176-48039176 on Build GRCh38
rs916700270 Chr.19: 48039299-48039299 on Build GRCh38
rs748393361 Chr.19: 48039221-48039221 on Build GRCh38
rs774758815 Chr.19: 48039235-48039235 on Build GRCh38
rs768584089 Chr.19: 48039127-48039127 on Build GRCh38
rs765980764 Chr.19: 48039362-48039362 on Build GRCh38
rs778777925 Chr.19: 48039190-48039190 on Build GRCh38
rs368985200 Chr.19: 48039225-48039225 on Build GRCh38
rs760403509 Chr.19: 48039274-48039274 on Build GRCh38
rs562348454 Chr.19: 48039049-48039049 on Build GRCh38
rs377627690 Chr.19: 48039357-48039357 on Build GRCh38
rs773720771 Chr.19: 48039275-48039275 on Build GRCh38
rs1019700044 Chr.19: 48039189-48039189 on Build GRCh38
rs775391933 Chr.19: 48039091-48039091 on Build GRCh38
rs557461807 Chr.19: 48039388-48039388 on Build GRCh38
rs914257531 Chr.19: 48039381-48039381 on Build GRCh38
rs750456 Chr.19: 48039052-48039052 on Build GRCh38
rs777720863 Chr.19: 48039161-48039161 on Build GRCh38
rs781264120 Chr.19: 48039336-48039336 on Build GRCh38
rs760400932 Chr.19: 48039194-48039204 on Build GRCh38
rs369912893 Chr.19: 48039164-48039164 on Build GRCh38
rs1045492617 Chr.19: 48039101-48039101 on Build GRCh38
rs765052835 Chr.19: 48039308-48039308 on Build GRCh38
rs928437821 Chr.19: 48039328-48039328 on Build GRCh38
rs139826866 Chr.19: 48039208-48039208 on Build GRCh38
rs181856283 Chr.19: 48039325-48039325 on Build GRCh38
rs757054510 Chr.19: 48039298-48039298 on Build GRCh38
rs750455 Chr.19: 48039075-48039075 on Build GRCh38
rs931463916 Chr.19: 48039047-48039047 on Build GRCh38
rs991098570 Chr.19: 48039293-48039293 on Build GRCh38
rs1024989918 Chr.19: 48039114-48039114 on Build GRCh38
rs889580241 Chr.19: 48039430-48039430 on Build GRCh38
rs376095493 Chr.19: 48039368-48039368 on Build GRCh38
rs34362586 Chr.19: 48039224-48039224 on Build GRCh38
rs778018878 Chr.19: 48039355-48039355 on Build GRCh38
rs372942092 Chr.19: 48039241-48039241 on Build GRCh38
rs577293433 Chr.19: 48039406-48039406 on Build GRCh38
rs533379420 Chr.19: 48039148-48039148 on Build GRCh38
rs775080126 Chr.19: 48039251-48039251 on Build GRCh38
rs769946190 Chr.19: 48039222-48039222 on Build GRCh38
rs370622082 Chr.19: 48039326-48039326 on Build GRCh38
rs974949323 Chr.19: 48039198-48039198 on Build GRCh38
rs773571363 Chr.19: 48039223-48039223 on Build GRCh38
rs749383840 Chr.19: 48039069-48039069 on Build GRCh38
rs374822522 Chr.19: 48039218-48039218 on Build GRCh38
rs73943516 Chr.19: 48039118-48039118 on Build GRCh38
rs893084229 Chr.19: 48039154-48039154 on Build GRCh38

View Full Product Details