Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 136239133-136239681 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MTFR2
Entrez Gene ID
113115
Gene Name
mitochondrial fission regulator 2
Gene Aliases
DUFD1, FAM54A
UniGene
Hs.121536
Gene Chromosome Location
Chr.6: 136231030-136253659 on Build GRCh38
Transcripts
NM_001099286.2 NM_001318738.1 NM_138419.4 XM_011535410.2 XM_011535412.2 XM_011535413.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6716322 c.790delA p.S264fs*14 Chr.6: 136239545-136239545 on Build GRCh38
4160209 c.691C>A p.P231T Chr.6: 136239644-136239644 on Build GRCh38
234697 c.829C>T p.L277L Chr.6: 136239506-136239506 on Build GRCh38
6716323 c.685G>A p.V229I Chr.6: 136239650-136239650 on Build GRCh38
6219229 c.868C>T p.R290W Chr.6: 136239467-136239467 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs565846288 Chr.6: 136239232-136239232 on Build GRCh38
rs564791862 Chr.6: 136239648-136239648 on Build GRCh38
rs1041685928 Chr.6: 136239191-136239191 on Build GRCh38
rs549219491 Chr.6: 136239218-136239218 on Build GRCh38
rs79006283 Chr.6: 136239369-136239369 on Build GRCh38
rs534952606 Chr.6: 136239291-136239291 on Build GRCh38
rs747717932 Chr.6: 136239499-136239500 on Build GRCh38
rs937491387 Chr.6: 136239163-136239163 on Build GRCh38
rs749548137 Chr.6: 136239568-136239568 on Build GRCh38
rs773326453 Chr.6: 136239625-136239625 on Build GRCh38
rs73779616 Chr.6: 136239306-136239306 on Build GRCh38
rs753103406 Chr.6: 136239549-136239549 on Build GRCh38
rs58671808 Chr.6: 136239479-136239479 on Build GRCh38
rs749351879 Chr.6: 136239472-136239472 on Build GRCh38
rs891095460 Chr.6: 136239200-136239200 on Build GRCh38
rs747532750 Chr.6: 136239424-136239424 on Build GRCh38
rs980849372 Chr.6: 136239398-136239398 on Build GRCh38
rs150862357 Chr.6: 136239537-136239537 on Build GRCh38
rs1039749539 Chr.6: 136239230-136239230 on Build GRCh38
rs1024911733 Chr.6: 136239328-136239328 on Build GRCh38
rs771098969 Chr.6: 136239578-136239578 on Build GRCh38
rs141467777 Chr.6: 136239509-136239509 on Build GRCh38
rs781605204 Chr.6: 136239540-136239540 on Build GRCh38
rs756588535 Chr.6: 136239283-136239283 on Build GRCh38
rs533416130 Chr.6: 136239651-136239651 on Build GRCh38
rs749920087 Chr.6: 136239652-136239652 on Build GRCh38
rs1026860931 Chr.6: 136239301-136239301 on Build GRCh38
rs146249691 Chr.6: 136239650-136239650 on Build GRCh38
rs768354788 Chr.6: 136239629-136239629 on Build GRCh38
rs755291469 Chr.6: 136239438-136239438 on Build GRCh38
rs369622195 Chr.6: 136239466-136239466 on Build GRCh38
rs368808981 Chr.6: 136239510-136239510 on Build GRCh38
rs984541413 Chr.6: 136239330-136239331 on Build GRCh38
rs149305211 Chr.6: 136239583-136239583 on Build GRCh38
rs776226724 Chr.6: 136239640-136239640 on Build GRCh38
rs948342010 Chr.6: 136239401-136239401 on Build GRCh38
rs970397026 Chr.6: 136239362-136239362 on Build GRCh38
rs995346426 Chr.6: 136239281-136239281 on Build GRCh38
rs1018782257 Chr.6: 136239272-136239272 on Build GRCh38
rs866937160 Chr.6: 136239461-136239461 on Build GRCh38
rs199867572 Chr.6: 136239643-136239643 on Build GRCh38
rs762439202 Chr.6: 136239653-136239653 on Build GRCh38
rs756478464 Chr.6: 136239554-136239554 on Build GRCh38
rs750806548 Chr.6: 136239261-136239261 on Build GRCh38
rs990757623 Chr.6: 136239164-136239164 on Build GRCh38
rs754664409 Chr.6: 136239453-136239462 on Build GRCh38
rs770609640 Chr.6: 136239607-136239607 on Build GRCh38
rs778122801 Chr.6: 136239565-136239565 on Build GRCh38
rs926383561 Chr.6: 136239159-136239159 on Build GRCh38
rs990315036 Chr.6: 136239455-136239455 on Build GRCh38
rs770045581 Chr.6: 136239468-136239468 on Build GRCh38
rs751988457 Chr.6: 136239320-136239320 on Build GRCh38
rs1028122860 Chr.6: 136239352-136239352 on Build GRCh38
rs528240471 Chr.6: 136239203-136239203 on Build GRCh38
rs775371775 Chr.6: 136239488-136239488 on Build GRCh38
rs777927099 Chr.6: 136239471-136239471 on Build GRCh38
rs766945649 Chr.6: 136239319-136239319 on Build GRCh38
rs374982682 Chr.6: 136239473-136239473 on Build GRCh38
rs543748793 Chr.6: 136239378-136239379 on Build GRCh38
rs147631179 Chr.6: 136239582-136239582 on Build GRCh38
rs200864358 Chr.6: 136239515-136239515 on Build GRCh38
rs145047825 Chr.6: 136239478-136239478 on Build GRCh38
rs906476868 Chr.6: 136239590-136239590 on Build GRCh38
rs760492794 Chr.6: 136239621-136239621 on Build GRCh38
rs886913582 Chr.6: 136239562-136239562 on Build GRCh38
rs139039815 Chr.6: 136239495-136239495 on Build GRCh38
rs369624300 Chr.6: 136239375-136239375 on Build GRCh38
rs914822867 Chr.6: 136239167-136239167 on Build GRCh38
rs372791220 Chr.6: 136239467-136239467 on Build GRCh38
rs771447481 Chr.6: 136239526-136239527 on Build GRCh38
rs374988588 Chr.6: 136239595-136239595 on Build GRCh38
rs951915771 Chr.6: 136239327-136239327 on Build GRCh38
rs778375122 Chr.6: 136239466-136239469 on Build GRCh38
rs535155884 Chr.6: 136239171-136239171 on Build GRCh38

View Full Product Details