Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 28742909-28743457 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EXTL3
Entrez Gene ID
2137
Gene Name
exostosin like glycosyltransferase 3
Gene Aliases
BOTV, EXTL1L, EXTR1, REGR, RPR
UniGene
Hs.491354
Gene Chromosome Location
Chr.8: 28701473-28753690 on Build GRCh38
Transcripts
NM_001440.3 NR_073468.1 NR_073469.1 XM_011544440.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6113152 c.2461A>G p.I821V Chr.8: 28743125-28743125 on Build GRCh38
6657442 c.2527A>G p.I843V Chr.8: 28743191-28743191 on Build GRCh38
3899448 c.2504C>T p.A835V Chr.8: 28743168-28743168 on Build GRCh38
5976810 c.2485A>G p.I829V Chr.8: 28743149-28743149 on Build GRCh38
6180708 c.2464C>T p.R822W Chr.8: 28743128-28743128 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs932278552 Chr.8: 28742998-28742998 on Build GRCh38
rs201767167 Chr.8: 28743064-28743064 on Build GRCh38
rs2048369 Chr.8: 28743055-28743055 on Build GRCh38
rs778619061 Chr.8: 28743164-28743164 on Build GRCh38
rs765685283 Chr.8: 28743178-28743178 on Build GRCh38
rs531707737 Chr.8: 28743112-28743112 on Build GRCh38
rs376962323 Chr.8: 28743229-28743229 on Build GRCh38
rs756004373 Chr.8: 28743135-28743135 on Build GRCh38
rs779877954 Chr.8: 28743254-28743254 on Build GRCh38
rs919611569 Chr.8: 28742991-28742991 on Build GRCh38
rs755949585 Chr.8: 28743237-28743237 on Build GRCh38
rs767721535 Chr.8: 28743007-28743007 on Build GRCh38
rs764705863 Chr.8: 28743056-28743056 on Build GRCh38
rs985121104 Chr.8: 28743300-28743300 on Build GRCh38
rs145489192 Chr.8: 28743341-28743341 on Build GRCh38
rs777054363 Chr.8: 28743203-28743203 on Build GRCh38
rs748805862 Chr.8: 28743148-28743148 on Build GRCh38
rs758692819 Chr.8: 28743286-28743286 on Build GRCh38
rs546255200 Chr.8: 28743422-28743422 on Build GRCh38
rs551396626 Chr.8: 28743283-28743283 on Build GRCh38
rs745569399 Chr.8: 28743175-28743175 on Build GRCh38
rs148468300 Chr.8: 28743410-28743410 on Build GRCh38
rs777707445 Chr.8: 28743139-28743139 on Build GRCh38
rs773355880 Chr.8: 28743222-28743222 on Build GRCh38
rs550423952 Chr.8: 28743369-28743369 on Build GRCh38
rs111821000 Chr.8: 28743302-28743302 on Build GRCh38
rs1025242546 Chr.8: 28743398-28743398 on Build GRCh38
rs746402794 Chr.8: 28743250-28743250 on Build GRCh38
rs1000230923 Chr.8: 28743339-28743339 on Build GRCh38
rs775933143 Chr.8: 28743181-28743181 on Build GRCh38
rs1012143313 Chr.8: 28743372-28743372 on Build GRCh38
rs145932235 Chr.8: 28743038-28743038 on Build GRCh38
rs747411243 Chr.8: 28743187-28743187 on Build GRCh38
rs766617724 Chr.8: 28743227-28743227 on Build GRCh38
rs530604002 Chr.8: 28743343-28743343 on Build GRCh38
rs201637637 Chr.8: 28743248-28743248 on Build GRCh38
rs199545704 Chr.8: 28743225-28743225 on Build GRCh38
rs959310547 Chr.8: 28743061-28743061 on Build GRCh38
rs767241080 Chr.8: 28743122-28743122 on Build GRCh38
rs759758365 Chr.8: 28743004-28743004 on Build GRCh38
rs983136504 Chr.8: 28743108-28743108 on Build GRCh38
rs1009801099 Chr.8: 28743054-28743055 on Build GRCh38
rs142387575 Chr.8: 28742980-28742980 on Build GRCh38
rs752492460 Chr.8: 28743133-28743133 on Build GRCh38
rs773921650 Chr.8: 28743035-28743035 on Build GRCh38
rs943788958 Chr.8: 28743307-28743307 on Build GRCh38
rs763985368 Chr.8: 28743246-28743246 on Build GRCh38
rs201017741 Chr.8: 28743249-28743249 on Build GRCh38
rs112933134 Chr.8: 28743079-28743079 on Build GRCh38
rs765301641 Chr.8: 28743207-28743207 on Build GRCh38
rs535962761 Chr.8: 28743419-28743419 on Build GRCh38
rs758087395 Chr.8: 28743252-28743252 on Build GRCh38
rs192509717 Chr.8: 28743071-28743071 on Build GRCh38
rs113776760 Chr.8: 28743320-28743320 on Build GRCh38
rs769238546 Chr.8: 28743197-28743197 on Build GRCh38
rs868606371 Chr.8: 28743401-28743401 on Build GRCh38
rs1022944298 Chr.8: 28743054-28743054 on Build GRCh38
rs756753935 Chr.8: 28743150-28743150 on Build GRCh38
rs983231906 Chr.8: 28743416-28743416 on Build GRCh38
rs552529834 Chr.8: 28742950-28742950 on Build GRCh38
rs575749917 Chr.8: 28743039-28743039 on Build GRCh38
rs776131467 Chr.8: 28743053-28743053 on Build GRCh38
rs747921132 Chr.8: 28742937-28742937 on Build GRCh38
rs183182702 Chr.8: 28743205-28743205 on Build GRCh38
rs922183697 Chr.8: 28743193-28743193 on Build GRCh38
rs754788898 Chr.8: 28743105-28743105 on Build GRCh38
rs1037328857 Chr.8: 28743188-28743188 on Build GRCh38
rs942302323 Chr.8: 28742933-28742933 on Build GRCh38
rs771863047 Chr.8: 28742938-28742938 on Build GRCh38
rs538977800 Chr.8: 28742988-28742988 on Build GRCh38
rs879127135 Chr.8: 28742958-28742958 on Build GRCh38
rs760831313 Chr.8: 28743000-28743000 on Build GRCh38
rs749958249 Chr.8: 28743251-28743251 on Build GRCh38
rs959555355 Chr.8: 28743361-28743361 on Build GRCh38
rs375145608 Chr.8: 28742940-28742940 on Build GRCh38

View Full Product Details