Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 128459974-128460546 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ETS1
Entrez Gene ID
2113
Gene Name
ETS proto-oncogene 1, transcription factor
Gene Aliases
ETS-1, EWSR2, c-ets-1, p54
UniGene
Hs.369438
Gene Chromosome Location
Chr.11: 128458761-128587593 on Build GRCh38
Transcripts
NM_001143820.1 NM_001162422.1 NM_005238.3 XM_011542649.2 XM_011542650.1 XM_011542651.1 XM_011542652.1 XM_017017314.1 XM_017017315.1 XM_017017316.1 XM_017017317.1

SNP Detail

SNP ID Chromosome Location
rs369942096 Chr.11: 128460093-128460097 on Build GRCh38
rs1041362097 Chr.11: 128460288-128460288 on Build GRCh38
rs896879948 Chr.11: 128460107-128460107 on Build GRCh38
rs750623611 Chr.11: 128460491-128460491 on Build GRCh38
rs757150817 Chr.11: 128460086-128460090 on Build GRCh38
rs762193361 Chr.11: 128460050-128460050 on Build GRCh38
rs1015303662 Chr.11: 128460102-128460102 on Build GRCh38
rs910089361 Chr.11: 128460344-128460344 on Build GRCh38
rs967916196 Chr.11: 128460194-128460194 on Build GRCh38
rs933013006 Chr.11: 128460332-128460332 on Build GRCh38
rs971441279 Chr.11: 128460091-128460092 on Build GRCh38
rs972750914 Chr.11: 128460054-128460054 on Build GRCh38
rs572855602 Chr.11: 128460354-128460354 on Build GRCh38
rs760335083 Chr.11: 128460115-128460116 on Build GRCh38
rs1050421244 Chr.11: 128460094-128460094 on Build GRCh38
rs981905590 Chr.11: 128460168-128460168 on Build GRCh38
rs796985611 Chr.11: 128460091-128460099 on Build GRCh38
rs571417131 Chr.11: 128460208-128460208 on Build GRCh38
rs57498864 Chr.11: 128460093-128460093 on Build GRCh38
rs990792550 Chr.11: 128460134-128460134 on Build GRCh38
rs577176917 Chr.11: 128460091-128460095 on Build GRCh38
rs1003180381 Chr.11: 128460109-128460109 on Build GRCh38
rs532211917 Chr.11: 128460248-128460248 on Build GRCh38
rs922663714 Chr.11: 128460253-128460253 on Build GRCh38
rs762284553 Chr.11: 128460271-128460271 on Build GRCh38
rs959284909 Chr.11: 128460128-128460128 on Build GRCh38
rs140391188 Chr.11: 128460087-128460088 on Build GRCh38
rs771600375 Chr.11: 128460089-128460090 on Build GRCh38
rs769523235 Chr.11: 128460088-128460089 on Build GRCh38
rs760153304 Chr.11: 128460311-128460311 on Build GRCh38
rs934757653 Chr.11: 128460217-128460217 on Build GRCh38
rs878953222 Chr.11: 128460072-128460079 on Build GRCh38
rs932639701 Chr.11: 128460272-128460272 on Build GRCh38
rs1026090457 Chr.11: 128460076-128460076 on Build GRCh38
rs979916532 Chr.11: 128460273-128460273 on Build GRCh38
rs1017100515 Chr.11: 128459997-128459997 on Build GRCh38
rs1037137597 Chr.11: 128460458-128460458 on Build GRCh38
rs867931177 Chr.11: 128460101-128460101 on Build GRCh38
rs369915212 Chr.11: 128460097-128460097 on Build GRCh38
rs773889631 Chr.11: 128460085-128460086 on Build GRCh38
rs897309833 Chr.11: 128460475-128460475 on Build GRCh38
rs1022593340 Chr.11: 128460168-128460168 on Build GRCh38
rs774630781 Chr.11: 128460099-128460099 on Build GRCh38
rs888866405 Chr.11: 128460392-128460392 on Build GRCh38
rs200782210 Chr.11: 128460090-128460091 on Build GRCh38
rs574009912 Chr.11: 128460004-128460004 on Build GRCh38
rs1050010554 Chr.11: 128460331-128460331 on Build GRCh38
rs924656964 Chr.11: 128460084-128460084 on Build GRCh38
rs546417796 Chr.11: 128460055-128460055 on Build GRCh38
rs867057840 Chr.11: 128460182-128460182 on Build GRCh38
rs544818439 Chr.11: 128460461-128460461 on Build GRCh38
rs773438481 Chr.11: 128460087-128460087 on Build GRCh38
rs4937332 Chr.11: 128460091-128460091 on Build GRCh38
rs888658769 Chr.11: 128460095-128460095 on Build GRCh38
rs775171430 Chr.11: 128460093-128460099 on Build GRCh38
rs142088976 Chr.11: 128460201-128460201 on Build GRCh38
rs1007008855 Chr.11: 128460423-128460423 on Build GRCh38
rs375149732 Chr.11: 128460091-128460091 on Build GRCh38
rs199537006 Chr.11: 128460072-128460077 on Build GRCh38
rs945977028 Chr.11: 128460241-128460241 on Build GRCh38
rs370869273 Chr.11: 128460220-128460220 on Build GRCh38
rs372827891 Chr.11: 128460077-128460077 on Build GRCh38
rs35034701 Chr.11: 128460485-128460485 on Build GRCh38
rs1034670833 Chr.11: 128460111-128460111 on Build GRCh38
rs535815822 Chr.11: 128460063-128460063 on Build GRCh38
rs772424268 Chr.11: 128460360-128460360 on Build GRCh38
rs111556815 Chr.11: 128460089-128460089 on Build GRCh38
rs879025698 Chr.11: 128460076-128460077 on Build GRCh38
rs11221316 Chr.11: 128460097-128460097 on Build GRCh38
rs927371030 Chr.11: 128460187-128460187 on Build GRCh38
rs914486930 Chr.11: 128460222-128460222 on Build GRCh38
rs1050562462 Chr.11: 128460334-128460334 on Build GRCh38
rs756340069 Chr.11: 128460088-128460092 on Build GRCh38
rs751634700 Chr.11: 128460093-128460093 on Build GRCh38

View Full Product Details