Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 176221280-176221860 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ASB5
Entrez Gene ID
140458
Gene Name
ankyrin repeat and SOCS box containing 5
Gene Aliases
ASB-5
UniGene
Hs.591712
Gene Chromosome Location
Chr.4: 176213673-176277571 on Build GRCh38
Transcripts
NM_080874.3 XM_005262759.1 XM_011531616.2 XM_011531617.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1053428 c.515C>T p.T172M Chr.4: 176221470-176221470 on Build GRCh38
3602176 c.388A>C p.N130H Chr.4: 176221597-176221597 on Build GRCh38
7245334 c.472T>A p.Y158N Chr.4: 176221513-176221513 on Build GRCh38
6252486 c.426C>A p.N142K Chr.4: 176221559-176221559 on Build GRCh38
367371 c.467T>A p.L156Q Chr.4: 176221518-176221518 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs988032632 Chr.4: 176221701-176221701 on Build GRCh38
rs963260051 Chr.4: 176221572-176221572 on Build GRCh38
rs554899118 Chr.4: 176221420-176221420 on Build GRCh38
rs71646876 Chr.4: 176221625-176221626 on Build GRCh38
rs946581410 Chr.4: 176221746-176221746 on Build GRCh38
rs143450675 Chr.4: 176221551-176221551 on Build GRCh38
rs138302221 Chr.4: 176221478-176221478 on Build GRCh38
rs17062631 Chr.4: 176221397-176221397 on Build GRCh38
rs772805804 Chr.4: 176221336-176221336 on Build GRCh38
rs748660120 Chr.4: 176221456-176221456 on Build GRCh38
rs950477105 Chr.4: 176221358-176221358 on Build GRCh38
rs28415362 Chr.4: 176221718-176221718 on Build GRCh38
rs938986870 Chr.4: 176221697-176221697 on Build GRCh38
rs537762433 Chr.4: 176221453-176221453 on Build GRCh38
rs149224494 Chr.4: 176221489-176221489 on Build GRCh38
rs116013117 Chr.4: 176221374-176221374 on Build GRCh38
rs139145486 Chr.4: 176221580-176221580 on Build GRCh38
rs201922691 Chr.4: 176221621-176221622 on Build GRCh38
rs544254554 Chr.4: 176221654-176221654 on Build GRCh38
rs570156844 Chr.4: 176221333-176221333 on Build GRCh38
rs746952957 Chr.4: 176221576-176221576 on Build GRCh38
rs545829322 Chr.4: 176221470-176221470 on Build GRCh38
rs150458674 Chr.4: 176221469-176221469 on Build GRCh38
rs1039875558 Chr.4: 176221689-176221689 on Build GRCh38
rs750662802 Chr.4: 176221419-176221419 on Build GRCh38
rs61748176 Chr.4: 176221577-176221577 on Build GRCh38
rs149547059 Chr.4: 176221629-176221629 on Build GRCh38
rs755503780 Chr.4: 176221326-176221326 on Build GRCh38
rs568685114 Chr.4: 176221682-176221682 on Build GRCh38
rs35698542 Chr.4: 176221460-176221460 on Build GRCh38
rs947737951 Chr.4: 176221422-176221422 on Build GRCh38
rs763092481 Chr.4: 176221491-176221491 on Build GRCh38
rs140659869 Chr.4: 176221463-176221463 on Build GRCh38
rs755618829 Chr.4: 176221459-176221459 on Build GRCh38
rs756157079 Chr.4: 176221412-176221412 on Build GRCh38
rs780447709 Chr.4: 176221324-176221324 on Build GRCh38
rs761951702 Chr.4: 176221412-176221412 on Build GRCh38
rs954975397 Chr.4: 176221368-176221368 on Build GRCh38
rs997207364 Chr.4: 176221496-176221496 on Build GRCh38
rs753807366 Chr.4: 176221465-176221465 on Build GRCh38
rs946524128 Chr.4: 176221545-176221545 on Build GRCh38
rs150772149 Chr.4: 176221495-176221495 on Build GRCh38
rs147166381 Chr.4: 176221559-176221559 on Build GRCh38
rs879669342 Chr.4: 176221625-176221626 on Build GRCh38
rs770678956 Chr.4: 176221339-176221339 on Build GRCh38
rs571295277 Chr.4: 176221692-176221692 on Build GRCh38
rs745493207 Chr.4: 176221564-176221564 on Build GRCh38
rs137892131 Chr.4: 176221527-176221527 on Build GRCh38
rs537600832 Chr.4: 176221683-176221683 on Build GRCh38
rs771131643 Chr.4: 176221587-176221587 on Build GRCh38
rs777769231 Chr.4: 176221438-176221441 on Build GRCh38
rs376426121 Chr.4: 176221639-176221639 on Build GRCh38
rs866689789 Chr.4: 176221480-176221480 on Build GRCh38
rs893980211 Chr.4: 176221477-176221477 on Build GRCh38
rs139542032 Chr.4: 176221748-176221748 on Build GRCh38
rs770575247 Chr.4: 176221466-176221466 on Build GRCh38
rs267600101 Chr.4: 176221461-176221461 on Build GRCh38
rs13120838 Chr.4: 176221626-176221626 on Build GRCh38
rs756854906 Chr.4: 176221635-176221635 on Build GRCh38
rs35545715 Chr.4: 176221485-176221485 on Build GRCh38
rs751906631 Chr.4: 176221426-176221426 on Build GRCh38
rs186980118 Chr.4: 176221620-176221620 on Build GRCh38
rs904643930 Chr.4: 176221836-176221836 on Build GRCh38
rs376452908 Chr.4: 176221606-176221606 on Build GRCh38
rs34057345 Chr.4: 176221655-176221656 on Build GRCh38
rs768506681 Chr.4: 176221569-176221569 on Build GRCh38
rs372759383 Chr.4: 176221402-176221402 on Build GRCh38
rs774753293 Chr.4: 176221588-176221588 on Build GRCh38
rs771837324 Chr.4: 176221474-176221474 on Build GRCh38
rs990699062 Chr.4: 176221381-176221381 on Build GRCh38
rs767371930 Chr.4: 176221403-176221403 on Build GRCh38
rs776701520 Chr.4: 176221408-176221408 on Build GRCh38
rs10032098 Chr.4: 176221625-176221625 on Build GRCh38
rs575064687 Chr.4: 176221421-176221421 on Build GRCh38
rs374709990 Chr.4: 176221418-176221418 on Build GRCh38
rs570133744 Chr.4: 176221325-176221325 on Build GRCh38
rs750657946 Chr.4: 176221308-176221308 on Build GRCh38
rs1015101945 Chr.4: 176221390-176221390 on Build GRCh38
rs113209719 Chr.4: 176221655-176221655 on Build GRCh38
rs756592467 Chr.4: 176221548-176221548 on Build GRCh38
rs540114501 Chr.4: 176221813-176221813 on Build GRCh38
rs767963342 Chr.4: 176221536-176221536 on Build GRCh38
rs996613212 Chr.4: 176221560-176221560 on Build GRCh38
rs750102564 Chr.4: 176221638-176221638 on Build GRCh38
rs753319825 Chr.4: 176221553-176221553 on Build GRCh38
rs1030113478 Chr.4: 176221348-176221348 on Build GRCh38
rs367961175 Chr.4: 176221665-176221665 on Build GRCh38
rs963509607 Chr.4: 176221376-176221376 on Build GRCh38
rs763151479 Chr.4: 176221586-176221586 on Build GRCh38
rs564000284 Chr.4: 176221600-176221600 on Build GRCh38
rs1021630943 Chr.4: 176221323-176221323 on Build GRCh38
rs376606057 Chr.4: 176221321-176221321 on Build GRCh38
rs751819078 Chr.4: 176221515-176221515 on Build GRCh38
rs934220670 Chr.4: 176221725-176221725 on Build GRCh38
rs12501772 Chr.4: 176221375-176221375 on Build GRCh38
rs543749428 Chr.4: 176221568-176221568 on Build GRCh38
rs1052265578 Chr.4: 176221475-176221475 on Build GRCh38
rs34466424 Chr.4: 176221337-176221337 on Build GRCh38
rs141181389 Chr.4: 176221341-176221341 on Build GRCh38
rs780855507 Chr.4: 176221332-176221332 on Build GRCh38
rs753304474 Chr.4: 176221537-176221537 on Build GRCh38
rs755556820 Chr.4: 176221431-176221431 on Build GRCh38

View Full Product Details