Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 158893332-158893893 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C4orf45
Entrez Gene ID
152940
Gene Name
chromosome 4 open reading frame 45
Gene Aliases
-
UniGene
Hs.415576
Gene Chromosome Location
Chr.4: 158893532-159082883 on Build GRCh38
Transcripts
NM_152543.2 XM_011531671.2 XM_011531672.2 XM_017007808.1 XM_017007809.1 XM_017007810.1 NM_001323916.1 NM_020840.2 XM_005263156.1 XM_005263158.1 XM_011532154.2 XM_017008487.1

Gene Symbol
FNIP2
Entrez Gene ID
57600
Gene Name
folliculin interacting protein 2
Gene Aliases
FNIPL, MAPO1
UniGene
Hs.652441
Gene Chromosome Location
Chr.4: 158768995-158908050 on Build GRCh38
Transcripts
NM_152543.2 XM_011531671.2 XM_011531672.2 XM_017007808.1 XM_017007809.1 XM_017007810.1 NM_001323916.1 NM_020840.2 XM_005263156.1 XM_005263158.1 XM_011532154.2 XM_017008487.1

SNP Detail

SNP ID Chromosome Location
rs1012533013 Chr.4: 158893852-158893852 on Build GRCh38
rs1006186805 Chr.4: 158893667-158893667 on Build GRCh38
rs953586603 Chr.4: 158893429-158893429 on Build GRCh38
rs924348579 Chr.4: 158893528-158893528 on Build GRCh38
rs749959104 Chr.4: 158893662-158893662 on Build GRCh38
rs755504427 Chr.4: 158893722-158893722 on Build GRCh38
rs547149609 Chr.4: 158893614-158893614 on Build GRCh38
rs559731374 Chr.4: 158893513-158893513 on Build GRCh38
rs527358730 Chr.4: 158893572-158893572 on Build GRCh38
rs372847692 Chr.4: 158893652-158893652 on Build GRCh38
rs749486687 Chr.4: 158893760-158893760 on Build GRCh38
rs773524982 Chr.4: 158893747-158893747 on Build GRCh38
rs902654088 Chr.4: 158893804-158893804 on Build GRCh38
rs986280901 Chr.4: 158893803-158893803 on Build GRCh38
rs1025002537 Chr.4: 158893511-158893511 on Build GRCh38
rs1001051936 Chr.4: 158893808-158893808 on Build GRCh38
rs764929726 Chr.4: 158893793-158893793 on Build GRCh38
rs189547099 Chr.4: 158893729-158893729 on Build GRCh38
rs574434600 Chr.4: 158893373-158893373 on Build GRCh38
rs986331894 Chr.4: 158893437-158893437 on Build GRCh38
rs771999762 Chr.4: 158893764-158893764 on Build GRCh38
rs549411773 Chr.4: 158893759-158893759 on Build GRCh38
rs779508915 Chr.4: 158893684-158893684 on Build GRCh38
rs902686221 Chr.4: 158893386-158893386 on Build GRCh38
rs908346221 Chr.4: 158893551-158893551 on Build GRCh38
rs1048315347 Chr.4: 158893649-158893649 on Build GRCh38
rs764815657 Chr.4: 158893653-158893653 on Build GRCh38
rs757866128 Chr.4: 158893678-158893678 on Build GRCh38
rs369800736 Chr.4: 158893736-158893736 on Build GRCh38
rs767781768 Chr.4: 158893774-158893774 on Build GRCh38
rs752007961 Chr.4: 158893406-158893406 on Build GRCh38
rs747806117 Chr.4: 158893503-158893508 on Build GRCh38
rs894283327 Chr.4: 158893635-158893636 on Build GRCh38
rs961205125 Chr.4: 158893571-158893571 on Build GRCh38
rs919856742 Chr.4: 158893591-158893591 on Build GRCh38
rs1012786544 Chr.4: 158893645-158893645 on Build GRCh38
rs1001082758 Chr.4: 158893387-158893387 on Build GRCh38
rs938420766 Chr.4: 158893545-158893545 on Build GRCh38
rs1021126207 Chr.4: 158893382-158893382 on Build GRCh38
rs1008006345 Chr.4: 158893369-158893369 on Build GRCh38
rs563856075 Chr.4: 158893631-158893631 on Build GRCh38
rs747519186 Chr.4: 158893695-158893695 on Build GRCh38
rs748486644 Chr.4: 158893734-158893734 on Build GRCh38
rs932609974 Chr.4: 158893646-158893646 on Build GRCh38
rs373687039 Chr.4: 158893800-158893800 on Build GRCh38
rs77228157 Chr.4: 158893481-158893481 on Build GRCh38
rs983542306 Chr.4: 158893533-158893533 on Build GRCh38
rs888385559 Chr.4: 158893659-158893659 on Build GRCh38
rs547814176 Chr.4: 158893859-158893859 on Build GRCh38
rs182999722 Chr.4: 158893813-158893813 on Build GRCh38
rs529350466 Chr.4: 158893475-158893475 on Build GRCh38
rs895372303 Chr.4: 158893846-158893846 on Build GRCh38
rs370810602 Chr.4: 158893474-158893474 on Build GRCh38
rs764550424 Chr.4: 158893814-158893814 on Build GRCh38
rs973933845 Chr.4: 158893574-158893574 on Build GRCh38
rs745968996 Chr.4: 158893704-158893706 on Build GRCh38

View Full Product Details