Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 79921500-79922085 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ELOVL4
Entrez Gene ID
6785
Gene Name
ELOVL fatty acid elongase 4
Gene Aliases
ADMD, CT118, ISQMR, SCA34, STGD2, STGD3
UniGene
Hs.101915
Gene Chromosome Location
Chr.6: 79914812-79947598 on Build GRCh38
Transcripts
NM_022726.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5061960 c.515G>T p.G172V Chr.6: 79921651-79921651 on Build GRCh38
3630111 c.380C>T p.A127V Chr.6: 79921786-79921786 on Build GRCh38
70603 c.507G>T p.W169C Chr.6: 79921659-79921659 on Build GRCh38
1081905 c.529G>A p.A177T Chr.6: 79921637-79921637 on Build GRCh38
4900017 c.395T>A p.F132Y Chr.6: 79921771-79921771 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs541690548 Chr.6: 79921589-79921589 on Build GRCh38
rs973821185 Chr.6: 79921737-79921737 on Build GRCh38
rs367615182 Chr.6: 79921608-79921608 on Build GRCh38
rs35163932 Chr.6: 79921913-79921914 on Build GRCh38
rs267601134 Chr.6: 79921674-79921674 on Build GRCh38
rs376188469 Chr.6: 79921596-79921596 on Build GRCh38
rs954200666 Chr.6: 79921807-79921807 on Build GRCh38
rs775696541 Chr.6: 79921849-79921849 on Build GRCh38
rs777096598 Chr.6: 79921609-79921609 on Build GRCh38
rs727503917 Chr.6: 79921699-79921699 on Build GRCh38
rs776610847 Chr.6: 79921845-79921845 on Build GRCh38
rs200484793 Chr.6: 79921644-79921644 on Build GRCh38
rs1002532899 Chr.6: 79921534-79921534 on Build GRCh38
rs183975666 Chr.6: 79921814-79921814 on Build GRCh38
rs987467575 Chr.6: 79921811-79921811 on Build GRCh38
rs775098373 Chr.6: 79921815-79921815 on Build GRCh38
rs1005323181 Chr.6: 79921581-79921581 on Build GRCh38
rs767298447 Chr.6: 79921612-79921612 on Build GRCh38
rs146327666 Chr.6: 79921869-79921869 on Build GRCh38
rs182978670 Chr.6: 79922039-79922039 on Build GRCh38
rs750084918 Chr.6: 79921677-79921677 on Build GRCh38
rs778754549 Chr.6: 79921675-79921675 on Build GRCh38
rs868561155 Chr.6: 79921957-79921957 on Build GRCh38
rs746276823 Chr.6: 79921583-79921583 on Build GRCh38
rs7770585 Chr.6: 79921913-79921913 on Build GRCh38
rs758898884 Chr.6: 79921678-79921678 on Build GRCh38
rs547259613 Chr.6: 79922038-79922038 on Build GRCh38
rs760471444 Chr.6: 79921619-79921619 on Build GRCh38
rs192103032 Chr.6: 79921796-79921796 on Build GRCh38
rs886061802 Chr.6: 79921664-79921664 on Build GRCh38
rs947475426 Chr.6: 79921893-79921893 on Build GRCh38
rs768992562 Chr.6: 79921692-79921692 on Build GRCh38
rs763682551 Chr.6: 79921626-79921626 on Build GRCh38
rs185730504 Chr.6: 79921862-79921862 on Build GRCh38
rs7770586 Chr.6: 79921914-79921914 on Build GRCh38
rs542126507 Chr.6: 79921562-79921562 on Build GRCh38
rs1034715298 Chr.6: 79921978-79921978 on Build GRCh38
rs1040409639 Chr.6: 79921880-79921880 on Build GRCh38
rs752286374 Chr.6: 79921960-79921960 on Build GRCh38
rs1049752838 Chr.6: 79921912-79921912 on Build GRCh38
rs1003693727 Chr.6: 79921974-79921979 on Build GRCh38
rs186000642 Chr.6: 79921564-79921564 on Build GRCh38
rs897804271 Chr.6: 79921889-79921889 on Build GRCh38
rs570505486 Chr.6: 79921996-79921996 on Build GRCh38
rs769438485 Chr.6: 79921731-79921731 on Build GRCh38
rs768785763 Chr.6: 79921591-79921591 on Build GRCh38
rs752688335 Chr.6: 79921618-79921618 on Build GRCh38
rs45466000 Chr.6: 79921565-79921565 on Build GRCh38
rs5877683 Chr.6: 79921962-79921962 on Build GRCh38
rs763980649 Chr.6: 79921827-79921827 on Build GRCh38
rs962483133 Chr.6: 79921676-79921676 on Build GRCh38
rs564561699 Chr.6: 79921698-79921698 on Build GRCh38
rs1004121401 Chr.6: 79922002-79922002 on Build GRCh38
rs981841446 Chr.6: 79921614-79921615 on Build GRCh38
rs769854532 Chr.6: 79921740-79921740 on Build GRCh38
rs1050477538 Chr.6: 79921859-79921859 on Build GRCh38
rs747465851 Chr.6: 79921590-79921590 on Build GRCh38
rs700483 Chr.6: 79921822-79921822 on Build GRCh38
rs550475226 Chr.6: 79922001-79922001 on Build GRCh38
rs780499701 Chr.6: 79921679-79921679 on Build GRCh38
rs1022135846 Chr.6: 79922005-79922005 on Build GRCh38
rs749448670 Chr.6: 79921804-79921804 on Build GRCh38
rs376142044 Chr.6: 79921841-79921841 on Build GRCh38
rs747560791 Chr.6: 79921680-79921680 on Build GRCh38
rs927613838 Chr.6: 79921616-79921616 on Build GRCh38
rs776513027 Chr.6: 79921595-79921595 on Build GRCh38
rs753248002 Chr.6: 79921629-79921629 on Build GRCh38
rs564560356 Chr.6: 79921735-79921735 on Build GRCh38
rs773493523 Chr.6: 79921746-79921746 on Build GRCh38
rs587777598 Chr.6: 79921662-79921662 on Build GRCh38
rs956272516 Chr.6: 79921835-79921835 on Build GRCh38
rs373287811 Chr.6: 79921967-79921967 on Build GRCh38
rs772540649 Chr.6: 79921584-79921584 on Build GRCh38
rs781360903 Chr.6: 79921693-79921693 on Build GRCh38
rs993786858 Chr.6: 79921906-79921906 on Build GRCh38
rs1028928248 Chr.6: 79921786-79921786 on Build GRCh38
rs956550498 Chr.6: 79921986-79921986 on Build GRCh38
rs937892059 Chr.6: 79921931-79921931 on Build GRCh38
rs1056700547 Chr.6: 79921961-79921961 on Build GRCh38
rs758425030 Chr.6: 79921965-79921965 on Build GRCh38
rs371762104 Chr.6: 79921643-79921643 on Build GRCh38
rs190439121 Chr.6: 79921867-79921867 on Build GRCh38
rs1003921690 Chr.6: 79921569-79921569 on Build GRCh38
rs757975990 Chr.6: 79921861-79921864 on Build GRCh38
rs896429438 Chr.6: 79921994-79921994 on Build GRCh38
rs769960749 Chr.6: 79921603-79921603 on Build GRCh38

View Full Product Details