Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 43784285-43784835 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C17orf105
Entrez Gene ID
284067
Gene Name
chromosome 17 open reading frame 105
Gene Aliases
-
UniGene
Hs.252707
Gene Chromosome Location
Chr.17: 43778391-43784842 on Build GRCh38
Transcripts
NM_001136483.1 XM_011524633.1 XM_011524634.2 XM_011524635.1 XM_011524636.1

SNP Detail

SNP ID Chromosome Location
rs754478913 Chr.17: 43784461-43784461 on Build GRCh38
rs1038209776 Chr.17: 43784799-43784799 on Build GRCh38
rs141559382 Chr.17: 43784685-43784685 on Build GRCh38
rs74371596 Chr.17: 43784578-43784578 on Build GRCh38
rs577107912 Chr.17: 43784357-43784357 on Build GRCh38
rs779636779 Chr.17: 43784400-43784400 on Build GRCh38
rs973071488 Chr.17: 43784556-43784556 on Build GRCh38
rs769464839 Chr.17: 43784332-43784332 on Build GRCh38
rs1023072632 Chr.17: 43784602-43784602 on Build GRCh38
rs545544191 Chr.17: 43784403-43784403 on Build GRCh38
rs1032780509 Chr.17: 43784348-43784348 on Build GRCh38
rs983372590 Chr.17: 43784608-43784608 on Build GRCh38
rs1009735554 Chr.17: 43784792-43784792 on Build GRCh38
rs779284614 Chr.17: 43784511-43784511 on Build GRCh38
rs75436311 Chr.17: 43784705-43784705 on Build GRCh38
rs748898610 Chr.17: 43784433-43784433 on Build GRCh38
rs979008640 Chr.17: 43784376-43784376 on Build GRCh38
rs543014121 Chr.17: 43784512-43784512 on Build GRCh38
rs1001251253 Chr.17: 43784691-43784691 on Build GRCh38
rs905258351 Chr.17: 43784674-43784674 on Build GRCh38
rs527758132 Chr.17: 43784456-43784456 on Build GRCh38
rs1017298833 Chr.17: 43784522-43784522 on Build GRCh38
rs905311119 Chr.17: 43784321-43784321 on Build GRCh38
rs1042614993 Chr.17: 43784810-43784810 on Build GRCh38
rs896817416 Chr.17: 43784506-43784506 on Build GRCh38
rs535919927 Chr.17: 43784750-43784750 on Build GRCh38
rs897029937 Chr.17: 43784808-43784808 on Build GRCh38
rs7215012 Chr.17: 43784316-43784316 on Build GRCh38
rs970262624 Chr.17: 43784373-43784373 on Build GRCh38
rs73985105 Chr.17: 43784521-43784521 on Build GRCh38
rs746273719 Chr.17: 43784611-43784611 on Build GRCh38
rs992923191 Chr.17: 43784351-43784351 on Build GRCh38
rs978974193 Chr.17: 43784624-43784625 on Build GRCh38
rs1021044094 Chr.17: 43784473-43784473 on Build GRCh38
rs894032831 Chr.17: 43784414-43784414 on Build GRCh38
rs1049561151 Chr.17: 43784431-43784431 on Build GRCh38
rs993925881 Chr.17: 43784320-43784320 on Build GRCh38
rs916095695 Chr.17: 43784352-43784352 on Build GRCh38
rs555081425 Chr.17: 43784435-43784435 on Build GRCh38
rs550059798 Chr.17: 43784639-43784639 on Build GRCh38
rs137968559 Chr.17: 43784625-43784626 on Build GRCh38
rs964844565 Chr.17: 43784532-43784532 on Build GRCh38
rs565790204 Chr.17: 43784745-43784745 on Build GRCh38
rs560653340 Chr.17: 43784418-43784418 on Build GRCh38
rs574959045 Chr.17: 43784642-43784642 on Build GRCh38
rs755663729 Chr.17: 43784335-43784335 on Build GRCh38
rs762446575 Chr.17: 43784626-43784627 on Build GRCh38
rs748081458 Chr.17: 43784658-43784658 on Build GRCh38
rs929207342 Chr.17: 43784586-43784586 on Build GRCh38
rs934690899 Chr.17: 43784399-43784399 on Build GRCh38
rs922636267 Chr.17: 43784412-43784412 on Build GRCh38
rs960490733 Chr.17: 43784349-43784349 on Build GRCh38
rs941214873 Chr.17: 43784789-43784789 on Build GRCh38
rs1038330709 Chr.17: 43784496-43784496 on Build GRCh38
rs925965647 Chr.17: 43784386-43784386 on Build GRCh38
rs911044918 Chr.17: 43784762-43784762 on Build GRCh38
rs1044281042 Chr.17: 43784644-43784644 on Build GRCh38
rs949856479 Chr.17: 43784641-43784641 on Build GRCh38
rs117962974 Chr.17: 43784707-43784707 on Build GRCh38
rs1033717046 Chr.17: 43784625-43784628 on Build GRCh38

View Full Product Details