Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 159893082-159893541 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CFAP45
Entrez Gene ID
25790
Gene Name
cilia and flagella associated protein 45
Gene Aliases
CCDC19, NESG1
UniGene
Hs.647705
Gene Chromosome Location
Chr.1: 159872364-159900116 on Build GRCh38
Transcripts
NM_012337.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
386155 c.59G>A p.R20K Chr.1: 159893250-159893250 on Build GRCh38
3369369 c.41C>T p.A14V Chr.1: 159893268-159893268 on Build GRCh38
6274268 c.68C>A p.A23D Chr.1: 159893241-159893241 on Build GRCh38
898089 c.90G>A p.V30V Chr.1: 159893219-159893219 on Build GRCh38
6871239 c.129+1G>A p.? Chr.1: 159893179-159893179 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs774749136 Chr.1: 159893278-159893278 on Build GRCh38
rs753347555 Chr.1: 159893243-159893243 on Build GRCh38
rs754042755 Chr.1: 159893462-159893462 on Build GRCh38
rs914610939 Chr.1: 159893447-159893447 on Build GRCh38
rs531140424 Chr.1: 159893438-159893438 on Build GRCh38
rs139643283 Chr.1: 159893146-159893146 on Build GRCh38
rs766633847 Chr.1: 159893434-159893434 on Build GRCh38
rs770086170 Chr.1: 159893326-159893326 on Build GRCh38
rs781407013 Chr.1: 159893335-159893335 on Build GRCh38
rs765459098 Chr.1: 159893133-159893133 on Build GRCh38
rs368850939 Chr.1: 159893232-159893232 on Build GRCh38
rs374757532 Chr.1: 159893191-159893191 on Build GRCh38
rs755615157 Chr.1: 159893180-159893180 on Build GRCh38
rs969138087 Chr.1: 159893373-159893373 on Build GRCh38
rs748727893 Chr.1: 159893216-159893216 on Build GRCh38
rs780327689 Chr.1: 159893187-159893187 on Build GRCh38
rs143593292 Chr.1: 159893221-159893221 on Build GRCh38
rs761820002 Chr.1: 159893131-159893131 on Build GRCh38
rs959015602 Chr.1: 159893433-159893433 on Build GRCh38
rs924663407 Chr.1: 159893512-159893513 on Build GRCh38
rs951235247 Chr.1: 159893397-159893397 on Build GRCh38
rs761340494 Chr.1: 159893320-159893320 on Build GRCh38
rs747383126 Chr.1: 159893192-159893192 on Build GRCh38
rs778993928 Chr.1: 159893458-159893458 on Build GRCh38
rs764766344 Chr.1: 159893322-159893322 on Build GRCh38
rs764030288 Chr.1: 159893312-159893312 on Build GRCh38
rs372198026 Chr.1: 159893184-159893184 on Build GRCh38
rs777385725 Chr.1: 159893183-159893183 on Build GRCh38
rs563923181 Chr.1: 159893384-159893384 on Build GRCh38
rs969108794 Chr.1: 159893455-159893455 on Build GRCh38
rs775416924 Chr.1: 159893293-159893293 on Build GRCh38
rs914642342 Chr.1: 159893125-159893125 on Build GRCh38
rs756128446 Chr.1: 159893239-159893239 on Build GRCh38
rs756840313 Chr.1: 159893246-159893246 on Build GRCh38
rs1002632312 Chr.1: 159893407-159893407 on Build GRCh38
rs774177149 Chr.1: 159893379-159893379 on Build GRCh38
rs372096436 Chr.1: 159893145-159893145 on Build GRCh38
rs763586832 Chr.1: 159893242-159893242 on Build GRCh38
rs778554723 Chr.1: 159893250-159893250 on Build GRCh38
rs193248121 Chr.1: 159893400-159893400 on Build GRCh38
rs1020348914 Chr.1: 159893341-159893341 on Build GRCh38
rs183394355 Chr.1: 159893222-159893222 on Build GRCh38
rs781655391 Chr.1: 159893256-159893256 on Build GRCh38
rs368787534 Chr.1: 159893158-159893158 on Build GRCh38
rs752205865 Chr.1: 159893163-159893163 on Build GRCh38
rs549353733 Chr.1: 159893500-159893500 on Build GRCh38
rs144965039 Chr.1: 159893259-159893259 on Build GRCh38
rs750022611 Chr.1: 159893323-159893323 on Build GRCh38
rs771744125 Chr.1: 159893285-159893285 on Build GRCh38
rs117846150 Chr.1: 159893205-159893205 on Build GRCh38
rs927270424 Chr.1: 159893391-159893391 on Build GRCh38
rs544005243 Chr.1: 159893266-159893266 on Build GRCh38
rs758125513 Chr.1: 159893330-159893330 on Build GRCh38
rs766402815 Chr.1: 159893421-159893424 on Build GRCh38
rs868231888 Chr.1: 159893271-159893271 on Build GRCh38
rs774925941 Chr.1: 159893202-159893202 on Build GRCh38
rs753111503 Chr.1: 159893343-159893343 on Build GRCh38
rs770401300 Chr.1: 159893261-159893261 on Build GRCh38
rs756966978 Chr.1: 159893185-159893185 on Build GRCh38
rs188930388 Chr.1: 159893253-159893253 on Build GRCh38
rs151098186 Chr.1: 159893255-159893255 on Build GRCh38
rs368573056 Chr.1: 159893136-159893136 on Build GRCh38
rs930106635 Chr.1: 159893509-159893509 on Build GRCh38
rs769182800 Chr.1: 159893154-159893154 on Build GRCh38
rs917456377 Chr.1: 159893464-159893464 on Build GRCh38
rs977523501 Chr.1: 159893504-159893504 on Build GRCh38
rs145788106 Chr.1: 159893282-159893282 on Build GRCh38
rs537736471 Chr.1: 159893155-159893155 on Build GRCh38
rs760460810 Chr.1: 159893306-159893306 on Build GRCh38
rs774231764 Chr.1: 159893233-159893233 on Build GRCh38
rs760993149 Chr.1: 159893305-159893305 on Build GRCh38
rs41264831 Chr.1: 159893238-159893238 on Build GRCh38
rs763313639 Chr.1: 159893137-159893137 on Build GRCh38
rs771229914 Chr.1: 159893267-159893267 on Build GRCh38
rs979433506 Chr.1: 159893129-159893129 on Build GRCh38
rs956516906 Chr.1: 159893303-159893303 on Build GRCh38
rs769153821 Chr.1: 159893193-159893193 on Build GRCh38
rs1010362955 Chr.1: 159893333-159893333 on Build GRCh38
rs753291903 Chr.1: 159893319-159893319 on Build GRCh38

View Full Product Details