Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 99719092-99719639 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FRRS1
Entrez Gene ID
391059
Gene Name
ferric chelate reductase 1
Gene Aliases
SDFR2, SDR2
UniGene
Hs.454779
Gene Chromosome Location
Chr.1: 99708699-99766630 on Build GRCh38
Transcripts
NM_001013660.2 XM_005270861.3 XM_011541451.2 XM_011541452.1 XM_017001271.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
423312 c.1070C>T p.S357F Chr.1: 99719584-99719584 on Build GRCh38
106921 c.1108C>T p.L370L Chr.1: 99719546-99719546 on Build GRCh38
6667232 c.1059T>C p.D353D Chr.1: 99719595-99719595 on Build GRCh38
333606 c.1092C>T p.S364S Chr.1: 99719562-99719562 on Build GRCh38
6667228 c.1087G>A p.G363R Chr.1: 99719567-99719567 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs777050021 Chr.1: 99719496-99719496 on Build GRCh38
rs762551202 Chr.1: 99719602-99719602 on Build GRCh38
rs149824246 Chr.1: 99719546-99719546 on Build GRCh38
rs763217995 Chr.1: 99719616-99719617 on Build GRCh38
rs755935419 Chr.1: 99719543-99719543 on Build GRCh38
rs373262536 Chr.1: 99719504-99719504 on Build GRCh38
rs771601736 Chr.1: 99719484-99719484 on Build GRCh38
rs372874869 Chr.1: 99719607-99719607 on Build GRCh38
rs565561582 Chr.1: 99719192-99719192 on Build GRCh38
rs1040943026 Chr.1: 99719299-99719299 on Build GRCh38
rs902575508 Chr.1: 99719388-99719389 on Build GRCh38
rs141962615 Chr.1: 99719572-99719572 on Build GRCh38
rs773697373 Chr.1: 99719495-99719496 on Build GRCh38
rs777159956 Chr.1: 99719173-99719173 on Build GRCh38
rs922716211 Chr.1: 99719330-99719330 on Build GRCh38
rs781718565 Chr.1: 99719582-99719582 on Build GRCh38
rs751099688 Chr.1: 99719520-99719520 on Build GRCh38
rs984059087 Chr.1: 99719279-99719279 on Build GRCh38
rs775654581 Chr.1: 99719335-99719335 on Build GRCh38
rs970982426 Chr.1: 99719502-99719502 on Build GRCh38
rs911091414 Chr.1: 99719214-99719214 on Build GRCh38
rs918298230 Chr.1: 99719539-99719539 on Build GRCh38
rs1005791691 Chr.1: 99719612-99719612 on Build GRCh38
rs758720376 Chr.1: 99719560-99719560 on Build GRCh38
rs1003312813 Chr.1: 99719564-99719564 on Build GRCh38
rs763544515 Chr.1: 99719516-99719516 on Build GRCh38
rs910964583 Chr.1: 99719606-99719606 on Build GRCh38
rs961436514 Chr.1: 99719312-99719312 on Build GRCh38
rs1009228162 Chr.1: 99719131-99719131 on Build GRCh38
rs561039727 Chr.1: 99719211-99719211 on Build GRCh38
rs1018085483 Chr.1: 99719435-99719435 on Build GRCh38
rs752909038 Chr.1: 99719551-99719551 on Build GRCh38
rs972708152 Chr.1: 99719323-99719323 on Build GRCh38
rs1020755029 Chr.1: 99719140-99719140 on Build GRCh38
rs746362263 Chr.1: 99719591-99719591 on Build GRCh38
rs959244920 Chr.1: 99719589-99719589 on Build GRCh38
rs775138695 Chr.1: 99719500-99719500 on Build GRCh38
rs770135851 Chr.1: 99719599-99719599 on Build GRCh38
rs372962149 Chr.1: 99719407-99719408 on Build GRCh38
rs746328637 Chr.1: 99719200-99719200 on Build GRCh38
rs756801082 Chr.1: 99719526-99719526 on Build GRCh38
rs149401212 Chr.1: 99719122-99719122 on Build GRCh38
rs750386280 Chr.1: 99719536-99719536 on Build GRCh38
rs959444633 Chr.1: 99719235-99719235 on Build GRCh38
rs879118460 Chr.1: 99719430-99719430 on Build GRCh38
rs567685364 Chr.1: 99719334-99719334 on Build GRCh38
rs112379586 Chr.1: 99719540-99719540 on Build GRCh38
rs1032850966 Chr.1: 99719461-99719461 on Build GRCh38
rs189194590 Chr.1: 99719349-99719349 on Build GRCh38
rs747474273 Chr.1: 99719483-99719483 on Build GRCh38
rs775738312 Chr.1: 99719601-99719601 on Build GRCh38
rs35626078 Chr.1: 99719427-99719428 on Build GRCh38
rs191189274 Chr.1: 99719514-99719514 on Build GRCh38
rs911396663 Chr.1: 99719338-99719338 on Build GRCh38
rs565943384 Chr.1: 99719579-99719579 on Build GRCh38
rs545335920 Chr.1: 99719154-99719154 on Build GRCh38
rs767420721 Chr.1: 99719528-99719528 on Build GRCh38
rs543518422 Chr.1: 99719395-99719395 on Build GRCh38
rs1042438505 Chr.1: 99719371-99719371 on Build GRCh38
rs530108974 Chr.1: 99719340-99719340 on Build GRCh38
rs931101703 Chr.1: 99719160-99719160 on Build GRCh38
rs774039436 Chr.1: 99719608-99719608 on Build GRCh38
rs564501634 Chr.1: 99719302-99719302 on Build GRCh38
rs936133801 Chr.1: 99719346-99719346 on Build GRCh38
rs557090762 Chr.1: 99719408-99719408 on Build GRCh38
rs71854810 Chr.1: 99719408-99719409 on Build GRCh38
rs376478764 Chr.1: 99719563-99719563 on Build GRCh38
rs1009260562 Chr.1: 99719449-99719449 on Build GRCh38
rs114530804 Chr.1: 99719499-99719499 on Build GRCh38
rs909758037 Chr.1: 99719283-99719283 on Build GRCh38
rs779719762 Chr.1: 99719545-99719545 on Build GRCh38
rs959530287 Chr.1: 99719494-99719494 on Build GRCh38
rs1033526354 Chr.1: 99719233-99719233 on Build GRCh38

View Full Product Details