Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 155921264-155921779 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KIAA0907
Entrez Gene ID
22889
Gene Name
KIAA0907
Gene Aliases
BLOM7
UniGene
Hs.24656
Gene Chromosome Location
Chr.1: 155913043-155934442 on Build GRCh38
Transcripts
NM_014949.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6588387 c.1226A>G p.Y409C Chr.1: 155921415-155921415 on Build GRCh38
1559933 c.1150G>A p.V384M Chr.1: 155921491-155921491 on Build GRCh38
6262830 c.1085C>A p.S362Y Chr.1: 155921556-155921556 on Build GRCh38
6563421 c.1117_1120delCCAG p.P373fs*10 Chr.1: 155921521-155921524 on Build GRCh38
6246693 c.1013-6C>A p.? Chr.1: 155921634-155921634 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs576721677 Chr.1: 155921377-155921377 on Build GRCh38
rs79601516 Chr.1: 155921598-155921598 on Build GRCh38
rs79641570 Chr.1: 155921588-155921588 on Build GRCh38
rs377674825 Chr.1: 155921614-155921614 on Build GRCh38
rs773860096 Chr.1: 155921645-155921645 on Build GRCh38
rs745928808 Chr.1: 155921350-155921350 on Build GRCh38
rs199908872 Chr.1: 155921353-155921353 on Build GRCh38
rs748143416 Chr.1: 155921444-155921444 on Build GRCh38
rs909402391 Chr.1: 155921530-155921530 on Build GRCh38
rs758473808 Chr.1: 155921584-155921584 on Build GRCh38
rs1033789265 Chr.1: 155921657-155921657 on Build GRCh38
rs752477359 Chr.1: 155921568-155921568 on Build GRCh38
rs145643261 Chr.1: 155921489-155921489 on Build GRCh38
rs757979048 Chr.1: 155921396-155921396 on Build GRCh38
rs757698179 Chr.1: 155921454-155921454 on Build GRCh38
rs913336120 Chr.1: 155921708-155921708 on Build GRCh38
rs770228111 Chr.1: 155921633-155921633 on Build GRCh38
rs773843754 Chr.1: 155921506-155921506 on Build GRCh38
rs139448031 Chr.1: 155921609-155921609 on Build GRCh38
rs887007384 Chr.1: 155921317-155921317 on Build GRCh38
rs765921105 Chr.1: 155921509-155921509 on Build GRCh38
rs185232337 Chr.1: 155921347-155921347 on Build GRCh38
rs751289168 Chr.1: 155921525-155921525 on Build GRCh38
rs996597699 Chr.1: 155921434-155921434 on Build GRCh38
rs920655990 Chr.1: 155921700-155921700 on Build GRCh38
rs200520724 Chr.1: 155921351-155921351 on Build GRCh38
rs762185860 Chr.1: 155921358-155921358 on Build GRCh38
rs946159497 Chr.1: 155921720-155921720 on Build GRCh38
rs780813220 Chr.1: 155921432-155921432 on Build GRCh38
rs769912567 Chr.1: 155921697-155921698 on Build GRCh38
rs746011529 Chr.1: 155921453-155921453 on Build GRCh38
rs756058552 Chr.1: 155921451-155921451 on Build GRCh38
rs749161002 Chr.1: 155921582-155921582 on Build GRCh38
rs769096231 Chr.1: 155921482-155921482 on Build GRCh38
rs959772483 Chr.1: 155921406-155921406 on Build GRCh38
rs754807393 Chr.1: 155921419-155921419 on Build GRCh38
rs796796426 Chr.1: 155921686-155921686 on Build GRCh38
rs963607979 Chr.1: 155921533-155921533 on Build GRCh38
rs747970490 Chr.1: 155921341-155921341 on Build GRCh38
rs112789044 Chr.1: 155921352-155921352 on Build GRCh38
rs371195265 Chr.1: 155921650-155921650 on Build GRCh38
rs983443319 Chr.1: 155921739-155921739 on Build GRCh38
rs754542490 Chr.1: 155921564-155921564 on Build GRCh38
rs201266630 Chr.1: 155921507-155921507 on Build GRCh38
rs771078033 Chr.1: 155921343-155921343 on Build GRCh38
rs370075138 Chr.1: 155921678-155921678 on Build GRCh38
rs775698695 Chr.1: 155921459-155921459 on Build GRCh38
rs777742302 Chr.1: 155921580-155921580 on Build GRCh38
rs766194829 Chr.1: 155921411-155921411 on Build GRCh38
rs776812398 Chr.1: 155921484-155921484 on Build GRCh38
rs777660143 Chr.1: 155921452-155921452 on Build GRCh38
rs759059825 Chr.1: 155921667-155921667 on Build GRCh38
rs748520545 Chr.1: 155921632-155921632 on Build GRCh38
rs377382718 Chr.1: 155921671-155921671 on Build GRCh38
rs751660660 Chr.1: 155921307-155921307 on Build GRCh38
rs776577661 Chr.1: 155921443-155921443 on Build GRCh38
rs777198452 Chr.1: 155921354-155921354 on Build GRCh38
rs751199199 Chr.1: 155921412-155921412 on Build GRCh38
rs770591884 Chr.1: 155921654-155921654 on Build GRCh38
rs552289126 Chr.1: 155921435-155921435 on Build GRCh38
rs376340342 Chr.1: 155921659-155921659 on Build GRCh38
rs764823479 Chr.1: 155921363-155921363 on Build GRCh38
rs762213514 Chr.1: 155921495-155921495 on Build GRCh38
rs781202684 Chr.1: 155921325-155921325 on Build GRCh38
rs373051632 Chr.1: 155921467-155921467 on Build GRCh38
rs750000844 Chr.1: 155921388-155921388 on Build GRCh38
rs536125490 Chr.1: 155921715-155921715 on Build GRCh38
rs921890959 Chr.1: 155921653-155921653 on Build GRCh38
rs767180734 Chr.1: 155921565-155921565 on Build GRCh38
rs765702011 Chr.1: 155921504-155921504 on Build GRCh38
rs755929581 Chr.1: 155921575-155921575 on Build GRCh38
rs1025399739 Chr.1: 155921433-155921433 on Build GRCh38

View Full Product Details