Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 61976093-61976621 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARID5B
Entrez Gene ID
84159
Gene Name
AT-rich interaction domain 5B
Gene Aliases
DESRT, MRF-2, MRF2
UniGene
Hs.535297
Gene Chromosome Location
Chr.10: 61901254-62096948 on Build GRCh38
Transcripts
NM_032199.2 XM_011540262.1

SNP Detail

SNP ID Chromosome Location
rs1055181514 Chr.10: 61976291-61976291 on Build GRCh38
rs373537497 Chr.10: 61976552-61976552 on Build GRCh38
rs895224094 Chr.10: 61976300-61976300 on Build GRCh38
rs78811275 Chr.10: 61976545-61976545 on Build GRCh38
rs962650302 Chr.10: 61976275-61976275 on Build GRCh38
rs912716328 Chr.10: 61976523-61976523 on Build GRCh38
rs558102317 Chr.10: 61976547-61976547 on Build GRCh38
rs778665681 Chr.10: 61976482-61976482 on Build GRCh38
rs972710168 Chr.10: 61976549-61976549 on Build GRCh38
rs532781682 Chr.10: 61976290-61976290 on Build GRCh38
rs987649289 Chr.10: 61976507-61976507 on Build GRCh38
rs191392113 Chr.10: 61976120-61976120 on Build GRCh38
rs912759749 Chr.10: 61976214-61976214 on Build GRCh38
rs1032240765 Chr.10: 61976351-61976351 on Build GRCh38
rs940196829 Chr.10: 61976531-61976531 on Build GRCh38
rs1011101012 Chr.10: 61976119-61976119 on Build GRCh38
rs940267616 Chr.10: 61976215-61976215 on Build GRCh38
rs752747630 Chr.10: 61976227-61976227 on Build GRCh38
rs371248560 Chr.10: 61976305-61976305 on Build GRCh38
rs139035104 Chr.10: 61976495-61976495 on Build GRCh38
rs533857906 Chr.10: 61976336-61976336 on Build GRCh38
rs973042973 Chr.10: 61976285-61976285 on Build GRCh38
rs371975152 Chr.10: 61976365-61976365 on Build GRCh38
rs372454285 Chr.10: 61976428-61976428 on Build GRCh38
rs893956107 Chr.10: 61976427-61976427 on Build GRCh38
rs561867280 Chr.10: 61976562-61976562 on Build GRCh38
rs1043023957 Chr.10: 61976442-61976442 on Build GRCh38
rs966977704 Chr.10: 61976331-61976331 on Build GRCh38
rs932777287 Chr.10: 61976188-61976188 on Build GRCh38
rs368070235 Chr.10: 61976320-61976320 on Build GRCh38
rs931586866 Chr.10: 61976548-61976548 on Build GRCh38
rs902323411 Chr.10: 61976591-61976591 on Build GRCh38
rs186020816 Chr.10: 61976326-61976326 on Build GRCh38
rs117142812 Chr.10: 61976222-61976222 on Build GRCh38
rs953863711 Chr.10: 61976202-61976202 on Build GRCh38
rs535750972 Chr.10: 61976452-61976452 on Build GRCh38
rs573269700 Chr.10: 61976550-61976550 on Build GRCh38
rs967108757 Chr.10: 61976150-61976150 on Build GRCh38
rs866449908 Chr.10: 61976502-61976502 on Build GRCh38
rs79872566 Chr.10: 61976472-61976472 on Build GRCh38
rs76983655 Chr.10: 61976191-61976191 on Build GRCh38
rs867811973 Chr.10: 61976148-61976148 on Build GRCh38
rs568274585 Chr.10: 61976404-61976404 on Build GRCh38
rs998313724 Chr.10: 61976165-61976165 on Build GRCh38
rs981484059 Chr.10: 61976579-61976579 on Build GRCh38
rs1037262830 Chr.10: 61976541-61976541 on Build GRCh38
rs529095944 Chr.10: 61976563-61976563 on Build GRCh38
rs182806583 Chr.10: 61976246-61976246 on Build GRCh38
rs552106359 Chr.10: 61976322-61976322 on Build GRCh38
rs145151505 Chr.10: 61976401-61976401 on Build GRCh38

View Full Product Details