Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 88607996-88608495 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AEN
Entrez Gene ID
64782
Gene Name
apoptosis enhancing nuclease
Gene Aliases
ISG20L1, pp12744
UniGene
Hs.436102
Gene Chromosome Location
Chr.15: 88604601-88632281 on Build GRCh38
Transcripts
XM_011521905.2 XM_017022489.1 XM_017022490.1 NR_031590.1

Gene Symbol
MIR1179
Entrez Gene ID
100302235
Gene Name
microRNA 1179
Gene Aliases
MIRN1179, hsa-mir-1179, mir-1179
UniGene
Gene Chromosome Location
Chr.15: 88608107-88608197 on Build GRCh38
Transcripts
XM_011521905.2 XM_017022489.1 XM_017022490.1 NR_031590.1

SNP Detail

SNP ID Chromosome Location
rs537995249 Chr.15: 88608402-88608402 on Build GRCh38
rs564506697 Chr.15: 88608049-88608049 on Build GRCh38
rs781692757 Chr.15: 88608224-88608224 on Build GRCh38
rs747724065 Chr.15: 88608160-88608164 on Build GRCh38
rs144212662 Chr.15: 88608064-88608064 on Build GRCh38
rs573528749 Chr.15: 88608024-88608025 on Build GRCh38
rs905980658 Chr.15: 88608381-88608381 on Build GRCh38
rs752798578 Chr.15: 88608065-88608065 on Build GRCh38
rs747520284 Chr.15: 88608116-88608116 on Build GRCh38
rs746265975 Chr.15: 88608134-88608134 on Build GRCh38
rs181571116 Chr.15: 88608377-88608377 on Build GRCh38
rs753496113 Chr.15: 88608097-88608097 on Build GRCh38
rs760714694 Chr.15: 88608176-88608176 on Build GRCh38
rs757905930 Chr.15: 88608113-88608113 on Build GRCh38
rs115724715 Chr.15: 88608415-88608415 on Build GRCh38
rs994440653 Chr.15: 88608400-88608400 on Build GRCh38
rs370629468 Chr.15: 88608182-88608182 on Build GRCh38
rs770110851 Chr.15: 88608245-88608245 on Build GRCh38
rs547779539 Chr.15: 88608205-88608205 on Build GRCh38
rs548720252 Chr.15: 88608382-88608382 on Build GRCh38
rs923179004 Chr.15: 88608298-88608298 on Build GRCh38
rs938817649 Chr.15: 88608387-88608387 on Build GRCh38
rs746238258 Chr.15: 88608230-88608230 on Build GRCh38
rs755910296 Chr.15: 88608080-88608080 on Build GRCh38
rs764278845 Chr.15: 88608174-88608174 on Build GRCh38
rs191321810 Chr.15: 88608404-88608404 on Build GRCh38
rs975923049 Chr.15: 88608267-88608267 on Build GRCh38
rs750440236 Chr.15: 88608079-88608079 on Build GRCh38
rs902368011 Chr.15: 88608213-88608213 on Build GRCh38
rs922258150 Chr.15: 88608051-88608051 on Build GRCh38
rs2350271 Chr.15: 88608031-88608031 on Build GRCh38
rs887456318 Chr.15: 88608420-88608420 on Build GRCh38
rs566795777 Chr.15: 88608367-88608367 on Build GRCh38
rs865926845 Chr.15: 88608144-88608144 on Build GRCh38
rs529316535 Chr.15: 88608180-88608180 on Build GRCh38
rs527744100 Chr.15: 88608120-88608120 on Build GRCh38
rs989195223 Chr.15: 88608348-88608348 on Build GRCh38
rs754639756 Chr.15: 88608105-88608105 on Build GRCh38
rs186623237 Chr.15: 88608388-88608388 on Build GRCh38
rs747465400 Chr.15: 88608220-88608220 on Build GRCh38
rs145052991 Chr.15: 88608390-88608390 on Build GRCh38
rs762981313 Chr.15: 88608066-88608066 on Build GRCh38
rs747691183 Chr.15: 88608112-88608112 on Build GRCh38
rs757739288 Chr.15: 88608221-88608221 on Build GRCh38
rs568494694 Chr.15: 88608123-88608123 on Build GRCh38
rs752307575 Chr.15: 88608198-88608198 on Build GRCh38
rs770025260 Chr.15: 88608136-88608136 on Build GRCh38
rs759834312 Chr.15: 88608057-88608057 on Build GRCh38
rs764931793 Chr.15: 88608197-88608197 on Build GRCh38
rs569000338 Chr.15: 88608024-88608024 on Build GRCh38
rs374549291 Chr.15: 88608195-88608195 on Build GRCh38
rs763072220 Chr.15: 88608157-88608157 on Build GRCh38
rs371419981 Chr.15: 88608208-88608208 on Build GRCh38
rs914586174 Chr.15: 88608352-88608352 on Build GRCh38
rs778195038 Chr.15: 88608114-88608114 on Build GRCh38
rs375375042 Chr.15: 88608169-88608169 on Build GRCh38
rs947419467 Chr.15: 88608353-88608353 on Build GRCh38
rs753641519 Chr.15: 88608192-88608192 on Build GRCh38
rs780029537 Chr.15: 88608084-88608084 on Build GRCh38
rs983652624 Chr.15: 88608023-88608024 on Build GRCh38
rs939437555 Chr.15: 88608216-88608216 on Build GRCh38
rs767554416 Chr.15: 88608067-88608067 on Build GRCh38
rs541445452 Chr.15: 88608441-88608441 on Build GRCh38
rs1044495189 Chr.15: 88608366-88608366 on Build GRCh38
rs966647102 Chr.15: 88608451-88608451 on Build GRCh38
rs368561451 Chr.15: 88608202-88608202 on Build GRCh38
rs932342577 Chr.15: 88608052-88608052 on Build GRCh38
rs560494434 Chr.15: 88608023-88608023 on Build GRCh38
rs1012211561 Chr.15: 88608341-88608341 on Build GRCh38
rs964514341 Chr.15: 88608223-88608223 on Build GRCh38
rs775638383 Chr.15: 88608145-88608145 on Build GRCh38
rs753533400 Chr.15: 88608231-88608231 on Build GRCh38
rs1056507136 Chr.15: 88608287-88608287 on Build GRCh38
rs934619407 Chr.15: 88608331-88608331 on Build GRCh38

View Full Product Details